1 - 30 of 30 products displayed
  • Mouse KC for Western Blot, IP, ELISA and Ctrl
  • Host Yeast
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # RMKCI
  • Size 20 ug
Applications
Control (Assay Dependent), ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Secretory protein N51; Platelet-derived growth factor-inducible protein KC; C-X-C motif chemokine 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
RMKCI 300.00
Check your price
  • CCL20 (mouse) for Ctrl
  • Host E. coli
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # RP8673
  • Size 20 µg
Applications
Control (Assay Dependent)
Alternate names:
Small-inducible cytokine A20; Macrophage inflammatory protein 3 alpha; Liver and activation-regulated chemokine; CC chemokine ST38; CC chemokine LARC; Beta-chemokine exodus-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP8673 275.00
Check your price
  • IP10 (mouse) for Ctrl
  • Host E. coli
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # RP8668
  • Size 25 µg
Applications
Control (Assay Dependent)
Alternate names:
Small-inducible cytokine B10; Interferon-gamma induced protein CRG-2; C7; 10 kDa interferon gamma-induced protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP8668 309.00
Check your price
  • CCL19 (mouse) for Ctrl
  • Host E. Coli
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # RP8669
  • Size 20 µg
Applications
Control (Assay Dependent)
Alternate names:
Small-inducible cytokine A19; Epstein-Barr virus-induced molecule 1 ligand chemokine
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP8669 275.00
Check your price
  • CXCL12 Polyclonal Antibody for Western Blot, IHC and IP
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129029
  • Size 100 µg
Applications
Immunohistochemistry (1:100-1:500), Immunoprecipitation (1:300-1:500), Western Blot (1:1000)
Alternate names:
Thymic lymphoma cell-stimulating factor; Pre-B cell growth-stimulating factor; C-X-C motif chemokine 12; 12-O-tetradecanoylphorbol 13-acetate repressed protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129029 349.00
Check your price
  • CCR2 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA127409
  • Size 100 µg
Applications
Flow Cytometry (1:40), Immunocytochemistry (1:400), Immunofluorescence (1:400), Immunohistochemistry (Paraffin) (1:250), Western Blot (Assay Dependent)
Alternate names:
Monocyte chemoattractant protein 1 receptor; MCP-1 receptor; JE/FIC receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127409 475.00
Check your price
  • ST2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520077
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Protein ST2; Protein T1; Lymphocyte antigen 84
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520077 349.00
Check your price
  • CXCR3 Polyclonal Antibody for Flow and ELISA
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 718800
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent)
Alternate names:
Interferon-inducible protein 10 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
718800 309.00
Check your price
  • CX3CL1 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA1031X
  • Size 20 µL
Applications
ELISA (Assay dependent), Western Blot (1:1000)
Alternate names:
Small-inducible cytokine D1; Neurotactin; CX3C membrane-anchored chemokine; C-X3-C motif chemokine 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1031X 119.00
Check your price
  • CX3CL1 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA1031
  • Size 100 µg
Applications
ELISA (Assay dependent), Western Blot (1:1000)
Alternate names:
Small-inducible cytokine D1; Neurotactin; CX3C membrane-anchored chemokine; C-X3-C motif chemokine 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1031 349.00
Check your price
  • CCR6 Monoclonal Antibody for IHC (F) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120156
  • Size 100 µg
Applications
Flow Cytometry (5-10 µg for 10^5 cells in 100 µl), Immunohistochemistry (Frozen) (Assay dependent)
Alternate names:
LARC receptor; GPR-CY4; G-protein coupled receptor 29; DRY6; Chemokine receptor-like 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120156 349.00
Check your price
  • CXCR2 Monoclonal Antibody for Flow and IA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA124669
  • Size 100 µg
Applications
Flow Cytometry (10 µg for 10^5 PBMC), Inhibition Assays (0.5-1.5 µg/ml)
Alternate names:
IL-8 receptor type 2; High affinity interleukin-8 receptor B; GRO/MGSA receptor; CDw128b
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124669 309.00
Check your price
  • CXCR1 Monoclonal Antibody for Flow and IA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA124668
  • Size 100 µg
Applications
Flow Cytometry (10 µl for 10^6 PBMC), Inhibition Assays (0.1-0.3 µg/ml)
Alternate names:
IL-8 receptor type 1; High affinity interleukin-8 receptor A; CDw128a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124668 375.00
Check your price
  • CCR2 Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120155
  • Size 100 µg
Applications
Flow Cytometry (5 µg for 10^5 cells), Western Blot (Assay-dependent)
Alternate names:
Monocyte chemoattractant protein 1 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120155 349.00
Check your price
  • CCR5 Monoclonal Antibody for IHC (P), Flow and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA124666
  • Size 100 µg
Applications
Flow Cytometry (5-10 µg/ml), Immunohistochemistry (Paraffin) (5 µg/ml), Neutralization (Assay-dependent)
Alternate names:
HIV-1 fusion coreceptor; CHEMR13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124666 349.00
Check your price
  • CXCL1 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523745
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.05-0.25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Secretory protein N51; Platelet-derived growth factor-inducible protein KC; C-X-C motif chemokine 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523745 349.00
Check your price
  • CCRD6 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA511867
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Chemokine-binding protein D6; Chemokine-binding protein 2; Chemokine receptor CCR-9; Chemokine receptor CCR-10; C-C chemokine receptor D6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511867 349.00
Check your price
  • CCRD6 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA511866
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Chemokine-binding protein D6; Chemokine-binding protein 2; Chemokine receptor CCR-9; Chemokine receptor CCR-10; C-C chemokine receptor D6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511866 349.00
Check your price
  • MCP-3 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546952
  • Size 100 µg
Applications
Neutralization (15-45 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small-inducible cytokine A7; Protein FIC; Monocyte chemotactic protein 3; Monocyte chemoattractant protein 3; Intercrine/chemokine MARC
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546952 375.00
Check your price
  • CCRD6 Polyclonal Antibody for IF, ICC, IHC (P), Flow and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA121612
  • Size 100 µg
Applications
ELISA (1:100,000), Flow Cytometry (1-5 µg/ml), Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (Paraffin) (1:250)
Alternate names:
Chemokine-binding protein D6; Chemokine-binding protein 2; C-C chemokine receptor D6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121612 399.00
Check your price
  • Eotaxin Polyclonal Antibody for Western Blot, IHC (F), ELISA and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546946
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunohistochemistry (Frozen) (5-15 µg/ml), Neutralization (0.1-0.5 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small-inducible cytokine A11; Eosinophil chemotactic protein; C-C motif chemokine 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546946 375.00
Check your price
  • CXCL13 Polyclonal Antibody for Western Blot, IHC (F) and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547018
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Neutralization (20-50 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small-inducible cytokine B13; CXC chemokine BLC; B lymphocyte chemoattractant
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547018 375.00
Check your price
  • CXCL13 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA129046
  • Size 100 µg
Applications
Western Blot (1:5000)
Alternate names:
Small-inducible cytokine B13; CXC chemokine BLC; B lymphocyte chemoattractant
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129046 375.00
Check your price
  • CCR9 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA121618
  • Size 100 µg
Applications
ELISA (1:100,000), Flow Cytometry (1:200), Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (Paraffin) (1:250), Western Blot (1:1000)
Alternate names:
Chemokine C-C receptor 10; GPR-9-6; G-protein coupled receptor 28
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121618 425.00
Check your price
  • CCR10 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA121617
  • Size 100 µg
Applications
ELISA (1:100,000), Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (Paraffin) (1:250), Western Blot (1:1000)
Alternate names:
G-protein coupled receptor 2; Chemokine C-C receptor 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121617 475.00
Check your price
  • CCL21 Monoclonal Antibody for Western Blot, IHC (F), Flow and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523755
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Frozen) (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
6Ckine, CKb9, ECL, SCYA21, SLC, TCA4, C-C motif chemokine 21, Efficient Chemoattractant for Lymphocytes, beta chemokine exodus-2, beta-chemokine exodus-2, exodus-2, secondary lymphoid tissue chemokine, secondary lymphoid-tissue chemokine, small inducible cytokine subfamily A (Cys-Cys), member 21, sma
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523755 349.00
Check your price
  • MCP-1 Polyclonal Antibody for Western Blot, ELISA and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546954
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Neutralization (0.5-2.5 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small-inducible cytokine A2; Platelet-derived growth factor-inducible protein JE; Monocyte chemotactic protein 1; Monocyte chemoattractant protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546954 375.00
Check your price
  • CXCL16 Polyclonal Antibody for Western Blot, Flow and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547688
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Neutralization (0.1-0.4 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Transmembrane chemokine CXCL16; Small-inducible cytokine B16; Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547688 349.00
Check your price
  • CXCR4 Polyclonal Antibody for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA121625
  • Size 100 µg
Applications
ELISA (1:50,000), Immunohistochemistry (1:300), Western Blot (1:1000)
Alternate names:
Stromal cell-derived factor 1 receptor; Pre-B-cell-derived chemokine receptor; Leukocyte-derived seven transmembrane domain receptor; Fusin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121625 475.00
Check your price
  • CXCR7 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA3069
  • Size 100 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Western Blot (1:4000-1:6000 )
Alternate names:
G-protein coupled receptor RDC1 homolog; Chemokine orphan receptor 1; C-X-C chemokine receptor type 7; G-protein coupled receptor 159
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3069 349.00
Check your price