1 - 6 of 6 products displayed
  • Myosin Skeletal Muscle Antibody Monoclonal (MY-32) (MA1-82136)
  • Host Mouse
  • Target Species Mouse, Rat, Non-human primate, Dog, Human, Rabbit
  • Conjugate
  • Catalog # MA182136
  • Size 100µL
Applications
Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (1:10)
Alternate names:
Myosin heavy chain, skeletal muscle, adult 1; Myosin heavy chain 2x; Myosin heavy chain 1; Myosin heavy chain IIx/d
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182136 189.00
Check your price
  • Myosin Skeletal Muscle Antibody (Monoclonal, 414)
  • Host Mouse
  • Target Species Mammal
  • Conjugate Unconjugated
  • Catalog # MA135718
  • Size 1 mL
Applications
Immunohistochemistry (Frozen) (1:5-1:10), Immunohistochemistry (Paraffin) (1:5-1:10), Western Blot (Assay Dependent)
Alternate names:
Myosin heavy chain, skeletal muscle, adult 1; Myosin heavy chain 2x; Myosin heavy chain 1; Myosin heavy chain IIx/d
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135718 399.00
Check your price
  • Myosin Skeletal/Smooth Muscle Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA128038
  • Size 100 µL
Applications
Immunocytochemistry (1:10), Immunofluorescence (1:10)
Alternate names:
Myosin heavy chain, skeletal muscle, adult 1; Myosin heavy chain 2x; Myosin heavy chain 1; Myosin heavy chain IIx/d; SMMHC; Myosin heavy chain, smooth muscle isoform; Myosin heavy chain 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128038 309.00
Check your price
  • Myosin Skeletal Muscle Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA531466
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Myosin heavy chain, skeletal muscle, adult 1; Myosin heavy chain 2x; Myosin heavy chain 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531466 349.00
Check your price
  • Myosin Skeletal Muscle Antibody (Monoclonal, MYSN02 (MY-32))
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511748
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:500-1:1000)
Alternate names:
Myosin heavy chain, skeletal muscle, adult 1; Myosin heavy chain IIx/d; Myosin heavy chain 2x; Myosin heavy chain 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511748 309.00
Check your price
  • Myosin Skeletal Muscle Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA128037
  • Size 100 µL
Applications
Immunocytochemistry (1:20), Immunofluorescence (1:20)
Alternate names:
Myosin heavy chain, skeletal muscle, adult 2; Myosin heavy chain IIa; Myosin heavy chain 2a; Myosin heavy chain 2; Myosin heavy chain, skeletal muscle, adult 1; Myosin heavy chain IIx/d; Myosin heavy chain 2x; Myosin heavy chain 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128037 309.00
Check your price