1 - 30 of 30 products displayed
  • GATA1 Antibody (Oligoclonal, 4HCLC)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710049
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Western Blot (1-5 ug/ml)
Alternate names:
NF-E1 DNA-binding protein; GATA-binding factor 1; Eryf1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710049 309.00
Check your price
  • GATA1 Antibody (Monoclonal, 4H15L5)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 700727
  • Size 100 µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
NF-E1 DNA-binding protein; GATA-binding factor 1; Eryf1
Catalog # Size Price (AUD) Your Price (AUD) Qty
700727 304.00
Check your price
  • GATA1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1099X
  • Size 20 µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Western Blot (1:500)
Alternate names:
NF-E1 DNA-binding protein; GATA-binding factor 1; Eryf1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1099X 119.00
Check your price
  • GATA1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1099
  • Size 100 µg
Applications
Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (1:50-1:500), Western Blot (1:500-1:1000)
Alternate names:
NF-E1 DNA-binding protein; GATA-binding factor 1; Eryf1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1099 349.00
Check your price
  • Phospho-GATA1 (Ser142) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536856
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
NF-E1 DNA-binding protein; GATA-binding factor 1; Eryf1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536856 349.00
Check your price
  • GATA1 Antibody (Monoclonal, 4F5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515638
  • Size 100 µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
NF-E1 DNA-binding protein; GATA-binding factor 1; Eryf1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515638 349.00
Check your price
  • Phospho-GATA1 (Ser142) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537581
  • Size 100 µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
NF-E1 DNA-binding protein; GATA-binding factor 1; Eryf1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537581 309.00
Check your price
  • Phospho-GATA1 (Ser310) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537582
  • Size 100 µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Western Blot (1:500-1:1000)
Alternate names:
NF-E1 DNA-binding protein; GATA-binding factor 1; Eryf1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537582 309.00
Check your price
  • GATA1 Antibody (Monoclonal, 4G1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515637
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
NF-E1 DNA-binding protein; GATA-binding factor 1; Eryf1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515637 349.00
Check your price
  • GATA1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518408
  • Size 100 ug
Applications
Western Blot (0.3-1.0 ug/ml)
Alternate names:
NF-E1 DNA-binding protein; GATA-binding factor 1; Eryf1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518408 349.00
Check your price
  • YY1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA529171
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Yin and yang 1; NF-E1; INO80 complex subunit S; Delta transcription factor; UCR-motif DNA-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529171 349.00
Check your price
  • YY1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA512206
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Yin and yang 1; NF-E1; INO80 complex subunit S; Delta transcription factor; UCR-motif DNA-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512206 349.00
Check your price
  • YY1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529138
  • Size 100 µL
Applications
Immunoprecipitation (1:100-1:500), Western Blot (1:500-1:3000)
Alternate names:
Yin and yang 1; NF-E1; INO80 complex subunit S; Delta transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529138 349.00
Check your price
  • YY1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519730
  • Size 100 µg
Applications
Immunofluorescence (5 µg/ml), Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (5 µg/ml)
Alternate names:
Yin and yang 1; NF-E1; INO80 complex subunit S; Delta transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519730 349.00
Check your price
  • YY1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512205
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Yin and yang 1; NF-E1; INO80 complex subunit S; Delta transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512205 349.00
Check your price
  • Phospho-beta Catenin (Ser86) Antibody (Monoclonal, 24 E1)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44210M
  • Size 50µL
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
44210M 309.00
Check your price
  • UBE2V1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA112841
  • Size 100 µg
Applications
Western Blot (Assay Dependent)
Alternate names:
CROC-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112841 309.00
Check your price
  • Estrogen Receptor alpha Antibody (Monoclonal, 33)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA1310
  • Size 100 µg
Applications
Immunofluorescence (10 µg/ml), Immunohistochemistry (1:20-1:100), Western Blot (4 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1310 375.00
Check your price
  • Estrogen Receptor alpha Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Sheep, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA516476
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (10-20 µg/mL), Immunoprecipitation (10 µg/ml), Western Blot (2.5-5.0 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516476 349.00
Check your price
  • Estrogen Receptor alpha Antibody
  • Host Rabbit
  • Target Species Hamster, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA516440
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:250-500), Immunoprecipitation (10 µg/ml), Western Blot (2-5µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516440 349.00
Check your price
  • Estrogen Receptor alpha Antibody (Monoclonal, 6F11)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA180216
  • Size 100 µL
Applications
Immunohistochemistry (Frozen) (1:40-1:60), Immunohistochemistry (Paraffin) (1:40-1:80), Immunoprecipitation (10 µl/mg lysate), Western Blot (1:50-1:100)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180216 159.00
Check your price
  • Estrogen Receptor alpha Antibody (Monoclonal, 1D5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513191
  • Size 500 µL
Applications
Immunocytochemistry (1:500-1:1000), Immunofluorescence (1:500-1:1000), Immunohistochemistry (Paraffin) (0.4-0.8 µg/mL), Western Blot (0.5-1.0 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513191 550.00
Check your price
  • UBC13 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529687
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Ubiquitin-protein ligase N; Ubiquitin carrier protein N; Ubc13; E2 ubiquitin-conjugating enzyme N; Bendless-like ubiquitin-conjugating enzyme; UbcH13
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529687 349.00
Check your price
  • UBC9 Antibody (Monoclonal, 67AT1273.95.90)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA511149
  • Size 400 µL
Applications
Western Blot (1:100-1:500)
Alternate names:
p18; Ubiquitin-protein ligase I; Ubiquitin-conjugating enzyme E2 I; Ubiquitin carrier protein I; Ubiquitin carrier protein 9; SUMO-protein ligase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511149 349.00
Check your price
  • Estrogen Receptor alpha Antibody (Cocktail, 1D5 + 6F11)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA514104
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:100-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514104 550.00
Check your price
  • Estrogen Receptor alpha Antibody (Monoclonal, 6F11)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA513304
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin, Frozen) (1:40-1:80), Immunoprecipitation (10 µl/mg lysate), Western Blot (1:100-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513304 425.00
Check your price
  • Estrogen Receptor alpha Antibody (Monoclonal, 6F11)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA127107
  • Size 250 µL
Applications
Immunohistochemistry (Frozen) (1:40-1:80), Immunohistochemistry (Paraffin) (1:40-1:80), Immunoprecipitation (10 µl/mg lysate), Western Blot (1:50-1:100)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA127107 425.00
Check your price
  • UBC13 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517010
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Ubiquitin-protein ligase N; Ubiquitin carrier protein N; E2 ubiquitin-conjugating enzyme N; Bendless-like ubiquitin-conjugating enzyme; Ubc13; UbcH13
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517010 275.00
Check your price
  • UBE2L3 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517234
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Ubiquitin-protein ligase L3; Ubiquitin-conjugating enzyme E2-F1; Ubiquitin carrier protein L3; UbcH7; L-UBC; E2 ubiquitin-conjugating enzyme L3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517234 275.00
Check your price
  • UBC9 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 383000
  • Size 100 µg
Applications
Western Blot (Assay Dependent)
Alternate names:
p18; Ubiquitin-protein ligase I; Ubiquitin-conjugating enzyme E2 I; Ubiquitin carrier protein I; Ubiquitin carrier protein 9; SUMO-protein ligase
Catalog # Size Price (AUD) Your Price (AUD) Qty
383000 309.00
Check your price