1 - 20 of 20 products displayed
  • NFATC1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141041
  • Size 100µg
Applications
Immunocytochemistry (1:10), Immunofluorescence (1:10), Western Blot (1-3 µg/ml)
Alternate names:
NFAT transcription complex cytosolic component
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141041 349.00
Check your price
  • NFATC1 Polyclonal Antibody for Western Blot, IHC and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA123423
  • Size 100µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Assay-Dependent), Western Blot (1:500-1:1000)
Alternate names:
NFAT transcription complex cytosolic component
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123423 375.00
Check your price
  • NFATC1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14195
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
NFAT transcription complex cytosolic component
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14195 349.00
Check your price
  • NFATC1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14196
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
NFAT transcription complex cytosolic component
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14196 309.00
Check your price
  • NFATC1 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), IHC (PFA), IP, ChIP and GS
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA3024
  • Size 100µL
Applications
ChIP assay (Assay dependent), Gel Shift (Assay dependent), Immunocytochemistry (1:250), Immunofluorescence (1:10-1:100), Immunohistochemistry (1:20-200), Immunohistochemistry (Paraffin) (Assay Dependent), Immunohistochemistry (PFA fixed) (Assay Dependent), Immunoprecipitation (Assay dependent), Western Blo
Alternate names:
NFAT transcription complex cytosolic component
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3024 375.00
Check your price
  • Phospho-NFATC1 (Ser294) Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538301
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
NFAT transcription complex cytosolic component
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538301 309.00
Check your price
  • NFAT5 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), IP and GS
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1023
  • Size 50µg
Applications
Gel Shift (Assay dependent), Immunocytochemistry (1:1,000), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:20), Immunoprecipitation (3 µg), Western Blot (1:1,000)
Alternate names:
Tonicity-responsive enhancer-binding protein; T-cell transcription factor NFAT5; Rel domain-containing transcription factor NFAT5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1023 375.00
Check your price
  • TRPM4 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534283
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml)
Alternate names:
Melastatin-4; Long transient receptor potential channel 4; Calcium-activated non-selective cation channel 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534283 349.00
Check your price
  • TRPM4 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534285
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (5-7.5 µg/ml)
Alternate names:
Melastatin-4; Long transient receptor potential channel 4; Calcium-activated non-selective cation channel 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534285 349.00
Check your price
  • TRPM4 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534284
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
Melastatin-4; Long transient receptor potential channel 4; Calcium-activated non-selective cation channel 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534284 349.00
Check your price
  • NFATC2 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P), IP, ChIP and GS
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1025
  • Size 100µg
Applications
ChIP assay (1-3 ul), Gel Shift (Assay dependent), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin, Frozen) (1:50-1:200), Immunoprecipitation (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
T-cell transcription factor NFAT1; NFAT pre-existing subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1025 375.00
Check your price
  • NFATC4 Polyclonal Antibody for Western Blot, IF, ICC, IHC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1021
  • Size 100µg
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (1:3000), Immunoprecipitation (3 µg), Western Blot (1:1000)
Alternate names:
T-cell transcription factor NFAT3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1021 349.00
Check your price
  • Phospho-IkB epsilon (Ser18, Ser22) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517532
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
I-kappa-B-epsilon
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517532 309.00
Check your price
  • Phospho-NFAT5 (Ser1197) Polyclonal Antibody for IF, ICC and pep-ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA538884
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), peptide-ELISA (1:1000)
Alternate names:
Tonicity-responsive enhancer-binding protein; T-cell transcription factor NFAT5; Rel domain-containing transcription factor NFAT5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538884 309.00
Check your price
  • Phospho-NFAT5 (Ser155) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539799
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Tonicity-responsive enhancer-binding protein; T-cell transcription factor NFAT5; Rel domain-containing transcription factor NFAT5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539799 309.00
Check your price
  • NFAT5 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538885
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Tonicity-responsive enhancer-binding protein; T-cell transcription factor NFAT5; Rel domain-containing transcription factor NFAT5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538885 309.00
Check your price
  • Phospho-NFATC2 (Ser54) Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44944G
  • Size 10blots
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Western Blot (Assay Dependent)
Alternate names:
T-cell transcription factor NFAT1; NFAT pre-existing subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
44944G 305.00
Check your price
  • Phospho-NFATC4 (Ser168, Ser170) Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539798
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
T-cell transcription factor NFAT3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539798 309.00
Check your price
  • Phospho-NFATC4 (Ser676) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538302
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
T-cell transcription factor NFAT3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538302 309.00
Check your price
  • TRPM4 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OSR00134W
  • Size 100µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Melastatin-4; Long transient receptor potential channel 4; Calcium-activated non-selective cation channel 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00134W 309.00
Check your price