1 - 22 of 22 products displayed
  • NGAL Monoclonal Antibody, Biotin conjugate for ELISA
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Biotin
  • Catalog # HYB21102B005
  • Size 50µl
Applications
ELISA (1:16000)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB21102B005 425.00
Check your price
  • NGAL Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Dog, Human, Pig
  • Conjugate Unconjugated
  • Catalog # ABS0381402
  • Size 200µL
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0381402 650.00
Check your price
  • NGAL Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # ABS0381502
  • Size 200µL
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0381502 650.00
Check your price
  • NGAL Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # ABS0623302
  • Size 200µL
Applications
ELISA (Assay Dependent)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0623302 650.00
Check your price
  • NGAL Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # ABS0621402
  • Size 200µL
Applications
ELISA (Assay Dependent)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0621402 650.00
Check your price
  • NGAL Monoclonal Antibody for Western Blot, IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB2110202
  • Size 200µl
Applications
ELISA (1:8,000), Immunohistochemistry (Assay dependent), Western Blot (1:50)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB2110202 650.00
Check your price
  • NGAL Monoclonal Antibody for Western Blot, IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB2110102
  • Size 200µl
Applications
ELISA (Assay dependent), Immunohistochemistry (Assay dependent), Western Blot (Assay dependent)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB2110102 650.00
Check your price
  • NGAL Monoclonal Antibody for ELISA and IB
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # ABS0380402
  • Size 200µL
Applications
ELISA (Assay Dependent), Immunoblot (Assay Dependent)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0380402 650.00
Check your price
  • NGAL Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # ABS0382602
  • Size 200µL
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0382602 650.00
Check your price
  • NGAL Monoclonal Antibody, Biotin conjugate for ELISA
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # ABS03932B005
  • Size 50µL
Applications
ELISA (1:4000)
Alternate names:
p25; Siderocalin LCN2; Lipocalin-2; Alpha-2U globulin-related protein; Alpha-2-microglobulin-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS03932B005 475.00
Check your price
  • NGAL Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # ABS0391502
  • Size 200µL
Applications
ELISA (Assay Dependent)
Alternate names:
p25; Siderocalin LCN2; Lipocalin-2; Alpha-2U globulin-related protein; Alpha-2-microglobulin-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0391502 650.00
Check your price
  • NGAL Monoclonal Antibody for Western Blot, IF and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # ABS0432902
  • Size 200µL
Applications
ELISA (1:4000), Immunofluorescence (1:1000), Western Blot (Assay dependent)
Alternate names:
p25; Siderocalin LCN2; SV-40-induced 24P3 protein; Lipocalin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0432902 600.00
Check your price
  • NGAL Monoclonal Antibody for Western Blot and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # ABS0431002
  • Size 200µL
Applications
ELISA (1:2000), Western Blot (Assay dependent)
Alternate names:
p25; Siderocalin LCN2; SV-40-induced 24P3 protein; Lipocalin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0431002 650.00
Check your price
  • NGAL Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546916
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.25-5 µg/ml)
Alternate names:
p25; Siderocalin LCN2; SV-40-induced 24P3 protein; Lipocalin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546916 375.00
Check your price
  • NGAL Polyclonal Antibody for Western Blot, IP and ELISA
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA546938
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
p25; Siderocalin LCN2; Lipocalin-2; Alpha-2U globulin-related protein; Alpha-2-microglobulin-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546938 375.00
Check your price
  • GLP1R Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Mammal
  • Conjugate Unconjugated
  • Catalog # ABS0460302
  • Size 200µL
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p25; Siderocalin LCN2; SV-40-induced 24P3 protein; Lipocalin-2; Alpha-2U globulin-related protein; Alpha-2-microglobulin-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0460302 650.00
Check your price
  • NGAL Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA518861
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
p25; Siderocalin LCN2; SV-40-induced 24P3 protein; Lipocalin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518861 349.00
Check your price
  • SLC22A17 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520543
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (20 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Neutrophil gelatinase-associated lipocalin receptor; Lipocalin-2 receptor; Brain-type organic cation transporter; 24p3 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520543 349.00
Check your price
  • NGAL Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Dog
  • Conjugate Unconjugated
  • Catalog # ABS0471402
  • Size 200µL
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Neutrophil gelatinase-associated lipocalin, HNL; p25, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, Lipocalin-2, Oncogene 24p3
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0471402 650.00
Check your price
  • NGAL Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Dog
  • Conjugate Unconjugated
  • Catalog # ABS0471702
  • Size 200µL
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Neutrophil gelatinase-associated lipocalin, HNL; p25, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, Lipocalin-2, Oncogene 24p3
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0471702 650.00
Check your price
  • NGAL Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Pig
  • Conjugate Unconjugated
  • Catalog # ABS0481702
  • Size 200µL
Applications
Western Blot (Assay Dependent)
Alternate names:
Neutrophil gelatinase-associated lipocalin, HNL; p25, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, Lipocalin-2, Oncogene 24p3
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0481702 650.00
Check your price
  • NGAL Monoclonal Antibody for Western Blot, IHC, IHC (P) and ELISA
  • Host Mouse
  • Target Species Pig
  • Conjugate Unconjugated
  • Catalog # ABS0482802
  • Size 200µL
Applications
ELISA (Assay Dependent), Immunohistochemistry (1:25), Immunohistochemistry (Paraffin) (1:1000), Western Blot (Assay Dependent)
Alternate names:
Neutrophil gelatinase-associated lipocalin, HNL; p25, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, Lipocalin-2, Oncogene 24p3
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0482802 650.00
Check your price