1 - 17 of 17 products displayed
  • NGAL (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP75672
  • Size 100µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75672 620.00
Check your price
  • NGAL (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP75671
  • Size 10µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Neurophil Gelatinase Associated Lipocalin
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75671 185.00
Check your price
  • NGAL (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP75673
  • Size 1mg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Neurophil Gelatinase Associated Lipocalin
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75673 2,900.00
Check your price
  • NGAL Monoclonal Antibody for Western Blot, IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB2110202
  • Size 200µl
Applications
ELISA (1:8,000), Immunohistochemistry (Assay dependent), Western Blot (1:50)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB2110202 650.00
Check your price
  • NGAL Monoclonal Antibody, Biotin conjugate for ELISA
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Biotin
  • Catalog # HYB21102B005
  • Size 50µl
Applications
ELISA (1:16000)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB21102B005 425.00
Check your price
  • NGAL Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # ABS0623302
  • Size 200µL
Applications
ELISA (Assay Dependent)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0623302 650.00
Check your price
  • NGAL Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # ABS0621402
  • Size 200µL
Applications
ELISA (Assay Dependent)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0621402 650.00
Check your price
  • NGAL Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Dog, Human, Pig
  • Conjugate Unconjugated
  • Catalog # ABS0381402
  • Size 200µL
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0381402 650.00
Check your price
  • NGAL Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # ABS0381502
  • Size 200µL
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0381502 650.00
Check your price
  • NGAL Monoclonal Antibody for Western Blot, IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB2110102
  • Size 200µl
Applications
ELISA (Assay dependent), Immunohistochemistry (Assay dependent), Western Blot (Assay dependent)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB2110102 650.00
Check your price
  • NGAL Monoclonal Antibody for ELISA and IB
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # ABS0380402
  • Size 200µL
Applications
ELISA (Assay Dependent), Immunoblot (Assay Dependent)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0380402 650.00
Check your price
  • NGAL Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # ABS0382602
  • Size 200µL
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0382602 650.00
Check your price
  • NGAL Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA175310
  • Size 500µg
Applications
ELISA (Assay Dependent)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175310 1,500.00
Check your price
  • NGAL Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # ABS0391502
  • Size 200µL
Applications
ELISA (Assay Dependent)
Alternate names:
p25; Siderocalin LCN2; Lipocalin-2; Alpha-2U globulin-related protein; Alpha-2-microglobulin-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0391502 650.00
Check your price
  • NGAL Monoclonal Antibody, Biotin conjugate for ELISA
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # ABS03932B005
  • Size 50µL
Applications
ELISA (1:4000)
Alternate names:
p25; Siderocalin LCN2; Lipocalin-2; Alpha-2U globulin-related protein; Alpha-2-microglobulin-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS03932B005 475.00
Check your price
  • NGAL Polyclonal Antibody for Western Blot, IP and ELISA
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA546938
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
p25; Siderocalin LCN2; Lipocalin-2; Alpha-2U globulin-related protein; Alpha-2-microglobulin-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546938 375.00
Check your price
  • NGAL Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546916
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.25-5 µg/ml)
Alternate names:
p25; Siderocalin LCN2; SV-40-induced 24P3 protein; Lipocalin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546916 375.00
Check your price