1 - 30 of 30 products displayed
  • Nuclear Matrix Protein p84 Antibody (Monoclonal, 5E10)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA123261
  • Size 100 µg
Applications
ChIP assay (Assay Dependent), Immunocytochemistry (0.5-2 ug/ml), Immunofluorescence (0.5-2 µg/ml), Immunohistochemistry (Assay Dependent), Immunoprecipitation (1:100-1:500), Western Blot (0.3-2 µg/ml)
Alternate names:
Nuclear matrix protein p84; hTREX84
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123261 309.00
Check your price
  • NUP107 Antibody (Monoclonal, 39C7)
  • Host Mouse
  • Target Species Human, Mouse, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # MA110031
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500)
Alternate names:
Nucleoporin Nup107; 107 kDa nucleoporin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110031 375.00
Check your price
  • NUP62 Antibody (Monoclonal, RL1)
  • Host Mouse
  • Target Species Amphibian, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # MA1071
  • Size 100 µL
Applications
Immunofluorescence (1:100), Immunohistochemistry (1:200), Immunoprecipitation (Assay dependent), Western Blot (1:1,000)
Alternate names:
Nucleoporin Nup62; 62 kDa nucleoporin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1071 275.00
Check your price
  • NUP62 Antibody (Monoclonal, RL31)
  • Host Hamster
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA1073
  • Size 100 µg
Applications
Immunofluorescence (1:50), Western Blot (1:200)
Alternate names:
Nucleoporin Nup62; 62 kDa nucleoporin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1073 275.00
Check your price
  • NUP50 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131827
  • Size 50 µg
Applications
Immunohistochemistry (Paraffin) (Assay-dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
Nucleoporin Nup50; Nuclear pore-associated protein 60 kDa-like; 50 kDa nucleoporin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131827 399.00
Check your price
  • NFYA Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124431
  • Size 100 µg
Applications
Immunohistochemistry (Assay-dependent), Western Blot (5-8 µg/ml)
Alternate names:
Nuclear transcription factor Y subunit A; CAAT box DNA-binding protein subunit A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124431 375.00
Check your price
  • Ubinuclein 1 Antibody (Monoclonal, UBN1-02)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110458
  • Size 100 ug
Applications
Immunoprecipitation (Assay-Dependent), Western Blot (1-5 ug/ml)
Alternate names:
Ubiquitously expressed nuclear protein; Protein VT4; HIRA-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110458 249.00
Check your price
  • Nurim Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141391
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (1:100-1:2000)
Alternate names:
Nuclear rim protein; Nuclear envelope membrane protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141391 309.00
Check your price
  • NFkB p52 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA130410
  • Size 1 mL
Applications
Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay-dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 2; DNA-binding factor KBF2; Oncogene Lyt-10; Lymphocyte translocation chromosome 10 protein; H2TF1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130410 425.00
Check your price
  • NFkB p65 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA130408
  • Size 1 mL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay-Dependent)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; Nuclear factor NF-kappa-B p65 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130408 475.00
Check your price
  • NFYA Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA132119
  • Size 100 µg
Applications
ELISA (1-2 µl), Gel Shift (Assay-dependent), Immunoprecipitation (1:1000), Western Blot (1:1000)
Alternate names:
Nuclear transcription factor Y subunit A; CAAT box DNA-binding protein subunit A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132119 550.00
Check your price
  • NUP107 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520562
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Nucleoporin Nup107; 107 kDa nucleoporin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520562 349.00
Check your price
  • NUP107 Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA520563
  • Size 100 µg
Applications
Immunocytochemistry (2.5 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Nucleoporin Nup107; 107 kDa nucleoporin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520563 349.00
Check your price
  • NUP160 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520565
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Nucleoporin Nup160; 160 kDa nucleoporin; Gene trap locus 1-13 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520565 349.00
Check your price
  • Nudel Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519082
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Mitosin-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519082 349.00
Check your price
  • NFIL3 Antibody
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131434
  • Size 50 µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
E4 promoter-binding protein 4; Transcriptional activator NF-IL3A; Interleukin-3-binding protein 1; Interleukin-3 promoter transcriptional activator; Embryo implantation-related NFIL3/E4BP4-like transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131434 349.00
Check your price
  • SET Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131457
  • Size 50 µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Template-activating factor I; Phosphatase 2A inhibitor I2PP2A; PHAPII; Inhibitor of granzyme A-activated DNase; HLA-DR-associated protein II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131457 399.00
Check your price
  • LAP1 Antibody (Monoclonal, RL13)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1074
  • Size 200 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:20-1:50), Immunoprecipitation (Assay dependent), Western Blot (1:200)
Alternate names:
Lamina-associated polypeptide 1B; Lamina-associated polypeptide 1C; Lamin-associated protein 1B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1074 275.00
Check your price
  • NOP1 Antibody (Monoclonal, 28F2)
  • Host Mouse
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # MA110025
  • Size 500 µL
Applications
Immunocytochemistry (1:2000-1:20,000), Immunofluorescence (1:2000-1:20,000), Immunoprecipitation (2-4 µl/lysate), Western Blot (1:10,000-1:25,000)
Alternate names:
U3 small nucleolar RNA-associated protein NOP1; Histone-glutamine methyltransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110025 375.00
Check your price
  • O-linked N-acetylglucosamine (O-GlcNAc) Antibody (Monoclonal, RL2)
  • Host Mouse
  • Target Species All
  • Conjugate Unconjugated
  • Catalog # MA1072
  • Size 100 µL
Applications
ChIP assay (Assay Dependent), Dot blot (1/800), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (Assay dependent), Western Blot (1:1,000)
Alternate names:
O-linked N-acetylglucosamine transferase 110 kDa subunit; O-GlcNAc transferase subunit p110
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1072 375.00
Check your price
  • Fibrillarin Antibody (Monoclonal, 38F3)
  • Host Mouse
  • Target Species Fruit fly, Human, Rat
  • Conjugate Unconjugated
  • Catalog # 480009
  • Size 100 µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Western Blot (1:1,000)
Alternate names:
Nucleolar protein 1; Histone-glutamine methyltransferase; 34 kDa nucleolar scleroderma antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
480009 309.00
Check your price
  • HHV-8 Antibody (Monoclonal, LN35)
  • Host Rat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA126628
  • Size 500 µL
Applications
Immunohistochemistry (Frozen) (1:25-1:50), Immunohistochemistry (Paraffin) (1:25-1:50)
Alternate names:
HHV-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126628 375.00
Check your price
  • Lamin B2 Antibody (Monoclonal, LN43)
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Pig, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA106104
  • Size 100 µL
Applications
Flow Cytometry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:100- 1:200), Western Blot (1:100-1:1000)
Alternate names:
LAMB2, LMN2, MGC2721
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106104 309.00
Check your price
  • NOP2 Antibody (Monoclonal, 22G1)
  • Host Mouse
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # MA110027
  • Size 500 µL
Applications
Immunocytochemistry (1:1000-1:5000), Immunofluorescence (1:1000-1:5000), Immunohistochemistry (1:1000-1:5000), Immunoprecipitation (Assay Dependent), Western Blot (1:2000-1:10,000)
Alternate names:
Nucleolar protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110027 309.00
Check your price
  • Lamin B1/B2 Antibody (Monoclonal, X223)
  • Host Mouse
  • Target Species Bovine, Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA15822
  • Size 2 mL
Applications
Immunofluorescence (Assay Dependent), Western Blot (1:50)
Alternate names:
B type lamin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15822 550.00
Check your price
  • HIF1A Antibody, Biotin (Monoclonal, H1alpha67)
  • Host Mouse
  • Target Species Avian, Bovine, Dog, Ferret, Human, Mouse, Non-human primate, Sheep, Pig, Rat
  • Conjugate Biotin
  • Catalog # MA146457
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:300), Immunoprecipitation (1:10), Western Blot (1:500)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146457 325.00
Check your price
  • CTCF Antibody
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA141663
  • Size 100 µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
CTCFL paralog; CCCTC-binding factor; 11-zinc finger protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141663 309.00
Check your price
  • Ki-67 Antibody, FITC (Monoclonal, 7B11)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # 334711
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml)
Alternate names:
antigen KI-67, proliferation-related Ki-67 antigen, RP11-380J17.2, KIA
Catalog # Size Price (AUD) Your Price (AUD) Qty
334711 329.00
Check your price
  • Ki-67 Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA516785
  • Size 500 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:300)
Alternate names:
antigen KI-67, proliferation-related Ki-67 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516785 309.00
Check your price
  • Nesprin 1 Antibody (Monoclonal, 6A)
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA113082X
  • Size 20 µL
Applications
Western Blot (1:1000)
Alternate names:
candidate plasticity gene 2, syne-1 fragment
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA113082X 119.00
Check your price