1 - 30 of 30 products displayed
  • Estrogen Receptor alpha Antibody, Biotin (Monoclonal, AER314)
  • Host Mouse
  • Target Species Rabbit, Rat, Human, Mouse, Bovine
  • Conjugate Biotin
  • Catalog # MA512325
  • Size 500 µL
Applications
Affinity Purification (Assay Dependent), Gel Shift (1mg/mL), Immunocytochemistry (1:500-1:2000), Immunofluorescence (1:500-1:2000), Immunoprecipitation (2 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512325 387.00
Check your price
  • Estrogen Receptor alpha Antibody (Monoclonal, AER314)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA512328
  • Size 500 µL
Applications
Gel Shift (1mg/mL), Immunoprecipitation (2 µg/ml), Western Blot (1:100-1:400)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512328 309.00
Check your price
  • Androgen Receptor Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA1110
  • Size 200 µL
Applications
Immunocytochemistry (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (1:10-1:100), Immunoprecipitation (Assay dependent), Western Blot (1:500)
Alternate names:
Nuclear receptor subfamily 3 group C member 4; Dihydrotestosterone receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1110 349.00
Check your price
  • Estrogen Receptor beta Antibody (Monoclonal, PPG5/10)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA127412
  • Size 500 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (1:20), Immunohistochemistry (Paraffin) (1:20)
Alternate names:
Nuclear receptor subfamily 3 group A member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA127412 425.00
Check your price
  • Phospho-Progesterone Receptor (Ser554) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA531313
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (1:100-1:500), Western Blot (1:500-1:20,000)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531313 349.00
Check your price
  • Phospho-Progesterone Receptor (Ser294) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537472
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537472 309.00
Check your price
  • Progesterone Receptor Antibody (Monoclonal, 4D8)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA123268
  • Size 200 µg
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (Assay dependent)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123268 600.00
Check your price
  • Progesterone Receptor Antibody (Monoclonal, SP2)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514505
  • Size 500 µL
Applications
Immunofluorescence (1-2 µg/ml), Immunohistochemistry (Paraffin) (1:100-1:400), Western Blot (1:1000-1:2000)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514505 700.00
Check your price
  • Progesterone Receptor Antibody Polyclonal (PA5-16709)
  • Host Rabbit
  • Target Species Human
  • Conjugate
  • Catalog # PA516709
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516709 339.00
Check your price
  • Estrogen Receptor alpha Antibody (Monoclonal, SP1)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA139540
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA139540 1
Check your price
  • Estrogen Receptor alpha Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137651
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137651 399.00
Check your price
  • Phospho-Androgen Receptor (Ser94) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538406
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500)
Alternate names:
Nuclear receptor subfamily 3 group C member 4; Dihydrotestosterone receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538406 309.00
Check your price
  • Phospho-Glucocorticoid Receptor (Ser211) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA517668
  • Size 100 µL
Applications
Immunocytochemistry (1:1600), Immunofluorescence (1:1600), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Nuclear receptor subfamily 3 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517668 309.00
Check your price
  • Phospho-Estrogen Receptor beta (Ser105) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538488
  • Size 100 µg
Applications
Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group A member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538488 309.00
Check your price
  • Phospho-Estrogen Receptor beta (Ser87) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539774
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Nuclear receptor subfamily 3 group A member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539774 309.00
Check your price
  • Phospho-Progesterone Receptor (Ser400) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537473
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537473 309.00
Check your price
  • Progesterone Receptor Antibody (Monoclonal, PgR636)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512581
  • Size 500 µL
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:10-1:20), Western Blot (1:500-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512581 850.00
Check your price
  • Progesterone Receptor Antibody (Monoclonal, SP42)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516394
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:400)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516394 700.00
Check your price
  • Progesterone Receptor Antibody (Monoclonal, SP2)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516393
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:400)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516393 700.00
Check your price
  • Progesterone Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531314
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531314 349.00
Check your price
  • Progesterone Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534849
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534849 349.00
Check your price
  • Progesterone Receptor Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA516435
  • Size 500 µL
Applications
Western Blot (5-10 µg/mL)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516435 349.00
Check your price
  • Phospho-Progesterone Receptor (Ser190) Antibody (Monoclonal, 1154)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA1413
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (2-4 ug/ml), Immunoprecipitation (Assay dependent), Western Blot (4 ug/ml)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1413 275.00
Check your price
  • Phospho-Progesterone Receptor (Ser294) Antibody (Monoclonal, 608)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA1414
  • Size 200 µg
Applications
ELISA (Assay dependent), Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1:20), Immunofluorescence (1:20), Immunohistochemistry (Paraffin) (1:500), Immunoprecipitation (Assay dependent), Western Blot (10 ug/ml)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1414 275.00
Check your price
  • Estrogen Receptor alpha Antibody (Monoclonal, AER311)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA112600
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112600 275.00
Check your price
  • Androgen Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517771
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Nuclear receptor subfamily 3 group C member 4; Dihydrotestosterone receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517771 275.00
Check your price
  • Glucocorticoid Receptor Antibody
  • Host Sheep
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA110856
  • Size 200 µL
Applications
Western Blot (1:500-1:1,000)
Alternate names:
Nuclear receptor subfamily 3 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA110856 475.00
Check your price
  • Glucocorticoid Receptor Antibody
  • Host Sheep
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA128366
  • Size 100 µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128366 375.00
Check your price
  • Glucocorticoid Receptor Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521341
  • Size 100 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (1:500-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Nuclear receptor subfamily 3 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521341 349.00
Check your price
  • Estrogen Receptor beta Antibody (Monoclonal, PPG5/10)
  • Host Mouse
  • Target Species Camelid, Human, Non-human primate, Sheep, Pig, Reptile, Rat
  • Conjugate Unconjugated
  • Catalog # MA181281
  • Size 100 µL
Applications
Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1/5-1/50)
Alternate names:
Nuclear receptor subfamily 3 group A member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181281 199.00
Check your price