1 - 20 of 20 products displayed
  • LCK Monoclonal Antibody for Western Blot, IF, ICC, Flow, IP and FN
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate
  • Catalog # AHO0472
  • Size 500µl
Applications
Flow Cytometry (Assay-Dependent), Functional Assay (Assay Dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (Assay-Dependent), Immunoprecipitation (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHO0472 Not Orderable Online
Check your price
  • LCK Monoclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA512303
  • Size 500µL
Applications
Flow Cytometry (1ug/test), Immunocytochemistry (1:20), Immunofluorescence (1:20), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512303 309.00
Check your price
  • Phospho-LCK (Tyr505) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44850G
  • Size 10blots
Applications
Western Blot (Assay Dependent)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
44850G 309.00
Check your price
  • LCK Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA141203
  • Size 200µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA141203 375.00
Check your price
  • Phospho-LCK (Tyr394) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA537628
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537628 309.00
Check your price
  • Phospho-LCK (Ser158) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44842G
  • Size 100µL
Applications
Western Blot (Assay Dependent)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
44842G 309.00
Check your price
  • Phospho-LCK (Tyr192) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44844G
  • Size 10blots
Applications
Western Blot (Assay Dependent)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
44844G 309.00
Check your price
  • Phospho-LCK (Tyr505) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA126703
  • Size 50µL
Applications
Western Blot (0.1-1 µg/ml)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126703 349.00
Check your price
  • Phospho-LCK (Tyr192) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA126702
  • Size 50µL
Applications
Western Blot (0.1-1 µg/ml)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126702 349.00
Check your price
  • LCK Monoclonal Antibody for Western Blot, ICC, Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119197
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119197 199.00
Check your price
  • Phospho-LCK (Tyr505) Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA517813
  • Size 100µL
Applications
Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517813 309.00
Check your price
  • Phospho-LCK (Tyr393) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539696
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539696 309.00
Check your price
  • LCK Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515295
  • Size 100µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515295 349.00
Check your price
  • Phospho-LCK (Ser59) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539760
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539760 309.00
Check your price
  • LCK Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA534653
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534653 349.00
Check your price
  • LCK Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA519422
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:200-1:500), Western Blot (1:200-1:500)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519422 349.00
Check your price
  • LCK Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524615
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524615 349.00
Check your price
  • LCK Monoclonal Antibody for Western Blot, IF, Flow and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A13995
  • Size 100µL
Applications
Flow Cytometry (1:400), Immunofluorescence (1:400), Immunoprecipitation (Assay Dependent), Western Blot (1:1,000)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
A13995 309.00
Check your price
  • LCK Monoclonal Antibody, Alexa Fluor® 647 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Alexa Fluor® 647
  • Catalog # MA518166
  • Size 100µg
Applications
Flow Cytometry (Assay-dependent)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518166 349.00
Check your price
  • Phospho-LCK (Ser158) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA126701
  • Size 50µL
Applications
Western Blot (0.1-1 µg/ml)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126701 349.00
Check your price