1 - 3 of 3 products displayed
  • LIMS1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530240
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Renal carcinoma antigen NY-REN-48; Particularly interesting new Cys-His protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530240 349.00
Check your price
  • LIMS1 Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515876
  • Size 100µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Renal carcinoma antigen NY-REN-48; Particularly interesting new Cys-His protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515876 349.00
Check your price
  • LIMS1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA529152
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Renal carcinoma antigen NY-REN-48; Particularly interesting new Cys-His protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529152 349.00
Check your price