1 - 30 of 30 products displayed
  • PYY Polyclonal Antibody, Biotin conjugate for ELISA
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # PA19590
  • Size 100 µL
Applications
ELISA (1:2000-1:10000)
Alternate names:
Peptide tyrosine tyrosine; PYY-I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19590 375.00
Check your price
  • PYY Polyclonal Antibody, Biotin conjugate for ELISA
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # PA19589
  • Size 100 µL
Applications
ELISA (1:2000-1:10000)
Alternate names:
Peptide tyrosine tyrosine; PYY-I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19589 375.00
Check your price
  • UBE2J1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519675
  • Size 200 µL
Applications
Immunocytochemistry (5 µg/ml), Immunofluorescence (5 µg/ml), Western Blot (1:250)
Alternate names:
Yeast ubiquitin-conjugating enzyme UBC6 homolog E; Non-canonical ubiquitin-conjugating enzyme 1; E2 ubiquitin-conjugating enzyme J1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519675 349.00
Check your price
  • C-Peptide Monoclonal Antibody, Biotin conjugate for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA110317
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (2-5 ug/ml)
Alternate names:
IDDM2, ILPR, IRDN, MODY10, proinsulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110317 249.00
Check your price
  • C-Peptide Monoclonal Antibody for ICC, IHC (P), ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119159
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (2-5 µg/ml), Radioimmune Assays (Assay Dependent)
Alternate names:
ILPR, IRDN
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119159 309.00
Check your price
  • UBC9 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519561
  • Size 100 µg
Applications
Immunocytochemistry (5 µg/ml), Immunofluorescence (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
p18; Ubiquitin-protein ligase I; Ubiquitin-conjugating enzyme E2 I; Ubiquitin carrier protein I; Ubiquitin carrier protein 9; SUMO-protein ligase; Ubiquitin-conjugating enzyme UbcE2A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519561 349.00
Check your price
  • UBC9 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519609
  • Size 100 µg
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Western Blot (1 µg/ml)
Alternate names:
p18; Ubiquitin-protein ligase I; Ubiquitin-conjugating enzyme E2 I; Ubiquitin carrier protein I; Ubiquitin carrier protein 9; SUMO-protein ligase; Ubiquitin-conjugating enzyme UbcE2A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519609 349.00
Check your price
  • beta Amyloid Monoclonal Antibody for IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191209
  • Size 500 µL
Applications
Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI; ABPP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191209 375.00
Check your price
  • Chromogranin B Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA110839
  • Size 200 µL
Applications
Immunohistochemistry (1:500-1:5000), Western Blot (1:2500-1:10000)
Alternate names:
Secretogranin I; Chromogranin-B; Secretogranin-I; Glucagonoma peptide
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA110839 449.00
Check your price
  • CGRP Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Rat
  • Conjugate
  • Catalog # PA519755
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:400), Western Blot (1-5 µg/ml)
Alternate names:
Calcitonin gene-related peptide I; Alpha-type CGRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519755 349.00
Check your price
  • JMJD6 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA124739
  • Size 100 µL
Applications
Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
Phosphatidylserine receptor; Peptide-lysine 5-dioxygenase JMJD6; Lysyl-hydroxylase JMJD6; Jumonji domain-containing protein 6; JmjC domain-containing protein 6; Histone arginine demethylase JMJD6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124739 375.00
Check your price
  • GRPR Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OPA115619
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
GRP-preferring bombesin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA115619 499.00
Check your price
  • UBE2A Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA534615
  • Size 100 µg
Applications
Immunofluorescence (1:200), Immunohistochemistry (Paraffin) (1 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Ubiquitin-protein ligase A; Ubiquitin carrier protein A; RAD6 homolog A; E2 ubiquitin-conjugating enzyme A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534615 349.00
Check your price
  • GPR10 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131068
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
hGR3; G-protein coupled receptor 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131068 399.00
Check your price
  • GAP43 Polyclonal Antibody for Western Blot, IF, ICC, IHC (F) and IHC (PFA)
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519470
  • Size 100 µg
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (Frozen) (1:500), Immunohistochemistry (PFA fixed) (1:300-1:1000), Western Blot (1 µg/ml)
Alternate names:
Protein F1; Growth-associated protein 43; Axonal membrane protein GAP-43; Calmodulin-binding protein P-57
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519470 349.00
Check your price
  • Glucagon-Like Peptide 1 Monoclonal Antibody, Biotin conjugate for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # ABS03310B005
  • Size 50 µl
Applications
ELISA (1:40,000)
Alternate names:
MGC138331
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS03310B005 449.00
Check your price
  • S-peptide Epitope Tag Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA1981
  • Size 100 µg
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Immunohistochemistry (10 µg/ml), Western Blot (1 µg/ml)
Alternate names:
S-tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1981 249.00
Check your price
  • BNP Monoclonal Antibody, HRP conjugate for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate HRP
  • Catalog # MA191673
  • Size 200 µg
Applications
ELISA (Assay-Dependent), Western Blot (Assay dependent)
Alternate names:
Gamma-brain natriuretic peptide
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191673 399.00
Check your price
  • TAP1 Polyclonal Antibody for Western Blot, Flow, IP and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131487
  • Size 100 µg
Applications
ELISA (Assay-dependent), Flow Cytometry (1:10,000), Immunoprecipitation (1:10,000), Western Blot (1:1000)
Alternate names:
Really interesting new gene 4 protein; Peptide transporter involved in antigen processing 1; Peptide transporter TAP1; Peptide transporter PSF1; Peptide supply factor 1; ATP-binding cassette sub-family B member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131487 550.00
Check your price
  • UBE2E1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542281
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Ubiquitin-protein ligase E1; Ubiquitin carrier protein E1; UbcH6; E2 ubiquitin-conjugating enzyme E1; (E3-independent) E2 ubiquitin-conjugating enzyme E1; UbcM3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542281 309.00
Check your price
  • NPR3 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515394
  • Size 400 µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Atrial natriuretic peptide receptor type C; Atrial natriuretic peptide clearance receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515394 349.00
Check your price
  • Calcitonin Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Dog, Horse, Human, Mouse, Non-human primate, Sheep, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA514552
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Calcitonin gene-related peptide I; Alpha-type CGRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514552 349.00
Check your price
  • UBE2D1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA511958
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Ubiquitin-protein ligase D1; Ubiquitin-conjugating enzyme E2-17 kDa 1; Ubiquitin-conjugating enzyme E2(17)KB 1; Ubiquitin carrier protein D1; UbcH5; UBC4/5 homolog; Stimulator of Fe transport; E2 ubiquitin-conjugating enzyme D1; (E3-independent) E2 ubiquitin-conjugating enzyme D1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511958 349.00
Check your price
  • UBE2D2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542279
  • Size 100 µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (1.25 µg/ml)
Alternate names:
p53-regulated ubiquitin-conjugating enzyme 1; Ubiquitin-protein ligase D2; Ubiquitin-conjugating enzyme E2-17 kDa 2; Ubiquitin-conjugating enzyme E2(17)KB 2; Ubiquitin carrier protein D2; E2 ubiquitin-conjugating enzyme D2; (E3-independent) E2 ubiquitin-conjugating enzyme D2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542279 309.00
Check your price
  • UBE2D3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542280
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Ubiquitin-protein ligase D3; Ubiquitin carrier protein D3; E2 ubiquitin-conjugating enzyme D3; (E3-independent) E2 ubiquitin-conjugating enzyme D3; Ubiquitin-conjugating enzyme E2-17 kDa 3; Ubiquitin-conjugating enzyme E2(17)KB 3; Phosphoarginine phosphatase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542280 309.00
Check your price
  • Glucagon Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513442
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Glucagon; Glicentin; Glicentin-related polypeptide; GRPP; Oxyntomodulin; OXY; OXM; Glucagon; Glucagon-like peptide 1; GLP-1; Glucagon-like peptide 1(7-37); GLP-1(7-37); Glucagon-like peptide 1(7-36); GLP-1(7-36); Glucagon-like peptide 2; GLP-2; GCG
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513442 349.00
Check your price
  • UBE2V2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131544
  • Size 50 µg
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Ubiquitin-conjugating enzyme variant MMS2; Vitamin D3-inducible protein; MMS2 homolog; Enterocyte differentiation-promoting factor 1; Enterocyte differentiation-associated factor 1; DDVit 1; Ubc-like protein MMS2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131544 399.00
Check your price
  • TAP1 Polyclonal Antibody for Western Blot, Flow, IP and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA186830
  • Size 50 µg
Applications
ELISA (1:10000), Flow Cytometry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
Really interesting new gene 4 protein; Peptide transporter involved in antigen processing 1; Peptide transporter TAP1; Peptide transporter PSF1; Peptide supply factor 1; ATP-binding cassette sub-family B member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186830 375.00
Check your price
  • E2EPF Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511972
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Ubiquitin-protein ligase S; Ubiquitin-conjugating enzyme E2-EPF5; Ubiquitin-conjugating enzyme E2-24 kDa; Ubiquitin carrier protein S; E2-EPF; E2 ubiquitin-conjugating enzyme S
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511972 349.00
Check your price
  • E2EPF Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511973
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Ubiquitin-protein ligase S; Ubiquitin-conjugating enzyme E2-EPF5; Ubiquitin-conjugating enzyme E2-24 kDa; Ubiquitin carrier protein S; E2-EPF; E2 ubiquitin-conjugating enzyme S
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511973 349.00
Check your price