1 - 30 of 30 products displayed
  • Thyroid Peroxidase Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA183490
  • Size 200µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
MSA, TDH2A, TPX
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183490 399.00
Check your price
  • Glutathione Peroxidase 4 (Hu) for Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # LFP0015
  • Size 500µg
Applications
Control (Assay Dependent)
Alternate names:
Glutathione peroxidase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFP0015 293.00
Check your price
  • GPX4 Monoclonal Antibody for IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFMA0085
  • Size 100µL
Applications
ELISA (Assay dependent), Immunoprecipitation (1-2 µl)
Alternate names:
Glutathione peroxidase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0085 309.00
Check your price
  • GPX4 Monoclonal Antibody for IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFMA0059
  • Size 100µL
Applications
ELISA (Assay dependent), Immunoprecipitation (1-2 µl)
Alternate names:
Glutathione peroxidase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0059 309.00
Check your price
  • PRDX1 Polyclonal Antibody for Western Blot, IF and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA3750
  • Size 100µL
Applications
Immunofluorescence (1:100), Immunohistochemistry (Assay-dependent), Western Blot (1:1,000)
Alternate names:
Thioredoxin-dependent peroxide reductase 2; Thioredoxin peroxidase 2; Proliferation-associated gene protein; Natural killer cell-enhancing factor A; Heme-binding 23 kDa protein; HBP23; Osteoblast-specific factor 3; Macrophage 23 kDa stress protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3750 375.00
Check your price
  • PRDX6 Monoclonal Antibody for Western Blot, IF, ICC, Flow, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFMA0018
  • Size 100µL
Applications
ELISA (Assay dependent), Immunoprecipitation (2 µg), Western Blot (0.5 ug/mL)
Alternate names:
Thiol-specific antioxidant protein; Non-selenium glutathione peroxidase; Antioxidant protein 2; Acidic calcium-independent phospholipase A2; 1-Cys peroxiredoxin; Red blood cells page spot 12; Liver 2D page spot 40; 24 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0018 349.00
Check your price
  • PRDX1 Monoclonal Antibody for Western Blot, Flow and ELISA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # LFMA0073
  • Size 100µL
Applications
ELISA (Assay dependent), Western Blot (1:1000)
Alternate names:
Thioredoxin-dependent peroxide reductase 2; Thioredoxin peroxidase 2; Proliferation-associated gene protein; Natural killer cell-enhancing factor A; Osteoblast-specific factor 3; Macrophage 23 kDa stress protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0073 309.00
Check your price
  • PRDX6 Monoclonal Antibody for ICC, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFMA0013
  • Size 100µL
Applications
ELISA (Assay dependent), Immunocytochemistry (20 µl/ml), Immunoprecipitation (2 µg)
Alternate names:
Red blood cells page spot 12; Non-selenium glutathione peroxidase; Liver 2D page spot 40; Antioxidant protein 2; Acidic calcium-independent phospholipase A2; 24 kDa protein; 1-Cys peroxiredoxin
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0013 309.00
Check your price
  • PRDX6 Monoclonal Antibody for IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFMA0067
  • Size 100µL
Applications
ELISA (Assay dependent), Immunoprecipitation (2 µg)
Alternate names:
Red blood cells page spot 12; Non-selenium glutathione peroxidase; Liver 2D page spot 40; Antioxidant protein 2; Acidic calcium-independent phospholipase A2; 24 kDa protein; 1-Cys peroxiredoxin
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0067 309.00
Check your price
  • PRDX6 Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFMA0104
  • Size 100µL
Applications
ELISA (Assay dependent), Immunoprecipitation (5 µg), Western Blot (0.5 ug/ml)
Alternate names:
Red blood cells page spot 12; Non-selenium glutathione peroxidase; Liver 2D page spot 40; Antioxidant protein 2; Acidic calcium-independent phospholipase A2; 24 kDa protein; 1-Cys peroxiredoxin
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0104 309.00
Check your price
  • PRDX1 Monoclonal Antibody for IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFMA0069
  • Size 100µL
Applications
ELISA (Assay dependent), Immunoprecipitation (2 µg)
Alternate names:
Thioredoxin-dependent peroxide reductase 2; Thioredoxin peroxidase 2; Proliferation-associated gene protein; Natural killer cell-enhancing factor A
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0069 309.00
Check your price
  • PRDX2 Polyclonal Antibody for Western Blot, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA3751
  • Size 100µL
Applications
Immunocytochemistry (1:500), Immunohistochemistry (Assay-dependent), Western Blot (1:10,000)
Alternate names:
Thioredoxin-dependent peroxide reductase 1; Thioredoxin peroxidase 1; Thiol-specific antioxidant protein; PRP; Natural killer cell-enhancing factor B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3751 Not Orderable Online
Check your price
  • PRDX4 Monoclonal Antibody for Western Blot, IF, ICC, Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFMA0014
  • Size 100µL
Applications
ELISA (Assay dependent), Immunoprecipitation (1-2 µl), Western Blot (1:2000)
Alternate names:
Thioredoxin-dependent peroxide reductase A0372; Thioredoxin peroxidase AO372; Peroxiredoxin IV; Antioxidant enzyme AOE372
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0014 309.00
Check your price
  • PRDX4 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA3753
  • Size 100µL
Applications
Immunocytochemistry (1:500), Immunohistochemistry (Assay-dependent), Western Blot (1:10,000)
Alternate names:
Thioredoxin-dependent peroxide reductase A0372; Thioredoxin peroxidase AO372; Peroxiredoxin IV; Antioxidant enzyme AOE372
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3753 349.00
Check your price
  • GPX1 Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFMA0206
  • Size 100µL
Applications
ELISA (Assay Dependent), Immunoprecipitation (1 µl), Western Blot (1:5000)
Alternate names:
Cellular glutathione peroxidase; Selenium-dependent glutathione peroxidase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0206 375.00
Check your price
  • GPX3 Monoclonal Antibody for IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFMA0145
  • Size 100µL
Applications
ELISA (Assay Dependent), Immunoprecipitation (1 µl )
Alternate names:
Plasma glutathione peroxidase; Extracellular glutathione peroxidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0145 309.00
Check your price
  • PRDX5 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFMA0017
  • Size 100µL
Applications
ELISA (Assay dependent), Western Blot (1:2000)
Alternate names:
Thioredoxin reductase; Thioredoxin peroxidase PMP20; TPx type VI; Peroxisomal antioxidant enzyme; Peroxiredoxin V; PLP; Liver tissue 2D-page spot 71B; Antioxidant enzyme B166; Alu corepressor 1; Liver tissue 2D-page spot 2D-0014IV
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0017 349.00
Check your price
  • PRDX5 Monoclonal Antibody for Western Blot, ICC, Flow, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # LFMA0002
  • Size 100µL
Applications
ELISA (Assay dependent), Immunocytochemistry (20 µl/ml), Immunoprecipitation (1-2 µl), Western Blot (1:1000)
Alternate names:
Thioredoxin reductase; Thioredoxin peroxidase PMP20; TPx type VI; Peroxisomal antioxidant enzyme; Peroxiredoxin V; PLP; Liver tissue 2D-page spot 71B; Antioxidant enzyme B166; Alu corepressor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0002 309.00
Check your price
  • PRDX5 Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFMA0001
  • Size 100µL
Applications
ELISA (Assay dependent), Immunoprecipitation (1-2 µl), Western Blot (1:1000)
Alternate names:
Thioredoxin reductase; Thioredoxin peroxidase PMP20; TPx type VI; Peroxisomal antioxidant enzyme; Peroxiredoxin V; PLP; Liver tissue 2D-page spot 71B; Antioxidant enzyme B166; Alu corepressor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0001 309.00
Check your price
  • PRDX2 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFMA0144
  • Size 100µL
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (1:2000-1:4000)
Alternate names:
Thioredoxin-dependent peroxide reductase 1; Thioredoxin peroxidase 1; Thiol-specific antioxidant protein; PRP; Natural killer cell-enhancing factor B
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0144 309.00
Check your price
  • PRDX1 Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFMA0214
  • Size 100µL
Applications
ELISA (Assay Dependent), Immunoprecipitation (1 µg), Western Blot (0.05 µg/ml)
Alternate names:
Thioredoxin-dependent peroxide reductase 2; Thioredoxin peroxidase 2; Proliferation-associated gene protein; Natural killer cell-enhancing factor A
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0214 309.00
Check your price
  • GPX7 Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA528854
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (Assay-Dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:500-1:3000)
Alternate names:
CL683, GPX6, GPx-7, GSHPx-7, NPGPx, glutathione peroxidase 6, non-selenocysteine containing phospholipid hydroperoxide glutathione peroxidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528854 349.00
Check your price
  • PRDX4 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFPA0009
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (1:2000)
Alternate names:
Thioredoxin-dependent peroxide reductase A0372; Thioredoxin peroxidase AO372; Peroxiredoxin IV; Antioxidant enzyme AOE372
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0009 375.00
Check your price
  • GPX3 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA522969
  • Size 100µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (0.5 µg/ml)
Alternate names:
Plasma glutathione peroxidase; Extracellular glutathione peroxidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522969 349.00
Check your price
  • HRP Monoclonal Antibody for Western Blot, ICC and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA110371
  • Size 100ug
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110371 159.00
Check your price
  • HRP Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # PA126409
  • Size 100µL
Applications
ELISA (1:16,000-1:64,000), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126409 349.00
Check your price
  • COX2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517614
  • Size 100µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Assay dependent), Western Blot (1:1000)
Alternate names:
Prostaglandin-endoperoxide synthase 2; Prostaglandin H2 synthase 2; PHS II; Cyclooxygenase-2; TIS10 protein; PES-2; Macrophage activation-associated marker protein P71/73; Gripghs; Glucocorticoid-regulated inflammatory cyclooxygenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517614 275.00
Check your price
  • SFPQ Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IP and RIA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA125325
  • Size 50µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (2-4 µg/ml)
Alternate names:
Polypyrimidine tract-binding protein-associated-splicing factor; DNA-binding p52/p100 complex, 100 kDa subunit; 100 kDa DNA-pairing protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125325 309.00
Check your price
  • COX1 Monoclonal Antibody for Western Blot, IF, ICC, IHC, Flow and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # 358100
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Prostaglandin-endoperoxide synthase 1; Prostaglandin H2 synthase 1; Cyclooxygenase-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
358100 307.00
Check your price
  • COX2 Monoclonal Antibody for Western Blot, IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 358200
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Prostaglandin-endoperoxide synthase 2; Prostaglandin H2 synthase 2; PHS II; Cyclooxygenase-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
358200 309.00
Check your price