1 - 30 of 30 products displayed
  • Phospho-PPP1R2 (Thr72) Polyclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 441160G
  • Size 10blots
Applications
Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Protein phosphatase inhibitor 2, IPP-2, PPP1R2, IPP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
441160G 309.00
Check your price
  • PPP1R11 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539367
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
T-complex testis expressed protein 5; Protein phosphatase inhibitor 3; Hemochromatosis candidate gene V protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539367 309.00
Check your price
  • Phospho-PPP1R2 (Ser120, Ser121) Polyclonal Antibody for IHC (P) and pep-ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538695
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), peptide-ELISA (1:5000)
Alternate names:
Protein phosphatase inhibitor 2, IPP-2, PPP1R2, IPP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538695 309.00
Check your price
  • PPP1R2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538696
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Protein phosphatase inhibitor 2, IPP-2, PPP1R2, IPP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538696 309.00
Check your price
  • PPP1R2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539805
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Protein phosphatase inhibitor 2, IPP-2, PPP1R2, IPP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539805 309.00
Check your price
  • Phospho-PPP1R2 (Ser121, Ser122) Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA540181
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
I-2, IPP-2, IPP2, Protein phosphatase inhibitor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540181 309.00
Check your price
  • PPP1R1A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537201
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Protein phosphatase inhibitor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537201 309.00
Check your price
  • LMTK2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519376
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Serine/threonine-protein kinase KPI-2; Lemur tyrosine kinase 2; Kinase/phosphatase/inhibitor 2; CDK5/p35-regulated kinase; Brain-enriched kinase; Apoptosis-associated tyrosine kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519376 349.00
Check your price
  • LMTK2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513935
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Serine/threonine-protein kinase KPI-2; Lemur tyrosine kinase 2; Kinase/phosphatase/inhibitor 2; CDK5/p35-regulated kinase; Brain-enriched kinase; Apoptosis-associated tyrosine kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513935 349.00
Check your price
  • LMTK2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513937
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Serine/threonine-protein kinase KPI-2; Lemur tyrosine kinase 2; Kinase/phosphatase/inhibitor 2; CDK5/p35-regulated kinase; Brain-enriched kinase; Apoptosis-associated tyrosine kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513937 349.00
Check your price
  • STYXL1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530426
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Map kinase phosphatase-like protein MK-STYX; Dual specificity protein phosphatase 24; Dual specificity phosphatase inhibitor MK-STYX
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530426 349.00
Check your price
  • PPP1R14D Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539368
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Gastrointestinal and brain-specific PP1-inhibitory protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539368 309.00
Check your price
  • AXIN1 Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710204
  • Size 100µg
Applications
Western Blot (2-5 ug/ml)
Alternate names:
Axis inhibition protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710204 309.00
Check your price
  • SET Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA141322
  • Size 100µg
Applications
Western Blot (1-2 µg/ml)
Alternate names:
Template-activating factor I; Phosphatase 2A inhibitor I2PP2A; PHAPII; Inhibitor of granzyme A-activated DNase; HLA-DR-associated protein II; Liver regeneration-related protein LRRGR00002
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141322 309.00
Check your price
  • SET Polyclonal Antibody for Western Blot and pep-ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517962
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Template-activating factor I; Phosphatase 2A inhibitor I2PP2A; PHAPII; Inhibitor of granzyme A-activated DNase; HLA-DR-associated protein II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517962 349.00
Check your price
  • Phospho-MYPT1 (Ser668) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517046
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Myosin phosphatase-targeting subunit 1; Protein phosphatase myosin-binding subunit; Serine/threonine protein phosphatase PP1 smooth muscle regulatory subunit M110; Protein phosphatase subunit 1M; MBSP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517046 309.00
Check your price
  • SET Polyclonal Antibody for Western Blot, IF, ICC, IHC and IP
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA544576
  • Size 100µl
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (1:500), Immunoprecipitation (1:500), Western Blot (0.2-1 µg/ml)
Alternate names:
Template-activating factor I; Phosphatase 2A inhibitor I2PP2A; PHAPII; Inhibitor of granzyme A-activated DNase; HLA-DR-associated protein II; Liver regeneration-related protein LRRGR00002
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544576 309.00
Check your price
  • SET Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131457
  • Size 50µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Template-activating factor I; Phosphatase 2A inhibitor I2PP2A; PHAPII; Inhibitor of granzyme A-activated DNase; HLA-DR-associated protein II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131457 399.00
Check your price
  • SET Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA129799
  • Size 200µL
Applications
Western Blot (1-2 µg/ml)
Alternate names:
Template-activating factor I; Phosphatase 2A inhibitor I2PP2A; PHAPII; Inhibitor of granzyme A-activated DNase; HLA-DR-associated protein II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129799 399.00
Check your price
  • SET Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521756
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Template-activating factor I; Phosphatase 2A inhibitor I2PP2A; PHAPII; Inhibitor of granzyme A-activated DNase; HLA-DR-associated protein II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521756 349.00
Check your price
  • Phospho-MYPT1 (Thr696) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A16679
  • Size 100µL
Applications
Western Blot (Assay Dependent)
Alternate names:
Protein phosphatase myosin-binding subunit; Myosin phosphatase-targeting subunit 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16679 309.00
Check your price
  • PPP1R1C Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539369
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Inhibitor-5 of protein phosphatase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539369 309.00
Check your price
  • MYPT1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Chicken, Hamster, Human, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA517164
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Myosin phosphatase-targeting subunit 1; Protein phosphatase myosin-binding subunit; PP1M subunit M110; 130 kDa myosin-binding subunit of smooth muscle myosin phosphatase; Serine/threonine protein phosphatase PP1 smooth muscle regulatory subunit M110; Protein phosphatase subunit 1M; MBSP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517164 275.00
Check your price
  • Phospho-MYPT1 (Thr696) Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538297
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Myosin phosphatase-targeting subunit 1; Protein phosphatase myosin-binding subunit; Serine/threonine protein phosphatase PP1 smooth muscle regulatory subunit M110; Protein phosphatase subunit 1M; MBSP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538297 309.00
Check your price
  • Phospho-MYPT1 (Thr853) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538299
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Myosin phosphatase-targeting subunit 1; Protein phosphatase myosin-binding subunit; Serine/threonine protein phosphatase PP1 smooth muscle regulatory subunit M110; Protein phosphatase subunit 1M; MBSP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538299 309.00
Check your price
  • MYPT1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA538298
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Myosin phosphatase-targeting subunit 1; Protein phosphatase myosin-binding subunit; Serine/threonine protein phosphatase PP1 smooth muscle regulatory subunit M110; Protein phosphatase subunit 1M; MBSP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538298 309.00
Check your price
  • MYPT1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538300
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Myosin phosphatase-targeting subunit 1; Protein phosphatase myosin-binding subunit; Serine/threonine protein phosphatase PP1 smooth muscle regulatory subunit M110; Protein phosphatase subunit 1M; MBSP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538300 309.00
Check your price
  • SH3RF2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521013
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
SH3 domain-containing RING finger protein 2; RING finger protein 158; Protein phosphatase 1 regulatory subunit 39; Heart protein phosphatase 1-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521013 349.00
Check your price
  • Phospho-MYPT1 (Thr853) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA540248
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Protein phosphatase myosin-binding subunit; Myosin phosphatase-targeting subunit 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540248 309.00
Check your price
  • PPME1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527754
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
PME-1, protein phosphatase methylesterase-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527754 349.00
Check your price