1 - 30 of 30 products displayed
  • PKA alpha/beta Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513737
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
cAMP-dependent protein kinase, alpha-catalytic subunit, PRKACA, PKACA, PKACB, PRKACB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513737 349.00
Check your price
  • PKA beta Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513794
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
cAMP-dependent protein kinase, beta-catalytic subunit; PKA C-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513794 349.00
Check your price
  • PKA alpha Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA130236
  • Size 100 µg
Applications
Immunoprecipitation (20-30 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
PKA, cAMP-Dependent Protein Kinase, Catalytic Subunit PKA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130236 425.00
Check your price
  • PKA alpha Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517626
  • Size 100 µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
PKA C-alpha; cAMP-dependent protein kinase catalytic subunit alpha; cAMP-dependent protein kinase catalytic subunit alpha, isoform 1; protein kinase A catalytic subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517626 275.00
Check your price
  • PKA alpha/beta Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538316
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
PRKC Pan, Protein kinase C alpha type, PKC-A, PKC-alpha, PRKCA, PKCA, PRKACA, PKCB, PRKCB2, Protein kinase C beta type, PKC-B, PKC-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538316 309.00
Check your price
  • Phospho-PKA alpha/beta (Thr197) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44988A
  • Size 10 blots
Applications
Western Blot (0.1-1.0 ug/ml)
Alternate names:
PRKC Pan, Protein kinase C alpha type, PKC-A, PKC-alpha, PRKCA, PKCA, PRKACA, PKCB, PRKCB2, Protein kinase C beta type, PKC-B, PKC-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
44988A 309.00
Check your price
  • Phospho-PKA alpha/beta (Thr197) Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA128490
  • Size 50 µL
Applications
Western Blot (0.1-0.75 µg/ml)
Alternate names:
PRKC Pan, Protein kinase C alpha type, PKC-A, PKC-alpha, PRKCA, PKCA, PRKACA, PKCB, PRKCB1, Protein kinase C beta type, PKC-B, PKC-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128490 349.00
Check your price
  • Phospho-PKA beta (Ser338) Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Pig
  • Conjugate Unconjugated
  • Catalog # PA126733
  • Size 50 µL
Applications
Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126733 349.00
Check your price
  • PKA beta Antibody (Monoclonal, 2G8-1D12)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA519698
  • Size 100 µg
Applications
ELISA (Assay-Dependent)
Alternate names:
PRKC Pan, Protein kinase C alpha type, PKC-A, PKC-alpha, PRKCA, PKCA, PRKACA, PKCB, PRKCB1, Protein kinase C beta type, PKC-B, PKC-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA519698 309.00
Check your price
  • PKA alpha Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA528160
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
PKACA, PKA C-alpha, cAMP-dependent protein kinase catalytic subunit alpha, protein kinase A catalytic subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528160 349.00
Check your price
  • Phospho-PKA alpha (Thr197) Antibody (Monoclonal, D45D3)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A16696
  • Size 100 µL
Applications
Western Blot (Assay Dependent)
Alternate names:
PKACA, PKA C-alpha, cAMP-dependent protein kinase catalytic subunit alpha, protein kinase A catalytic subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16696 309.00
Check your price
  • Phospho-PKA beta (Ser338) Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 44992G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
cAMP-dependent protein kinase, beta-catalytic subunit, PKA C-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
44992G 309.00
Check your price
  • PKA alpha Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521842
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
PKACA, PKA C-alpha, cAMP-dependent protein kinase catalytic subunit alpha, protein kinase A catalytic subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521842 349.00
Check your price
  • Phospho-PKA 2 beta pSer114 Antibody
  • Host Rabbit
  • Target Species Rat, Bovine, Human, Mouse, Rabbit
  • Conjugate
  • Catalog # PA126735
  • Size 50µL
Applications
Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126735 359.00
Check your price
  • PKA gamma Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528148
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
KAPG, PKACg, PKA C-gamma, cAMP-dependent protein kinase catalytic subunit gamma, serine(threonine) protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528148 349.00
Check your price
  • PKA alpha Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528604
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
PKACA, PKA C-alpha, cAMP-dependent protein kinase catalytic subunit alpha, protein kinase A catalytic subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528604 349.00
Check your price
  • PKA alpha Antibody (Monoclonal, 6D2.1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119732
  • Size 100 µg
Applications
Dot blot (Assay-dependent), Western Blot (0.5-1 µg/ml)
Alternate names:
MGC102831, MGC48865, PKACA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119732 309.00
Check your price
  • Adenylate Kinase 5 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Goat, Human, Mouse, Sheep, Rabbit, Rat, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA544218
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
ATP-AMP transphosphorylase 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544218 309.00
Check your price
  • Adenylate Kinase 5 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA515422
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
ATP-AMP transphosphorylase 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515422 349.00
Check your price
  • PKA gamma Antibody (Monoclonal, 2E4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517158
  • Size 100 µL
Applications
Flow Cytometry (1:200-1:400), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517158 309.00
Check your price
  • Phospho-PKC alpha/beta-2 (Thr638, Thr641) Antibody
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517904
  • Size 100 µL
Applications
Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
PRKC Pan, Protein kinase C alpha type, PKC-A, PKC-alpha, PRKCA, PKCA, PRKACA, PKCB, PRKCB2, Protein kinase C beta type, PKC-B, PKC-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517904 309.00
Check your price
  • PRKAR2B Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA528266
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
PRKAR2, RII-BETA, H_RG363E19.2, WUGSC:H_RG363E19.2, cAMP-dependent protein kinase type II-beta regulatory chain, cAMP-dependent protein kinase type II-beta regulatory subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528266 349.00
Check your price
  • PRKAR2B Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513799
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
cAMP-dependent protein kinase type II-beta regulatory chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513799 349.00
Check your price
  • PRKAR1B Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513798
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
cAMP-dependent protein kinase type I-beta regulatory chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513798 349.00
Check your price
  • PRKAR1B Antibody
  • Host Sheep
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA547592
  • Size 100 µg
Applications
Western Blot (1-10 µg/ml)
Alternate names:
cAMP-dependent protein kinase type I-beta regulatory chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547592 349.00
Check your price
  • PRKAR2A Antibody (Monoclonal, H2 006 D4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA3517
  • Size 100 µL
Applications
Western Blot (1:1,000)
Alternate names:
Protein Kinase A Type 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3517 349.00
Check your price
  • Phospho-PRKAR2B (Ser113) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA537787
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
PRKAR2, RII-BETA, H_RG363E19.2, WUGSC:H_RG363E19.2, cAMP-dependent protein kinase type II-beta regulatory chain, cAMP-dependent protein kinase type II-beta regulatory subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537787 309.00
Check your price
  • Phospho-PRKAR2B (Ser113) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA538508
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
PRKAR2, RII-BETA, H_RG363E19.2, WUGSC:H_RG363E19.2, cAMP-dependent protein kinase type II-beta regulatory chain, cAMP-dependent protein kinase type II-beta regulatory subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538508 309.00
Check your price
  • PRKAR1A Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515368
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Tissue-specific extinguisher 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515368 349.00
Check your price
  • Phospho-MYLK (Ser1760) Antibody
  • Host Rabbit
  • Target Species Bovine, Mouse, Rabbit
  • Conjugate Unconjugated
  • Catalog # 441085G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
Telokin
Catalog # Size Price (AUD) Your Price (AUD) Qty
441085G 304.00
Check your price