1 - 30 of 30 products displayed
  • PKC alpha Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529228
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
AAG6, PKC-alpha, PKCA, PRKACA, PKC-A, aging-associated gene 6, protein kinase C alpha type
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529228 349.00
Check your price
  • Phospho-PKC alpha (Tyr657) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537691
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
EC 2.7.11.13, KPCA, PKC III, PKC-A, PKC-III, PKC-alpha, PRKACA, PRKCA, Protein kinase C, alpha type, kinase PKC-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537691 309.00
Check your price
  • PKC alpha Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA522160
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
AAG6, PKC-alpha, PKCA, PRKACA, PKC-A, aging-associated gene 6, protein kinase C alpha type
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522160 349.00
Check your price
  • Phospho-PKC alpha/beta-2 (Thr638, Thr641) Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517904
  • Size 100µL
Applications
Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
PRKC Pan, Protein kinase C alpha type, PKC-A, PKC-alpha, PRKCA, PKCA, PRKACA, PKCB, PRKCB2, Protein kinase C beta type, PKC-B, PKC-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517904 309.00
Check your price
  • PKC alpha Monoclonal Antibody for Western Blot and IHC (F)
  • Host Mouse
  • Target Species Mammal
  • Conjugate Unconjugated
  • Catalog # MA125286
  • Size 1mL
Applications
Immunohistochemistry (Frozen) (1:10), Western Blot (1:10-1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125286 349.00
Check your price
  • PKC alpha Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA1157
  • Size 100µg
Applications
Immunocytochemistry (1:50-1:100), Immunofluorescence (1:50-1:100), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (5 µg), Western Blot (1:1000-1:2000)
Alternate names:
protein kinase C alpha, PKCA, PKC-alpha, PRKACA, AAG6, EC 2.7.11.13, aging-associated gene 6, protein kinase C alpha type
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1157 349.00
Check your price
  • Phospho-PKC alpha (Thr638) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44962G
  • Size 100µL
Applications
Western Blot (Assay Dependent)
Alternate names:
EC 2.7.11.13, KPCA, PKC III, PKC-A, PKC-III, PKC-alpha, PRKACA, PRKCA, Protein kinase C, alpha type, kinase PKC-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
44962G 309.00
Check your price
  • PKC alpha Polyclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513739
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Protein kinase C, alpha type; PRKCA; PKCA; PRKACA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513739 349.00
Check your price
  • PKC alpha Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517551
  • Size 100µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
PKC-A; aging-associated gene 6; protein kinase C alpha type
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517551 275.00
Check your price
  • PKC alpha Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFPA0175
  • Size 100µL
Applications
Western Blot (1:2000)
Alternate names:
FHC, FTH, FTHL6, MGC104426, PIG15, PLIF
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0175 309.00
Check your price
  • PKC Pan Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536757
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
nPKC-zeta; nPKC-eta; PKC-L; nPKC-epsilon; nPKC-delta; nPKC-theta; Tyrosine-protein kinase PRKCD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536757 349.00
Check your price
  • Phospho-PKC Pan (Thr497) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538418
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
nPKC-zeta; nPKC-eta; PKC-L; nPKC-epsilon; nPKC-delta; nPKC-theta; Tyrosine-protein kinase PRKCD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538418 309.00
Check your price
  • Phospho-PKC gamma (Thr514) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44956
  • Size 10blots
Applications
Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (0.1-1 µg/ml)
Alternate names:
PRKC Pan, Protein kinase C alpha type, PKC-A, PKC-alpha, PRKCA, PKCA, PRKACA, PKCB, PRKCB1, Protein kinase C beta type, PKC-B, PKC-beta, nPKC-delta, PRKCD, Sphingosine-dependent protein kinase-1, SDK-1, Tyrosine-protein kinase PRKCD, Protein kinase C delta type, Protein kinase C epsilon type, nPKC-epsilon, PRKCE, PKCE, Protein kinase C gamma type, PKC-gamma, PRKCG, PKCG, Protein kinase C eta type, PKC-L, nPKC-eta, PRKCH, PKCL, PRKCL, Protein kinase C iota type, PRKCI, DXS1179E, Protein kinase C theta type, nPKC-theta, PRKCQ, PRKCT, Protein kinase C zeta type, nPKC-zeta, PRKCZ, PKC2
Catalog # Size Price (AUD) Your Price (AUD) Qty
44956 309.00
Check your price
  • Phospho-PKC gamma (Thr655) Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44965
  • Size 10blots
Applications
Flow Cytometry (3-5 µg/million cells), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (0.1-1.0 ug/ml)
Alternate names:
PRKC Pan, Protein kinase C alpha type, PKC-A, PKC-alpha, PRKCA, PKCA, PRKACA, PKCB, PRKCB1, Protein kinase C beta type, PKC-B, PKC-beta, nPKC-delta, PRKCD, Sphingosine-dependent protein kinase-1, SDK-1, Tyrosine-protein kinase PRKCD, Protein kinase C delta type, Protein kinase C epsilon type, nPKC-epsilon, PRKCE, PKCE, Protein kinase C gamma type, PKC-gamma, PRKCG, PKCG, Protein kinase C eta type, PKC-L, nPKC-eta, PRKCH, PKCL, PRKCL, Protein kinase C iota type, PRKCI, DXS1179E, Protein kinase C theta type, nPKC-theta, PRKCQ, PRKCT, Protein kinase C zeta type, nPKC-zeta, PRKCZ, PKC2
Catalog # Size Price (AUD) Your Price (AUD) Qty
44965 309.00
Check your price
  • PKC gamma Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 133800
  • Size 200µg
Applications
Immunohistochemistry (Assay Dependent), Western Blot (2-5 ug/ml)
Alternate names:
PRKC Pan, Protein kinase C alpha type, PKC-A, PKC-alpha, PRKCA, PKCA, PRKACA, PKCB, PRKCB1, Protein kinase C beta type, PKC-B, PKC-beta, nPKC-delta, PRKCD, Sphingosine-dependent protein kinase-1, SDK-1, Tyrosine-protein kinase PRKCD, Protein kinase C delta type, Protein kinase C epsilon type, nPKC-epsilon, PRKCE, PKCE, Protein kinase C gamma type, PKC-gamma, PRKCG, PKCG, Protein kinase C eta type, PKC-L, nPKC-eta, PRKCH, PKCL, PRKCL, Protein kinase C iota type, PRKCI, DXS1179E, Protein kinase C theta type, nPKC-theta, PRKCQ, PRKCT, Protein kinase C zeta type, nPKC-zeta, PRKCZ, PKC2
Catalog # Size Price (AUD) Your Price (AUD) Qty
133800 304.00
Check your price
  • Phospho-PKC zeta (Thr560) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA538419
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
nPKC-zeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538419 309.00
Check your price
  • Phospho-PKC gamma (Thr674) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44975G
  • Size 10blots
Applications
Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (Assay Dependent)
Alternate names:
PRKC Pan, Protein kinase C alpha type, PKC-A, PKC-alpha, PRKCA, PKCA, PRKACA, PKCB, PRKCB1, Protein kinase C beta type, PKC-B, PKC-beta, nPKC-delta, PRKCD, Sphingosine-dependent protein kinase-1, SDK-1, Tyrosine-protein kinase PRKCD, Protein kinase C delta type, Protein kinase C epsilon type, nPKC-epsilon, PRKCE, PKCE, Protein kinase C gamma type, PKC-gamma, PRKCG, PKCG, Protein kinase C eta type, PKC-L, nPKC-eta, PRKCH, PKCL, PRKCL, Protein kinase C iota type, PRKCI, DXS1179E, Protein kinase C theta type, nPKC-theta, PRKCQ, PRKCT, Protein kinase C zeta type, nPKC-zeta, PRKCZ, PKC2
Catalog # Size Price (AUD) Your Price (AUD) Qty
44975G 309.00
Check your price
  • Phospho-PKC beta (Thr641) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537692
  • Size 100µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Protein kinase C beta type, PKC-B, PKC-beta, PRKCB, PKCB, PRKCB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537692 309.00
Check your price
  • Phospho-PKC eta (Thr655) Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44969
  • Size 10blots
Applications
Flow Cytometry (3-5 µg/million cells), Western Blot (0.1-0.75 ug/ml)
Alternate names:
nPKC-eta; PKC-L
Catalog # Size Price (AUD) Your Price (AUD) Qty
44969 309.00
Check your price
  • Phospho-PKC theta (Ser676) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537696
  • Size 100µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
nPKC-theta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537696 309.00
Check your price
  • Phospho-PKC theta (Ser695) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537697
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
nPKC-theta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537697 309.00
Check your price
  • Phospho-PKC mu (Ser738, Ser742) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538386
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
nPKC-D2; nPKC-mu; nPKC-D1; Protein kinase D; Protein kinase C mu type; nPKC-nu; Protein kinase C nu type; Protein kinase EPK2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538386 309.00
Check your price
  • Phospho-PKC mu (Ser205) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538420
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
nPKC-mu; nPKC-D1; Protein kinase D; Protein kinase C mu type
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538420 309.00
Check your price
  • Phospho-PKC mu (Ser910) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538388
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
nPKC-mu; nPKC-D1; Protein kinase D; Protein kinase C mu type
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538388 309.00
Check your price
  • PKC mu Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538387
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
nPKC-mu; nPKC-D1; Protein kinase D; Protein kinase C mu type
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538387 309.00
Check your price
  • PKA alpha/beta Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538316
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
PRKC Pan, Protein kinase C alpha type, PKC-A, PKC-alpha, PRKCA, PKCA, PRKACA, PKCB, PRKCB2, Protein kinase C beta type, PKC-B, PKC-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538316 309.00
Check your price
  • Phospho-PKA alpha/beta (Thr197) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44988A
  • Size 10blots
Applications
Western Blot (0.1-1.0 ug/ml)
Alternate names:
PRKC Pan, Protein kinase C alpha type, PKC-A, PKC-alpha, PRKCA, PKCA, PRKACA, PKCB, PRKCB2, Protein kinase C beta type, PKC-B, PKC-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
44988A 309.00
Check your price
  • Phospho-PKA alpha/beta (Thr197) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA128490
  • Size 50µL
Applications
Western Blot (0.1-0.75 µg/ml)
Alternate names:
PRKC Pan, Protein kinase C alpha type, PKC-A, PKC-alpha, PRKCA, PKCA, PRKACA, PKCB, PRKCB1, Protein kinase C beta type, PKC-B, PKC-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128490 349.00
Check your price
  • PKC mu Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536113
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
nPKC-D2; nPKC-mu; nPKC-D1; Protein kinase D; Protein kinase C mu type; nPKC-nu; Protein kinase C nu type; Protein kinase EPK2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536113 309.00
Check your price
  • Phospho-PKC delta/theta (Ser643, Ser676) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA517905
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (1:1000)
Alternate names:
nPKC-delta; nPKC-theta; Tyrosine-protein kinase PRKCD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517905 309.00
Check your price