1 - 3 of 3 products displayed
  • LNPEP Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523777
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Placental leucine aminopeptidase; Oxytocinase; Insulin-responsive aminopeptidase; Insulin-regulated membrane aminopeptidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523777 349.00
Check your price
  • LNPEP Polyclonal Antibody for Western Blot, IF and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534458
  • Size 100 ug
Applications
Immunofluorescence (20 ug/mL), Immunohistochemistry (20 ug/ml), Western Blot (1 ug/mL)
Alternate names:
Placental leucine aminopeptidase; Oxytocinase; Insulin-responsive aminopeptidase; Insulin-regulated membrane aminopeptidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534458 349.00
Check your price
  • LNPEP Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A16604
  • Size 100 µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Placental leucine aminopeptidase; Oxytocinase; Insulin-responsive aminopeptidase; Insulin-regulated membrane aminopeptidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16604 Not Orderable Online
Check your price