1 - 30 of 30 products displayed
  • POLR2A Antibody (Monoclonal, 4H8)
  • Host Mouse
  • Target Species Human, Mouse, Yeast
  • Conjugate Unconjugated
  • Catalog # MA146093
  • Size 100 µg
Applications
ChIP assay (1-10 ul), ELISA (1:100-1:2000), Immunocytochemistry (1:50-1:100), Immunofluorescence (1:50-1:100), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (1:10-500), Western Blot (1:1000-1:20000)
Alternate names:
DNA-directed RNA polymerase III largest subunit; DNA-directed RNA polymerase II subunit A; RNA-directed RNA polymerase II subunit RPB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146093 375.00
Check your price
  • POLR2A Antibody (Monoclonal)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523510
  • Size 50 µg
Applications
ChIP assay (1-2 µg), ChIP-sequencing (Assay dependant), Immunocytochemistry (1:500), Immunofluorescence (1:500), peptide-ELISA (1:3000), Western Blot (1:1000)
Alternate names:
RNA-directed RNA polymerase II subunit RPB1; DNA-directed RNA polymerase III largest subunit; DNA-directed RNA polymerase II subunit A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523510 309.00
Check your price
  • POLR2A Antibody (Monoclonal)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 491033
  • Size 100 µL
Applications
ChIP assay (2-3 ul)
Alternate names:
RNA-directed RNA polymerase II subunit RPB1; DNA-directed RNA polymerase III largest subunit; DNA-directed RNA polymerase II subunit A
Catalog # Size Price (AUD) Your Price (AUD) Qty
491033 309.00
Check your price
  • POLR2A Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528087
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
RNA-directed RNA polymerase II subunit RPB1; DNA-directed RNA polymerase III largest subunit; DNA-directed RNA polymerase II subunit A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528087 325.00
Check your price
  • Phospho-POLR2A (Ser5) Antibody (Monoclonal, 4H8)
  • Host Mouse
  • Target Species C. elegans, Fruit fly, Human, Mouse, Non-human primate, Plant, Yeast
  • Conjugate Unconjugated
  • Catalog # MA518089
  • Size 100 µL
Applications
ChIP assay (Assay-dependent), ELISA (Assay-dependent), Flow Cytometry (Assay-dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (Assay-dependent)
Alternate names:
DNA-directed RNA polymerase III largest subunit; DNA-directed RNA polymerase II subunit A; RNA-directed RNA polymerase II subunit RPB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518089 425.00
Check your price
  • Phospho-POLR2A (Ser5) Antibody (Monoclonal, 8A7)
  • Host Mouse
  • Target Species Human, Yeast
  • Conjugate Unconjugated
  • Catalog # MA518090
  • Size 100 µL
Applications
ELISA (Assay-dependent), Flow Cytometry (Assay-dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (Assay-dependent)
Alternate names:
RNA-directed RNA polymerase II subunit RPB1; DNA-directed RNA polymerase III largest subunit; DNA-directed RNA polymerase II subunit A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518090 425.00
Check your price
  • Phospho-POLR2A (Ser1619) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536723
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
DNA-directed RNA polymerase III largest subunit; DNA-directed RNA polymerase II subunit A; RNA-directed RNA polymerase II subunit RPB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536723 325.00
Check your price
  • POLR2A Antibody (Monoclonal, 8WG16)
  • Host Mouse
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # MA110882
  • Size 200 ug
Applications
Western Blot (Assay dependent)
Alternate names:
DNA-directed RNA polymerase III largest subunit; DNA-directed RNA polymerase II subunit A; RNA-directed RNA polymerase II subunit RPB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110882 375.00
Check your price
  • POLR2A Antibody (Monoclonal, 8WG16)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA126249
  • Size 100 µg
Applications
ChIP assay (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
DNA-directed RNA polymerase III largest subunit; DNA-directed RNA polymerase II subunit A; RNA-directed RNA polymerase II subunit RPB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126249 349.00
Check your price
  • Phospho-POLR2A (Ser1619) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA538091
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
DNA-directed RNA polymerase III largest subunit; DNA-directed RNA polymerase II subunit A; RNA-directed RNA polymerase II subunit RPB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538091 309.00
Check your price
  • Phospho-POLR2A (Ser2, Ser5) Antibody
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # PA517563
  • Size 100 µL
Applications
ChIP assay (1-3µl), Immunohistochemistry (Paraffin) (1:25), Western Blot (1:1000)
Alternate names:
DNA-directed RNA polymerase III largest subunit; DNA-directed RNA polymerase II subunit A; RNA-directed RNA polymerase II subunit RPB1; DNA polymerase II subunit A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517563 309.00
Check your price
  • Phospho-POLR2A (Ser5) Antibody (Monoclonal, 8A7)
  • Host Mouse
  • Target Species Human, Yeast
  • Conjugate Unconjugated
  • Catalog # MA15875
  • Size 50 µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
RNA-directed RNA polymerase II subunit RPB1; DNA-directed RNA polymerase III largest subunit; DNA-directed RNA polymerase II subunit A; DNA polymerase II subunit A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15875 375.00
Check your price
  • FCP1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528074
  • Size 100 µL
Applications
ChIP assay (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
TFIIF-associating CTD phosphatase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528074 349.00
Check your price
  • FCP1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530302
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
TFIIF-associating CTD phosphatase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530302 325.00
Check your price
  • Cdc73 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526189
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Hyperparathyroidism 2 protein; Cell division cycle protein 73 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526189 325.00
Check your price
  • Phospho-LEO1 (Ser551) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512993
  • Size 400 µL
Applications
Dot blot (1:500)
Alternate names:
Replicative senescence down-regulated leo1-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512993 325.00
Check your price
  • INTS10 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526920
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
C8orf35, INT10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526920 325.00
Check your price
  • Cdc73 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517158
  • Size 100 µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Hyperparathyroidism 2 protein homolog; Cell division cycle protein 73 homolog; Hyperparathyroidism 2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517158 299.00
Check your price
  • Cdc73 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517159
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Hyperparathyroidism 2 protein homolog; Cell division cycle protein 73 homolog; Hyperparathyroidism 2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517159 299.00
Check your price
  • CPSF3L Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535035
  • Size 400 µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (1:1000)
Alternate names:
Protein related to CPSF subunits of 68 kDa; Cleavage and polyadenylation-specific factor 3-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535035 325.00
Check your price
  • SND1 Antibody (Monoclonal, 2D7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515420
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
p100 co-activator; Tudor domain-containing protein 11; EBNA2 coactivator p100; 100 kDa coactivator
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515420 325.00
Check your price
  • Phospho-LEO1 (Ser10) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513010
  • Size 400 µL
Applications
Dot blot (1:500)
Alternate names:
Replicative senescence down-regulated leo1-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513010 325.00
Check your price
  • LEO1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511820
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Replicative senescence down-regulated leo1-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511820 325.00
Check your price
  • Cyclin L1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536070
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Cyclin Ania-6a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536070 309.00
Check your price
  • Phospho-CDK7 (Thr170) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538452
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
TFIIH basal transcription factor complex kinase subunit; Protein-tyrosine kinase MPK-7; Cell division protein kinase 7; CR4 protein kinase; CDK-activating kinase; 39 kDa protein kinase; Serine/threonine-protein kinase 1; CDK-activating kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538452 309.00
Check your price
  • CDK7 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA534791
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
TFIIH basal transcription factor complex kinase subunit; Protein-tyrosine kinase MPK-7; Cell division protein kinase 7; CR4 protein kinase; CDK-activating kinase; 39 kDa protein kinase; Serine/threonine-protein kinase 1; CDK-activating kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534791 349.00
Check your price
  • SETD2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534934
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
p231HBP; SET domain-containing protein 2; Lysine N-methyltransferase 3A; Huntingtin-interacting protein B; Huntingtin-interacting protein 1; Huntingtin yeast partner B; HIF-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534934 349.00
Check your price
  • SETD2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534935
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
p231HBP; SET domain-containing protein 2; Lysine N-methyltransferase 3A; Huntingtin-interacting protein B; Huntingtin-interacting protein 1; Huntingtin yeast partner B; HIF-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534935 349.00
Check your price
  • SETD2 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA543071
  • Size 100 µl
Applications
Immunohistochemistry (4-8 µg/ml), Western Blot (Assay-Dependent)
Alternate names:
SET domain-containing protein 2; Lysine N-methyltransferase 3A; p231HBP; Huntingtin-interacting protein B; Huntingtin-interacting protein 1; Huntingtin yeast partner B; HIF-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543071 309.00
Check your price
  • CDK7 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124252
  • Size 100 µg
Applications
Immunohistochemistry (Assay-dependent), Western Blot (1 µg/ml)
Alternate names:
TFIIH basal transcription factor complex kinase subunit; Serine/threonine-protein kinase 1; Cell division protein kinase 7; CDK-activating kinase 1; 39 kDa protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124252 349.00
Check your price