1 - 2 of 2 products displayed
  • PROSC Antibody (Monoclonal, 2G1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA521300
  • Size 100 µg
Applications
ELISA (Assay-Dependent)
Alternate names:
proline synthase co-transcribed bacterial homolog protein, proline synthetase co-transcribed (bacterial homolog), proline synthetase co-transcribed bacterial homolog protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA521300 309.00
Check your price
  • PROSC Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA532036
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
proline synthase co-transcribed bacterial homolog protein, proline synthetase co-transcribed (bacterial homolog), proline synthetase co-transcribed bacterial homolog protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532036 349.00
Check your price