1 - 30 of 30 products displayed
  • Caspase 9 (active) Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA126434
  • Size 50µg
Applications
Immunocytochemistry (10-20 µg/ml), Immunofluorescence (10-20 ug/ml), Immunoprecipitation (10-20 µg/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126434 399.00
Check your price
  • active Caspase 9 Polyclonal Antibody for Western Blot, IF, ICC
  • Host Rabbit
  • Target Species Rat
  • Conjugate
  • Catalog # PA126435
  • Size 50µg
Applications
Immunocytochemistry (10-20 µg/ml), Immunofluorescence (10-20 ug/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
RP11-265F14.3, APAF-3, APAF3, CASPASE-9c, ICE-LAP6, MCH6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126435 Not Orderable Online
Check your price
  • HIV1 Protease Monoclonal Antibody for Western Blot, ELISA, DB and FN
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA119015
  • Size 100µg
Applications
Dot blot (0.2-2 ug/ml), ELISA (Assay Dependent), Functional Assay (Assay-Dependent), Western Blot (0.5µg/mL)
Alternate names:
Human immunodeficiency virus, HIV protease, HIV1gp1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119015 309.00
Check your price
  • HGF Activator Polyclonal Antibody for Western Blot, IHC (F) and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547187
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
HGFA, Hgfac, HGF activator, HGFA, HGFAC
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547187 349.00
Check your price
  • HGF Activator Polyclonal Antibody for Western Blot, IHC and IP
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547147
  • Size 100µg
Applications
Immunohistochemistry (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
HGFA, Hgfac, HGF activator, HGFA, HGFAC
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547147 349.00
Check your price
  • Caspase 7 (active) Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA126433
  • Size 50µg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (10-20 µg/ml), Immunoprecipitation (10-20 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Cysteine protease LICE2; Apoptotic protease Mch-3; ICE-like apoptotic protease 3; CMH-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126433 399.00
Check your price
  • Caspase 3 (active/pro) Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191637
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (2-3 µg/ml)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191637 425.00
Check your price
  • APAF1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532266
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
APAF-1, CED4, apoptotic protease-activating factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532266 275.00
Check your price
  • NLRC4 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520014
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (1-2 µg/ml)
Alternate names:
Ice protease-activating factor; Caspase recruitment domain-containing protein 12; CARD, LRR, and NACHT-containing protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520014 349.00
Check your price
  • Phospho-Caspase 9 (Thr125) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536710
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:500-1:1000)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536710 349.00
Check your price
  • Phospho-Caspase 9 (Tyr153) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536020
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536020 349.00
Check your price
  • Phospho-Caspase 9 (Ser144) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538437
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538437 309.00
Check your price
  • Phospho-Caspase 9 (Tyr153) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538436
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538436 309.00
Check your price
  • Caspase 9 Monoclonal Antibody for Western Blot, IF, ICC, IHC and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA116842
  • Size 200µL
Applications
Immunocytochemistry (1:50-1:250), Immunofluorescence (1:50-1:250), Western Blot (1:500-1:1000)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116842 349.00
Check your price
  • NLRC4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527686
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Ice protease-activating factor; Caspase recruitment domain-containing protein 12; CARD, LRR, and NACHT-containing protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527686 349.00
Check your price
  • NLRC4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14264
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Ice protease-activating factor; Caspase recruitment domain-containing protein 12; CARD, LRR, and NACHT-containing protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14264 349.00
Check your price
  • Caspase 9 Polyclonal Antibody, Biotin conjugate for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Sheep, Rat
  • Conjugate Biotin
  • Catalog # PA516355
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (5-10 µg/mL), Immunoprecipitation (10 µg/ml), Western Blot (2.5-5.0 µg/ml)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516355 399.00
Check your price
  • Caspase 9 (Cleaved Asp315) Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517913
  • Size 100µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517913 309.00
Check your price
  • Caspase 9 (Cleaved Asp353) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA517605
  • Size 100µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Western Blot (1:1000)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517605 309.00
Check your price
  • Caspase 9 (Cleaved Asp330) Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517911
  • Size 100µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517911 309.00
Check your price
  • Caspase 9 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA112562
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112562 309.00
Check your price
  • Phospho-Caspase 9 (Ser196) Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512558
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512558 349.00
Check your price
  • Phospho-Caspase 9 (Thr125) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537505
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537505 309.00
Check your price
  • Phospho-Caspase 9 (Ser196) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA540222
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540222 309.00
Check your price
  • Caspase 9 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA141415
  • Size 200µL
Applications
Western Blot (1:250-1:1000)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141415 349.00
Check your price
  • Caspase 9 Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519904
  • Size 100ug
Applications
Immunocytochemistry (2 µg/ml), Immunofluorescence (20 ug/ml), Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519904 349.00
Check your price
  • Caspase 9 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519903
  • Size 100ug
Applications
Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519903 349.00
Check your price
  • Caspase 9 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), IP and HA
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # PA516358
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (5-10 µg/mL), Immunoprecipitation (10µg/mg protein lysate), Western Blot (2.5-5.0 µg/ml)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516358 349.00
Check your price
  • Caspase 9 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112506
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112506 375.00
Check your price
  • Caspase 9 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112505
  • Size 100µg
Applications
Western Blot (0.25-0.5µg/ml)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112505 325.00
Check your price