1 - 2 of 2 products displayed
  • HEY2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA525807
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Protein gridlock homolog; Hairy-related transcription factor 2; Hairy and enhancer of split-related protein 2; HES-related repressor protein 2; Class B basic helix-loop-helix protein 32; Cardiovascular helix-loop-helix factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525807 349.00
Check your price
  • HEY2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534957
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Protein gridlock homolog; Hairy-related transcription factor 2; Hairy and enhancer of split-related protein 2; HES-related repressor protein 2; Class B basic helix-loop-helix protein 32; Cardiovascular helix-loop-helix factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534957 349.00
Check your price