1 - 30 of 30 products displayed
  • p70 S6 Kinase Oligoclonal Antibody for Western Blot, IF, ICC, Flow and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710285
  • Size 100µg
Applications
ELISA (Assay-Dependent), Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:100-1:1000)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710285 304.00
Check your price
  • Phospho-p70 S6 Kinase (Thr421, Ser424) Monoclonal Antibody for Western Blot, Flow and ChIP
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 701083
  • Size 100µg
Applications
ChIP assay (1 ul), Flow Cytometry (1:100-1:500), Western Blot (1:500-1:5000)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
701083 Contact Us ›
Check your price
  • LCK Monoclonal Antibody for Western Blot, IF, ICC, Flow, IP and FN
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate
  • Catalog # AHO0472
  • Size 500µl
Applications
Flow Cytometry (Assay-Dependent), Functional Assay (Assay Dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (Assay-Dependent), Immunoprecipitation (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHO0472 Not Orderable Online
Check your price
  • CDK5 Monoclonal Antibody for Western Blot, IF, Flow and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA511291
  • Size 500µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunofluorescence (Assay Dependent), Immunoprecipitation (6 ug/mg protein lysate), Western Blot (1-2 µg/ml)
Alternate names:
Tau protein kinase II catalytic subunit; Serine/threonine-protein kinase PSSALRE; Cell division protein kinase 5; CR6 protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511291 349.00
Check your price
  • CDK5 Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # AHZ0492
  • Size 500µl
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (Assay-Dependent), Immunoprecipitation (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
Tau protein kinase II catalytic subunit; Serine/threonine-protein kinase PSSALRE; Cell division protein kinase 5; CR6 protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHZ0492 303.00
Check your price
  • ERK2 Polyclonal Antibody for Western Blot, IHC (P), IP and ChIP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121016
  • Size 500µL
Applications
ChIP assay (Assay-Dependent), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121016 349.00
Check your price
  • CDK1 Monoclonal Antibody for Western Blot, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA119057
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (10µg/ml), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p34 protein kinase; Cell division protein kinase 1; Cell division control protein 2 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119057 199.00
Check your price
  • GSK3 beta Monoclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Mouse
  • Target Species Human, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA17621
  • Size 100ug
Applications
ELISA (1:1000), Immunohistochemistry (Assay Dependent), Immunoprecipitation (5-10 µg), Western Blot (Assay-Dependent)
Alternate names:
Xgsk-3 protein; Serine/threonine-protein kinase GSK3B; Factor A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17621 375.00
Check your price
  • CaMKII alpha Monoclonal Antibody for Western Blot, IF, IHC and IP
  • Host Mouse
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1048
  • Size 100µL
Applications
Immunofluorescence (1:100), Immunohistochemistry (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (0.5-1 µg/mL)
Alternate names:
Calcium/calmodulin-dependent protein kinase type 2, CaMKII
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1048 375.00
Check your price
  • CDK4 Monoclonal Antibody for Western Blot, IF, Flow and IP
  • Host Mouse
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA512984
  • Size 500µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunofluorescence (Assay Dependent), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
PSK-J3; Cell division protein kinase 4; CRK3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512984 349.00
Check your price
  • TTK Monoclonal Antibody for Western Blot, IF, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 359100
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Phosphotyrosine picked threonine-protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
359100 303.00
Check your price
  • ERK1/ERK2 Oligoclonal Antibody for Western Blot and ChIP
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 710214
  • Size 100µg
Applications
ChIP assay (1 ul), Western Blot (1-2 ug/ml)
Alternate names:
Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1; p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2
Catalog # Size Price (AUD) Your Price (AUD) Qty
710214 309.00
Check your price
  • CaMKII alpha Monoclonal Antibody for Western Blot, IHC, IHC (P), IP and ELISA
  • Host Mouse
  • Target Species Fish, Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # 137300
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, calcium/calmodulin-dependent protein kinase type II alpha chain, Calci
Catalog # Size Price (AUD) Your Price (AUD) Qty
137300 304.00
Check your price
  • Aurora A Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132191
  • Size 100ug
Applications
ELISA (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Serine/threonine-protein kinase aurora-A; Serine/threonine-protein kinase 6; Serine/threonine-protein kinase 15; Breast tumor-amplified kinase; Aurora/IPL1-related kinase 1; Aurora 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132191 399.00
Check your price
  • PKN1 Monoclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA519703
  • Size 100µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Immunoprecipitation (Assay-Dependent), Western Blot (1-5 µg/ml), Western Blot (Assay-Dependent)
Alternate names:
Serine-threonine protein kinase N; Protein-kinase C-related kinase 1; Protein kinase PKN-alpha; Protein kinase C-like PKN; Protein kinase C-like 1; Protease-activated kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA519703 309.00
Check your price
  • CDK9 Polyclonal Antibody for Western Blot, IHC (F), IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129427
  • Size 50µL
Applications
ELISA (1:10,000-1:50,000), Immunohistochemistry (Frozen) (1:200-1:1000), Immunohistochemistry (Paraffin) (1:200-1:1000), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:3000)
Alternate names:
Cell division protein kinase 9; Tat-associated kinase complex catalytic subunit; Serine/threonine-protein kinase PITALRE; Cell division cycle 2-like protein kinase 4; C-2K
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129427 375.00
Check your price
  • CDK6 Monoclonal Antibody for Western Blot, IF, IHC (F) and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA513338
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Serine/threonine-protein kinase PLSTIRE; Cell division protein kinase 6; CR2 protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513338 349.00
Check your price
  • CDK1 Monoclonal Antibody for Western Blot, IF, IHC (F), IHC (P), Flow, IP and ELISA
  • Host Mouse
  • Target Species Amphibian, Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 331800
  • Size 200µg
Applications
ELISA (0.1-1.0 ug/ml), Flow Cytometry (3-5 µg/million cells), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (5-10 ug/ml), Immunohistochemistry (Paraffin) (1:10-1:50), Immunoprecipitation (5 ug), Western Blot (0.5-2 µg/ml)
Alternate names:
p34 protein kinase; Cell division protein kinase 1; Cell division control protein 2 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
331800 305.00
Check your price
  • CDK6 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # AHZ0232
  • Size 500µl
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (1:10-1:200), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunoprecipitation (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
Serine/threonine-protein kinase PLSTIRE; Cell division protein kinase 6; CR2 protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHZ0232 306.00
Check your price
  • ATM Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow, IP, ChIP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA123152
  • Size 100µl
Applications
ChIP assay (Assay Dependent), ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:500-1:3000)
Alternate names:
Ataxia telangiectasia mutated; Ataxia telangiectasia mutated homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123152 349.00
Check your price
  • PLK1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 377000
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Serine/threonine-protein kinase 13; Polo-like kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
377000 305.00
Check your price
  • CDK4 Monoclonal Antibody, Biotin conjugate for Western Blot, IF and IP
  • Host Mouse
  • Target Species Mouse, Pig, Rat, Human
  • Conjugate Biotin
  • Catalog # MA512981
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
PSK-J3; Cell division protein kinase 4; CRK3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512981 399.00
Check your price
  • CDK4 Monoclonal Antibody for Western Blot, IF, Flow and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA513720
  • Size 500µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunofluorescence (Assay Dependent), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
PSK-J3; Cell division protein kinase 4; CRK3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513720 349.00
Check your price
  • CDK4 Monoclonal Antibody for Western Blot, IF and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA112299
  • Size 100µg
Applications
Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
PSK-J3; Cell division protein kinase 4; CRK3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112299 309.00
Check your price
  • RSK2/RSK3 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA517212
  • Size 100µL
Applications
ELISA (Assay Dependent), Western Blot (2 µg/ml)
Alternate names:
pp90RSK3; Ribosomal S6 kinase 3; MAP kinase-activated protein kinase 1c; 90 kDa ribosomal protein S6 kinase 2; pp90RSK2; Ribosomal S6 kinase 2; MAP kinase-activated protein kinase 1b; Insulin-stimulated protein kinase 1; 90 kDa ribosomal protein S6 kinase 3; Protein-tyrosine kinase Mpk-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517212 349.00
Check your price
  • Aurora B Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA517226
  • Size 100µL
Applications
ELISA (Assay Dependent), Western Blot (2 µg/ml)
Alternate names:
Serine/threonine-protein kinase aurora-B; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase 12; STK-1; Aurora/IPL1-related kinase 2; Aurora- and IPL1-like midbody-associated protein 1; Aurora 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517226 349.00
Check your price
  • CDK7 Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517210
  • Size 100µL
Applications
ELISA (Assay Dependent), Immunoprecipitation (2 µg), Western Blot (0.2 µg/ml)
Alternate names:
TFIIH basal transcription factor complex kinase subunit; Serine/threonine-protein kinase 1; Cell division protein kinase 7; CDK-activating kinase 1; 39 kDa protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517210 349.00
Check your price
  • CDK7 Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517209
  • Size 100µL
Applications
ELISA (Assay Dependent), Immunoprecipitation (2 µg), Western Blot (0.2 µg/ml)
Alternate names:
TFIIH basal transcription factor complex kinase subunit; Serine/threonine-protein kinase 1; Cell division protein kinase 7; CDK-activating kinase 1; 39 kDa protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517209 349.00
Check your price
  • CDK7 Polyclonal Antibody for Western Blot, IP and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA129162
  • Size 500µL
Applications
ChIP assay (1-3 ul), Immunoprecipitation (1:1000), Western Blot (1:1000)
Alternate names:
TFIIH basal transcription factor complex kinase subunit; Serine/threonine-protein kinase 1; Cell division protein kinase 7; CDK-activating kinase 1; 39 kDa protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129162 349.00
Check your price
  • EphA4 Monoclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Mouse
  • Target Species Chicken, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 371600
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tyrosine-protein kinase receptor SEK; Tyrosine-protein kinase TYRO1; EPH-like kinase 8; Tyrosine-protein kinase receptor SEK-1; Tyrosine-protein kinase receptor MPK-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
371600 304.00
Check your price