1 - 30 of 30 products displayed
  • Golgi protein 58k Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IM
  • Host Mouse
  • Target Species Bovine, Dog, Hamster, Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA122144
  • Size 100 µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:250), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay dependent), Western Blot (1:5000)
Alternate names:
LCHC1; Formiminotransferase-cyclodeaminase; 58 kDa microtubule-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122144 399.00
Check your price
  • Nuclear Matrix Protein p84 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IP and ChIP
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA123261
  • Size 100 µg
Applications
ChIP assay (Assay Dependent), Immunocytochemistry (0.5-2 ug/ml), Immunofluorescence (0.5-2 µg/ml), Immunohistochemistry (Assay Dependent), Immunoprecipitation (1:100-1:500), Western Blot (0.3-2 µg/ml)
Alternate names:
Nuclear matrix protein p84; hTREX84
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123261 309.00
Check your price
  • XPC Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA123328
  • Size 100 µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Paraffin) (1/100), Western Blot (1/1000)
Alternate names:
p125; Xeroderma pigmentosum group C-complementing protein; Xeroderma pigmentosum group C-complementing protein homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123328 309.00
Check your price
  • GFAP Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516367
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Glial Fibrillary Acidic Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516367 499.00
Check your price
  • LC3A/LC3B Polyclonal Antibody, DyLight® 650 conjugate for Western Blot, IHC, IHC (P) and IP
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Rat, Zebrafish
  • Conjugate DyLight® 650
  • Catalog # PA522730
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:200-1:400), Immunoprecipitation (20 µg/500 µg of protein), Western Blot (2 µg/ml)
Alternate names:
Microtubule-associated protein 1 light chain 3 alpha; MAP1A/MAP1B light chain 3 A; MAP1 light chain 3-like protein 1; Autophagy-related ubiquitin-like modifier LC3 A; Autophagy-related protein LC3 A; Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522730 375.00
Check your price
  • LC3A/LC3B Polyclonal Antibody, DyLight® 550 conjugate for Western Blot, IHC, IHC (P) and IP
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Rat, Zebrafish
  • Conjugate DyLight® 550
  • Catalog # PA522733
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:200-1:400), Immunoprecipitation (20 µg/500 µg of protein), Western Blot (2 µg/ml)
Alternate names:
Microtubule-associated protein 1 light chain 3 alpha; MAP1A/MAP1B light chain 3 A; MAP1 light chain 3-like protein 1; Autophagy-related ubiquitin-like modifier LC3 A; Autophagy-related protein LC3 A; Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522733 375.00
Check your price
  • LC3A/LC3B Polyclonal Antibody, DyLight® 488 conjugate for Western Blot, IHC, IHC (P) and IP
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Rat, Zebrafish
  • Conjugate DyLight® 488
  • Catalog # PA522731
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:200-1:400), Immunoprecipitation (20 µg/500 µg of protein), Western Blot (2 µg/ml)
Alternate names:
Microtubule-associated protein 1 light chain 3 alpha; MAP1A/MAP1B light chain 3 A; MAP1 light chain 3-like protein 1; Autophagy-related ubiquitin-like modifier LC3 A; Autophagy-related protein LC3 A; Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522731 375.00
Check your price
  • LC3B Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ChIP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 700712
  • Size 100 µg
Applications
ChIP assay (1 ul), Flow Cytometry (3-5 ug x 10^6 cells), Immunocytochemistry (1-3 ug/ml), Immunofluorescence (1-3 ug/ml), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (1-5 ug/ml)
Alternate names:
Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
700712 303.00
Check your price
  • KPNA2 Polyclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Goat
  • Target Species Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA132094
  • Size 100 ug
Applications
ELISA (Assay-dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (0.1-0.5 µg/ml)
Alternate names:
SRP1-alpha; RAG cohort protein 1; Pore targeting complex 58 kDa subunit; Pendulin; Karyopherin subunit alpha-2; Importin alpha P1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132094 399.00
Check your price
  • S100 Monoclonal Antibody for Western Blot, IF, IHC (Free), IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA122734
  • Size 200 µg
Applications
ELISA (Assay dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry-Free Floating (Assay-Dependent), Immunohistochemistry (Paraffin) (Assay dependent), Western Blot (Assay dependent)
Alternate names:
S100 calcium-binding protein A1; S-100 protein subunit alpha; S-100 protein alpha chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122734 349.00
Check your price
  • Myogenin Monoclonal Antibody for Western Blot, IF, IHC (P), Flow, IP and GS
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA511658
  • Size 500 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Gel Shift (1mg/mL), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:50-1:100), Immunoprecipitation (3 ug/mg protein lysate), Western Blot (1-2 ug/ml)
Alternate names:
Myogenic factor 4; Class C basic helix-loop-helix protein 3; MYOD1-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511658 309.00
Check your price
  • PDIA2 Oligoclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710081
  • Size 100 µg
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Pancreas-specific protein disulfide isomerase
Catalog # Size Price (AUD) Your Price (AUD) Qty
710081 309.00
Check your price
  • LC3A/LC3B Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P), Flow, IP and ELISA
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA116931
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:300), Immunofluorescence (1:100-1:300), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (1:200-1:400), Immunoprecipitation (20 µg/500 µg of lysate), Western Blot (2 µg/ml)
Alternate names:
Microtubule-associated protein 1 light chain 3 alpha; MAP1A/MAP1B light chain 3 A; MAP1 light chain 3-like protein 1; Autophagy-related ubiquitin-like modifier LC3 A; Autophagy-related protein LC3 A; Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116931 375.00
Check your price
  • LC3B Polyclonal Antibody, DyLight® 650 conjugate for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate DyLight® 650
  • Catalog # PA522937
  • Size 100 µL
Applications
Immunocytochemistry (1:2000), Immunofluorescence (1:2000), Immunohistochemistry (1:2000), Western Blot (1:3000)
Alternate names:
Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522937 375.00
Check your price
  • AGR2/AGR3 Monoclonal Antibody for Western Blot, ICC and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110047
  • Size 100 ug
Applications
Immunocytochemistry (1 ug/ml), Immunohistochemistry (Paraffin) (5 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Protein disulfide isomerase family A, member 18; Breast cancer membrane protein 11; Anterior gradient 3 homolog; Secreted cement gland protein XAG-2 homolog; HPC8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110047 309.00
Check your price
  • Prohibitin Monoclonal Antibody, Biotin conjugate for Western Blot, IF and IHC (P)
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Pig, Rat
  • Conjugate Biotin
  • Catalog # MA512855
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (2-4 µg/ml), Western Blot (0.5-1.0 µg/ml)
Alternate names:
Repressor of estrogen receptor activity; D-prohibitin; B-cell receptor-associated protein BAP37; B-cell receptor-associated protein 32
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512855 399.00
Check your price
  • Prohibitin Monoclonal Antibody for Western Blot, IF and IHC (P)
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA512858
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (2-4 µg/ml), Western Blot (0.5-1.0 µg/ml)
Alternate names:
Repressor of estrogen receptor activity; D-prohibitin; B-cell receptor-associated protein BAP37; B-cell receptor-associated protein 32
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512858 349.00
Check your price
  • PTK7 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529839
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Tyrosine-protein kinase-like 7; Pseudo tyrosine kinase receptor 7; Protein-tyrosine kinase 7; Colon carcinoma kinase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529839 349.00
Check your price
  • Actin Monoclonal Antibody for Western Blot, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Dog, Chicken, Cat, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA110046
  • Size 100 ug
Applications
Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (1 ug/ml)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Gamma-actin; Alpha-cardiac actin; Alpha-actin; Alpha-actin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110046 249.00
Check your price
  • 14.3.3 Pan Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA512242
  • Size 500 µL
Applications
Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
SEZ-2; Protein kinase C inhibitor protein 1; Stratifin; Protein 1054; Epithelial cell marker protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512242 349.00
Check your price
  • NEFM Monoclonal Antibody for Western Blot, ICC and IHC (P)
  • Host Mouse
  • Target Species Mammal
  • Conjugate Unconjugated
  • Catalog # MA119225
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Neurofilament triplet M protein; 160 kDa neurofilament protein; Neurofilament 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119225 199.00
Check your price
  • Factor XIII A Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA516423
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (10-20 µg/mL), Western Blot (5-10 µg/mL)
Alternate names:
Transglutaminase A chain; Protein-glutamine gamma-glutamyltransferase A chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516423 309.00
Check your price
  • Cdc73 Polyclonal Antibody for Western Blot, IP and ChIP
  • Host Rabbit
  • Target Species Fruit fly, Human
  • Conjugate Unconjugated
  • Catalog # PA540082
  • Size 50 µg
Applications
ChIP assay (1-5 µg), Immunoprecipitation (12 µg), Western Blot (1:1000)
Alternate names:
Hyperparathyroidism 2 protein; Cell division cycle protein 73 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540082 309.00
Check your price
  • PGP9.5 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532567
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
Ubiquitin thioesterase L1; PGP 9.5; Neuron cytoplasmic protein 9.5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532567 275.00
Check your price
  • PGP9.5 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA516825
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Ubiquitin thioesterase L1; PGP 9.5; Neuron cytoplasmic protein 9.5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516825 309.00
Check your price
  • MYOD Monoclonal Antibody for IF, IP and IM
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA512907
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunomicroscopy (Assay Dependent), Immunoprecipitation (2 ug/ml)
Alternate names:
Myogenic factor 3; Class C basic helix-loop-helix protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512907 309.00
Check your price
  • p73 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA123147
  • Size 100 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Paraffin) (Assay-dependent), Immunoprecipitation (Assay dependent), Western Blot (1:500)
Alternate names:
p53-related protein; p53-like transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123147 399.00
Check your price
  • CENPF Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA123185
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:500)
Alternate names:
Mitosin; Kinetochore protein CENPF; AH antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123185 309.00
Check your price
  • 14-3-3 sigma Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511663
  • Size 500 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1 µg/mL)
Alternate names:
Stratifin; Epithelial cell marker protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511663 349.00
Check your price
  • GFAP Monoclonal Antibody, Alexa Fluor® 594 conjugate for IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate Alexa Fluor® 594
  • Catalog # A21295
  • Size 50 µL
Applications
Immunocytochemistry (1-10 ug/ml), Immunofluorescence (Assay Dependent)
Alternate names:
Glial fibrillary acidic protein, GFAP, GFAP
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21295 326.00
Check your price