1 - 3 of 3 products displayed
  • LCN1 Oligoclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710406
  • Size 100 µg
Applications
Immunocytochemistry (1 ug/ml), Immunofluorescence (1 ug/ml), Western Blot (2-3 ug/ml)
Alternate names:
Von Ebner gland protein; Tear prealbumin; Tear lipocalin
Catalog # Size Price (AUD) Your Price (AUD) Qty
710406 309.00
Check your price
  • LCN1 Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515501
  • Size 100 µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Von Ebner gland protein; Tear prealbumin; Tear lipocalin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515501 349.00
Check your price
  • LCN1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530077
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Von Ebner gland protein; Tear prealbumin; Tear lipocalin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530077 349.00
Check your price