1 - 30 of 30 products displayed
  • Phospho-p70 S6 Kinase (Thr389) Antibody (Monoclonal, R.566.2)
  • Host Mouse
  • Target Species Fruit fly, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515202
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515202 309.00
Check your price
  • Phospho-ERK1/ERK2 (Thr202, Tyr204) Antibody (Monoclonal, S.812.9)
  • Host Rabbit
  • Target Species Fruit fly, Hamster, Human, Mink, Mouse, Non-human primate, Pig, Rat, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA515173
  • Size 200 µL
Applications
ChIP assay (1-3 µl), Immunohistochemistry (Paraffin) (1:400), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2; Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1; Protein rolled; Extracellular-regulated kinase A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515173 309.00
Check your price
  • Phospho-ERK1/ERK2 (Tyr204) Antibody (Monoclonal, B.742.5)
  • Host Rabbit
  • Target Species Fruit fly, Human, Mink, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA515174
  • Size 200 µL
Applications
Flow Cytometry (1:200), Immunocytochemistry (1:200), Immunofluorescence (1:200), Western Blot (1:1000)
Alternate names:
p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2; Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1; Protein rolled; Extracellular-regulated kinase A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515174 309.00
Check your price
  • ERK1/ERK2 Antibody (Monoclonal, K.913.4)
  • Host Rabbit
  • Target Species Bovine, Dog, C. elegans, Chicken, Fruit fly, Hamster, Human, Mink, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA515134
  • Size 200 µL
Applications
Flow Cytometry (1:100), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:250), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2; Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1; Protein rolled; Extracellular-regulated kinase A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515134 275.00
Check your price
  • Phospho-p70 S6 Kinase (Thr421, Ser424) Antibody (Monoclonal, B12H16L8)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 701083
  • Size 100 µg
Applications
ChIP assay (1 ul), Flow Cytometry (1:100-1:500), Western Blot (1:500-1:5000)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
701083 Contact Us ›
Check your price
  • Phospho-p70 S6 Kinase (Thr389) Antibody (Oligoclonal, B2HCLC)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 710095
  • Size 100 µg
Applications
ChIP assay (1 ul), Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:5000)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710095 309.00
Check your price
  • p70 S6 Kinase Antibody (Oligoclonal, 15HCLC)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710285
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:100-1:1000)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710285 304.00
Check your price
  • Phospho-p70 S6 Kinase (Thr389) Antibody (Monoclonal, B2H9L2)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701064
  • Size 100 µg
Applications
Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:5000)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
701064 303.00
Check your price
  • p70 S6 Kinase Antibody (Monoclonal, 15H5L2)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 701261
  • Size 100 µg
Applications
Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:5000)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
701261 309.00
Check your price
  • Human FGFR2 Recombinant Protein
  • Host E. coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP10919
  • Size 25 µg
Applications
Control (Assay Dependent)
Alternate names:
FGF-10
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP10919 307.00
Check your price
  • c-Abl Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532281
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
p150; Proto-oncogene c-Abl; Abelson tyrosine-protein kinase 1; Abelson murine leukemia viral oncogene homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532281 275.00
Check your price
  • c-Abl Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137196
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
p150; Proto-oncogene c-Abl; Abelson tyrosine-protein kinase 1; Abelson murine leukemia viral oncogene homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137196 375.00
Check your price
  • LCK Antibody (Monoclonal, 3A5)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # AHO0472
  • Size 500µl
Applications
Flow Cytometry (Assay-Dependent), Functional Assay (Assay Dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (Assay-Dependent), Immunoprecipitation (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHO0472 304.00
Check your price
  • LCK Antibody (Monoclonal, 3A5)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA512303
  • Size 500 µL
Applications
Flow Cytometry (1ug/test), Immunocytochemistry (1:20), Immunofluorescence (1:20), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512303 309.00
Check your price
  • Phospho-LCK (Tyr505) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44850G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
44850G 309.00
Check your price
  • CSK Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA119417
  • Size 100 µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Protein-tyrosine kinase CYL; C-Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA119417 225.00
Check your price
  • LCK Antibody (Monoclonal, 33D196)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA141203
  • Size 200 µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA141203 375.00
Check your price
  • Phospho-HER-2 / ErbB2 pTyr1112 Antibody Polyclonal (PA5-35806)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate
  • Catalog # PA535806
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
p185erbB2; Tyrosine kinase-type cell surface receptor HER2; Proto-oncogene c-ErbB-2; Proto-oncogene Neu; Metastatic lymph node gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535806 339.00
Check your price
  • FAK Antibody
  • Host Rabbit
  • Target Species Amphibian, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA516676
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Western Blot (5-10 µg/mL)
Alternate names:
pp125FAK; p125FAK; Protein-tyrosine kinase 2; Focal adhesion kinase-related nonkinase; Protein phosphatase 1 regulatory subunit 71
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516676 309.00
Check your price
  • Phospho-FAK (Tyr576) Antibody
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # 44652G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
pp125FAK; p125FAK; Protein-tyrosine kinase 2; Focal adhesion kinase-related nonkinase; Protein phosphatase 1 regulatory subunit 71
Catalog # Size Price (AUD) Your Price (AUD) Qty
44652G 304.00
Check your price
  • Phospho-FAK (Tyr577) Antibody
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Xenopus
  • Conjugate Unconjugated
  • Catalog # 44614G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
pp125FAK; p125FAK; Protein-tyrosine kinase 2; Focal adhesion kinase-related nonkinase; Protein phosphatase 1 regulatory subunit 71
Catalog # Size Price (AUD) Your Price (AUD) Qty
44614G 309.00
Check your price
  • Phospho-LCK (Tyr394) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA537628
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537628 309.00
Check your price
  • Phospho-LCK (Ser158) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44842G
  • Size 100 µL
Applications
Western Blot (Assay Dependent)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
44842G 309.00
Check your price
  • Phospho-LCK (Tyr192) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44844G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
44844G 309.00
Check your price
  • FAK Antibody (Oligoclonal, 5HCLC)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710119
  • Size 100 µg
Applications
Flow Cytometry (2-4µg x 10^6 cells), Immunocytochemistry (2-4µg), Immunofluorescence (2-4µg), Western Blot (1-3µg/ml)
Alternate names:
pp125FAK; p125FAK; Protein-tyrosine kinase 2; Protein phosphatase 1 regulatory subunit 71; Focal adhesion kinase-related nonkinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
710119 309.00
Check your price
  • FAK Antibody (Monoclonal, 5H18L19)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701094
  • Size 100 µg
Applications
Flow Cytometry (2-4µg x 10^6 cells), Immunocytochemistry (2-4µg), Immunofluorescence (2-4µg), Western Blot (1-3 ug/ml)
Alternate names:
pp125FAK; p125FAK; Protein-tyrosine kinase 2; Protein phosphatase 1 regulatory subunit 71; Focal adhesion kinase-related nonkinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
701094 309.00
Check your price
  • FRK Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534782
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Protein-tyrosine kinase 5; Nuclear tyrosine protein kinase RAK; FYN-related kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534782 349.00
Check your price
  • FRK Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528573
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Protein-tyrosine kinase 5; Nuclear tyrosine protein kinase RAK; FYN-related kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528573 349.00
Check your price
  • FRK Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528373
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Protein-tyrosine kinase 5; Nuclear tyrosine protein kinase RAK; FYN-related kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528373 349.00
Check your price
  • Phospho-p70 S6 Kinase pThr389 / pThr412 Antibody Polyclonal (PA5-35701)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate
  • Catalog # PA535701
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535701 339.00
Check your price