1 - 30 of 30 products displayed
  • Rabbit anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 755 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Sheep
  • Conjugate DyLight® 755
  • Catalog # SA510059
  • Size 500ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510059 160.00
Check your price
  • Rabbit anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 680 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Sheep
  • Conjugate DyLight® 680
  • Catalog # SA510058
  • Size 500ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510058 158.00
Check your price
  • Rabbit anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 650 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Sheep
  • Conjugate DyLight® 650
  • Catalog # SA510057
  • Size 500ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510057 156.00
Check your price
  • Rabbit anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 550 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Sheep
  • Conjugate DyLight® 550
  • Catalog # SA510055
  • Size 500µg
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510055 156.00
Check your price
  • Rabbit anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 800 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Sheep
  • Conjugate DyLight® 800
  • Catalog # SA510060
  • Size 500µg
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510060 158.00
Check your price
  • Rabbit anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 594 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Sheep
  • Conjugate DyLight® 594
  • Catalog # SA510056
  • Size 500µg
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510056 158.00
Check your price
  • Rabbit anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 488 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Sheep
  • Conjugate DyLight® 488
  • Catalog # SA510054
  • Size 500ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510054 157.00
Check your price
  • Rabbit anti-Sheep IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Sheep
  • Conjugate HRP
  • Catalog # 31480
  • Size 1.5mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31480 149.00
Check your price
  • Rabbit anti-Sheep IgG (H+L) Secondary Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Sheep
  • Conjugate Unconjugated
  • Catalog # 31240
  • Size 2mg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31240 136.00
Check your price
  • Rabbit anti-Sheep IgG (H+L) Secondary Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Rabbit
  • Target Species Sheep
  • Conjugate Biotin
  • Catalog # 31840
  • Size 1.5mg
Applications
ELISA (1:20,000-1:400,000), Flow Cytometry (1:200-1:1,000), Immunocytochemistry (1:500-1:5,000), Immunofluorescence (1:500-1:5,000), Immunohistochemistry (1:500-1:5,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:20,000-1:400,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31840 195.00
Check your price
  • Rabbit anti-Sheep IgG (H+L) Secondary Antibody, FITC conjugate for IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Sheep
  • Conjugate FITC
  • Catalog # 31627
  • Size 1.5mg
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31627 113.00
Check your price
  • Rabbit anti-Sheep IgG (H+L) Secondary Antibody, AP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Sheep
  • Conjugate AP
  • Catalog # 31360
  • Size 1mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:50,000), Immunofluorescence (1:5,000-1:50,000), Immunohistochemistry (1:5,000-1:50,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:50,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31360 181.00
Check your price
  • Rabbit anti-Sheep IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Rabbit
  • Target Species Sheep
  • Conjugate HRP
  • Catalog # 818620
  • Size 1.5mg
Applications
ELISA (1:200-1:20,000), Immunohistochemistry (1:200-1:20,000), Western Blot (1:200-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
818620 149.00
Check your price
  • Rabbit anti-Sheep IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IF, IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Sheep
  • Conjugate HRP
  • Catalog # 618620
  • Size 1mL
Applications
ELISA (1:2,000-1:4,000), Immunofluorescence (1:2,000-1:4,000), Immunohistochemistry (Paraffin) (1:2,000-1:4,000), Immunoprecipitation (1:2,000-1:4,000), Western Blot (1:2,000-1:4,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
618620 149.00
Check your price
  • Rabbit anti-Sheep IgG (H+L) Secondary Antibody, FITC conjugate for IF and Flow
  • Host Rabbit
  • Target Species Sheep
  • Conjugate
  • Catalog # 818611
  • Size 1.5mg
Applications
Flow Cytometry (1:50 to 1:500), Immunofluorescence (1:50 to 1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
818611 Not Orderable Online
Check your price
  • Rabbit anti-Sheep IgG (H+L) Secondary Antibody, AP conjugate for ELISA
  • Host Rabbit
  • Target Species Sheep
  • Conjugate AP
  • Catalog # 618622
  • Size 1mL
Applications
ELISA (1:1,000), Immunofluorescence (1:1,000), Immunohistochemistry (Paraffin) (1:1,000), Immunoprecipitation (1:1,000), Western Blot (1:1,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
618622 139.00
Check your price
  • Donkey anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 488 conjugate for IF, ICC and Flow
  • Host Donkey
  • Target Species Sheep
  • Conjugate Alexa Fluor® 488
  • Catalog # A11015
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11015 209.00
Check your price
  • Donkey anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 594 conjugate for IF, ICC and Flow
  • Host Donkey
  • Target Species Sheep
  • Conjugate Alexa Fluor® 594
  • Catalog # A11016
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11016 209.00
Check your price
  • Donkey anti-Sheep IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Donkey
  • Target Species Sheep
  • Conjugate HRP
  • Catalog # A16047
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16047 179.00
Check your price
  • Donkey anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 680 conjugate for Western Blot, IF and ICC
  • Host Donkey
  • Target Species Sheep
  • Conjugate Alexa Fluor® 680
  • Catalog # A21102
  • Size 500µL
Applications
Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL), Western Blot (0.04-0.2 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21102 209.00
Check your price
  • Donkey anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 647 conjugate for IF, ICC and IHC
  • Host Donkey
  • Target Species Sheep
  • Conjugate Alexa Fluor® 647
  • Catalog # A21448
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21448 209.00
Check your price
  • Donkey anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 633 conjugate for IF, ICC and IHC
  • Host Donkey
  • Target Species Sheep
  • Conjugate Alexa Fluor® 633
  • Catalog # A21100
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21100 209.00
Check your price
  • Donkey anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 568 conjugate for IF, ICC and IHC
  • Host Donkey
  • Target Species Sheep
  • Conjugate Alexa Fluor® 568
  • Catalog # A21099
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21099 209.00
Check your price
  • Donkey anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 555 conjugate for IF, ICC and IHC
  • Host Donkey
  • Target Species Sheep
  • Conjugate Alexa Fluor® 555
  • Catalog # A21436
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21436 209.00
Check your price
  • Donkey anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 546 conjugate for IF, ICC and IHC
  • Host Donkey
  • Target Species Sheep
  • Conjugate Alexa Fluor® 546
  • Catalog # A21098
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21098 209.00
Check your price
  • Donkey anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 350 conjugate for IF, ICC and IHC
  • Host Donkey
  • Target Species Sheep
  • Conjugate Alexa Fluor® 350
  • Catalog # A21097
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21097 209.00
Check your price
  • Donkey anti-Sheep IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Donkey
  • Target Species Sheep
  • Conjugate Biotin
  • Catalog # A16052
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16052 158.00
Check your price
  • Donkey anti-Sheep IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, TRITC conjugate for IF
  • Host Donkey
  • Target Species Sheep
  • Conjugate TRITC
  • Catalog # A16053
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16053 136.00
Check your price
  • Donkey anti-Sheep IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, FITC conjugate for IF and Flow
  • Host Donkey
  • Target Species Sheep
  • Conjugate FITC
  • Catalog # A16049
  • Size 1mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16049 139.00
Check your price
  • Donkey anti-Sheep IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Donkey
  • Target Species Sheep
  • Conjugate AP
  • Catalog # A16051
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16051 200.00
Check your price