1 - 30 of 30 products displayed
  • Ki-67 Antibody (Monoclonal, SP6)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514520
  • Size 500 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunohistochemistry (Paraffin) (1:100-200)
Alternate names:
antigen KI-67, proliferation-related Ki-67 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514520 375.00
Check your price
  • Ki-67 Antibody (Monoclonal, 12H15 L5)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701198
  • Size 100 µg
Applications
ChIP assay (1 ul), Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500)
Alternate names:
antigen KI-67, proliferation-related Ki-67 antigen, RP11-380J17.2, KIA
Catalog # Size Price (AUD) Your Price (AUD) Qty
701198 303.00
Check your price
  • CD56 Antibody (Monoclonal, 3H15L12)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 701379
  • Size 100 µg
Applications
Flow Cytometry (1 ug/ml X 10^6 cells), Immunocytochemistry (1 ug/ml), Immunofluorescence (1 ug/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
701379 303.00
Check your price
  • CD99 Antibody (Monoclonal, SP119)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516416
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
T-cell surface glycoprotein E2; Protein MIC2; E2 antigen; 120000000
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516416 499.00
Check your price
  • Aggrecan Antibody (Monoclonal, BC-3)
  • Host Mouse
  • Target Species Bovine, Dog, Horse, Cat, Guinea pig, Human, Sheep, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA316888
  • Size 100 µg
Applications
ELISA (1:1000-1:5000), Immunofluorescence (1:100-1:500), Immunohistochemistry (Frozen) (1:10-1:2000), Immunohistochemistry (Paraffin) (1:10-1:2000), Western Blot (1:500-1:1000)
Alternate names:
Cartilage-specific proteoglycan core protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA316888 309.00
Check your price
  • SERCA1 ATPase Antibody (Monoclonal, VE121G9)
  • Host Mouse
  • Target Species Amphibian, Dog, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3912
  • Size 100 µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (1:20), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (1:500-1:5000)
Alternate names:
Endoplasmic reticulum class 1/2 Ca(2+) ATPase; Calcium-transporting ATPase sarcoplasmic reticulum type, fast twitch skeletal muscle isoform; Calcium pump 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3912 375.00
Check your price
  • SERCA1 ATPase Antibody (Monoclonal, IIH11)
  • Host Mouse
  • Target Species Dog, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3911
  • Size 100 µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (Frozen) (1:20-200), Western Blot (1:500-1:5000)
Alternate names:
Endoplasmic reticulum class 1/2 Ca(2+) ATPase; Calcium-transporting ATPase sarcoplasmic reticulum type, fast twitch skeletal muscle isoform; Calcium pump 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3911 375.00
Check your price
  • Dihydropyridine Receptor alpha-1 Antibody (Monoclonal, 1A)
  • Host Mouse
  • Target Species Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3920
  • Size 200 µL
Applications
Flow Cytometry (1ug/test), Immunohistochemistry (Frozen) (1:200), Immunohistochemistry (Paraffin) (1:20), Immunoprecipitation (Assay dependent), Western Blot (1:500)
Alternate names:
Voltage-gated calcium channel subunit alpha Cav1.1; Calcium channel, L type, alpha-1 polypeptide, isoform 3, skeletal muscle
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3920 349.00
Check your price
  • Calsequestrin Antibody (Monoclonal, VIIID12)
  • Host Mouse
  • Target Species Dog, Chicken, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3913
  • Size 100 µL
Applications
Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (1:20), Western Blot (1:1,000)
Alternate names:
Laminin-binding protein; Calsequestrin, skeletal muscle isoform; Aspartactin; Calmitine
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3913 375.00
Check your price
  • CACNA2D1 Antibody (Monoclonal, 20A)
  • Host Mouse
  • Target Species Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3921
  • Size 100 µL
Applications
Flow Cytometry (1:100-1:200), Immunocytochemistry (1:250), Immunofluorescence (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (1:500), Western Blot (1:500)
Alternate names:
Voltage-gated calcium channel subunit alpha-2/delta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3921 375.00
Check your price
  • Sarcalumenin Antibody (Monoclonal, XIIC4)
  • Host Mouse
  • Target Species Dog, Guinea pig, Mouse, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA3932
  • Size 100 µL
Applications
Immunofluorescence (1/500), Western Blot (1:500)
Alternate names:
SRL
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3932 349.00
Check your price
  • Triadin Antibody (Monoclonal, GE 4.90)
  • Host Mouse
  • Target Species Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3927
  • Size 200 µL
Applications
ELISA (Assay dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (1:50), Immunoprecipitation (Assay dependent), Western Blot (1:500)
Alternate names:
TRDN, TRISK
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3927 349.00
Check your price
  • Triadin Antibody (Monoclonal, IIG12)
  • Host Mouse
  • Target Species Mouse, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA3931
  • Size 100 µL
Applications
Immunofluorescence (1/1000), Immunohistochemistry (1:500), Western Blot (1:1,000)
Alternate names:
TRDN, TRISK
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3931 349.00
Check your price
  • CD4 Antibody (Monoclonal, KEN-4)
  • Host Mouse
  • Target Species Rabbit
  • Conjugate Unconjugated
  • Catalog # MA181754
  • Size 100 µg
Applications
Flow Cytometry (1:25-1:200), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
T-cell surface antigen T4/Leu-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181754 309.00
Check your price
  • Sodium/Calcium Exchanger Antibody (Monoclonal, 6H2)
  • Host Mouse
  • Target Species Rabbit
  • Conjugate Unconjugated
  • Catalog # MA112751
  • Size 10 Miniblots
Applications
Western Blot (1:1000)
Alternate names:
Sodium/Calcium Exchanger 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112751 425.00
Check your price
  • JSRP1 Antibody (Monoclonal, VF1c)
  • Host Mouse
  • Target Species Dog, Guinea pig, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3933
  • Size 100 µL
Applications
Immunofluorescence (1:250), Western Blot (1:1,500)
Alternate names:
Junctional SR
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3933 349.00
Check your price
  • CD98 Antibody (Monoclonal, 44D7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516514
  • Size 200 ug
Applications
Flow Cytometry (1:50-1:200), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
Solute carrier family 3 member 2; Lymphocyte activation antigen 4F2 large subunit; 4F2 heavy chain antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516514 375.00
Check your price
  • CD98 Antibody (Monoclonal, 44D7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA134546
  • Size 100 µg
Applications
Flow Cytometry (1:50-1:200 (10 µl for 10^6 cells in 100 µl)), Immunohistochemistry (Frozen) (Assay dependent), Immunoprecipitation (Assay dependent)
Alternate names:
Solute carrier family 3 member 2; Lymphocyte activation antigen 4F2 large subunit; 4F2 heavy chain antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134546 309.00
Check your price
  • CD51/CD61 Antibody (Monoclonal, 23C6)
  • Host Mouse
  • Target Species Chicken, Human
  • Conjugate Unconjugated
  • Catalog # MA516470
  • Size 200 ug
Applications
Flow Cytometry (1:40-1:80), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
Platelet membrane glycoprotein IIIa; Vitronectin receptor subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516470 375.00
Check your price
  • Sarcomeric alpha Actin Antibody (Monoclonal, 5C5)
  • Host Mouse
  • Target Species Bovine, Fish, Guinea pig, Human, Sheep, Pig, Rabbit, Reptile, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA121597
  • Size 6 mL
Applications
Immunohistochemistry (Frozen) (Neat), Immunohistochemistry (Paraffin) (Neat)
Alternate names:
Alpha-cardiac actin; Actin alpha 1; Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121597 349.00
Check your price
  • Cytokeratin 3 Antibody (Monoclonal, AE5)
  • Host Mouse
  • Target Species Bovine, Human, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA15763
  • Size 100 µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (1:1000-1:3000)
Alternate names:
Type-II keratin Kb3; Keratin-3; Cytokeratin-3; 65 kDa cytokeratin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15763 600.00
Check your price
  • FKBP4 Antibody (Monoclonal, KN382/EC1)
  • Host Mouse
  • Target Species Mink, Mouse, Non-human primate, Rabbit, Bovine, Guinea pig, Hamster, Human
  • Conjugate Unconjugated
  • Catalog # MA1111
  • Size 200 µg
Applications
Immunocytochemistry (1:10-1:1000), Immunofluorescence (1:10-1:1000), Immunohistochemistry (Paraffin) (1:50-1:5000), Immunoprecipitation (10 ug/ml), Western Blot (1:1000)
Alternate names:
Rotamase; Immunophilin FKBP52; HSP-binding immunophilin; FKBP59; FK506-binding protein 4; 59 kDa immunophilin; 52 kDa FK506-binding protein; 51 kDa FK506-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1111 425.00
Check your price
  • Sarcomeric alpha Actin Antibody (Monoclonal, 5C5)
  • Host Mouse
  • Target Species Bovine, Fish, Guinea pig, Human, Mouse, Sheep, Pig, Rabbit, Reptile, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA126928
  • Size 500 µL
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (1:25-1:50), Immunohistochemistry (Paraffin) (1:25-1:50)
Alternate names:
Alpha-cardiac actin; Actin alpha 1; Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126928 399.00
Check your price
  • Metallothionein Antibody (Monoclonal, UC1MT)
  • Host Mouse
  • Target Species Horse, Fish, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA125479
  • Size 50 µg
Applications
Immunohistochemistry (Assay Dependent), Western Blot (1 µg/ml)
Alternate names:
Metallothionein-I; Metallothionein-IA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125479 399.00
Check your price
  • MMP2 Antibody (Monoclonal, 101)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rabbit, Rhesus monkey, Rat
  • Conjugate Unconjugated
  • Catalog # 436000
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:20-1:200), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Matrix metalloproteinase-2; Gelatinase A; 72 kDa gelatinase; TBE-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
436000 309.00
Check your price
  • MYL1 Antibody (Monoclonal, MY32)
  • Host Mouse
  • Target Species Guinea pig, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA190701
  • Size 6 mL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (Neat)
Alternate names:
Myosin light chain alkali 1/2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190701 309.00
Check your price
  • GAPDH Antibody (Monoclonal, 6C5)
  • Host Mouse
  • Target Species Amphibian, Dog, Chicken, Fish, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # AM4300
  • Size 100 µg
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Western Blot (Assay Dependent)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH
Catalog # Size Price (AUD) Your Price (AUD) Qty
AM4300 181.00
Check your price
  • Cardiac Troponin T Antibody (Monoclonal, 13-11)
  • Host Mouse
  • Target Species Avian, Dog, Chicken, Fish, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA512960
  • Size 500 µL
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunomicroscopy (Assay Dependent)
Alternate names:
Cardiac muscle troponin T
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512960 309.00
Check your price
  • NHE3 Antibody (Monoclonal, 19F5)
  • Host Mouse
  • Target Species Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA146355
  • Size 100 µL
Applications
ELISA (Assay-Dependent), Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (1:10-1:500), Immunohistochemistry (Paraffin) (1:10-1:500), Immunoprecipitation (1:10-1:500), Western Blot (0.5 µg/ml)
Alternate names:
Solute carrier family 9 member 3; Na(+)/H(+) exchanger 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146355 309.00
Check your price
  • ATP1B1 Antibody (Monoclonal, 464.8 (aka 8A))
  • Host Mouse
  • Target Species Bovine, Dog, Hamster, Human, Pig, Rabbit, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA116732
  • Size 100 µL
Applications
Immunocytochemistry (1:500-1:1000), Immunofluorescence (1:500-1:1000), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:1000-1:5000)
Alternate names:
Sodium/potassium-dependent ATPase subunit beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116732 375.00
Check your price