1 - 30 of 30 products displayed
  • Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 488 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Rat
  • Conjugate DyLight® 488
  • Catalog # SA510018
  • Size 500ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510018 157.00
Check your price
  • F(ab')2-Goat anti-Rat IgG Cross Adsorbed Secondary Antibody, FITC conjugate for Flow
  • Host Goat
  • Target Species Rat
  • Conjugate FITC
  • Catalog # PA533282
  • Size 500µL
Applications
Flow Cytometry (1:25-1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533282 225.00
Check your price
  • Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 800 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Rat
  • Conjugate DyLight® 800
  • Catalog # SA510024
  • Size 500ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510024 156.00
Check your price
  • Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 680 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Rat
  • Conjugate DyLight® 680
  • Catalog # SA510022
  • Size 500ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510022 156.00
Check your price
  • Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 650 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Rat
  • Conjugate DyLight® 650
  • Catalog # SA510021
  • Size 500ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510021 157.00
Check your price
  • Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 594 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Rat
  • Conjugate DyLight® 594
  • Catalog # SA510020
  • Size 500ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510020 157.00
Check your price
  • Chicken anti-Rat IgG Secondary Antibody, Texas Red® conjugate for IHC (F), IHC (P), Flow and IM
  • Host Chicken
  • Target Species Rat
  • Conjugate Texas Red®
  • Catalog # PA130360
  • Size 1mg
Applications
Flow Cytometry (Assay-dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay-dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130360 349.00
Check your price
  • Chicken anti-Rat IgG Secondary Antibody, Biotin conjugate for Western Blot, IHC (F), IHC (P), ELISA and IM
  • Host Chicken
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # PA130361
  • Size 1mg
Applications
ELISA (1:20,000-1:400,000), Immunohistochemistry (Frozen) (1:500-1:5000), Immunohistochemistry (Paraffin) (1:500-1:5000), Immunomicroscopy (Assay-dependent), Western Blot (1:20,000-1:400,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130361 349.00
Check your price
  • Chicken anti-Rat IgG Secondary Antibody, TRITC conjugate for IHC (F), IHC (P), Flow and IM
  • Host Chicken
  • Target Species Rat
  • Conjugate TRITC
  • Catalog # PA129949
  • Size 1mg
Applications
Flow Cytometry (Assay-dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay-dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129949 309.00
Check your price
  • Chicken anti-Rat IgG Secondary Antibody, HRP conjugate for Western Blot, IHC (F), IHC (P), ELISA and IM
  • Host Chicken
  • Target Species Rat
  • Conjugate HRP
  • Catalog # PA130362
  • Size 1mg
Applications
ELISA (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130362 379.00
Check your price
  • Chicken anti-Rat IgG Secondary Antibody, AP conjugate for Western Blot, IHC (F), IHC (P), ELISA and IM
  • Host Chicken
  • Target Species Rat
  • Conjugate AP
  • Catalog # PA130741
  • Size 1mg
Applications
ELISA (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130741 375.00
Check your price
  • Chicken anti-Rat IgG Secondary Antibody for IHC (F) and IHC (P)
  • Host Chicken
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA128630
  • Size 1mg
Applications
Immunodiffusion (Assay-dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunoprecipitation (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128630 198.00
Check your price
  • Rat IgG Isotype Control for IF, ICC, IHC and BLOCK
  • Host Rat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # 31933
  • Size 10mg
Applications
Blocking Assay (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
Negative control, Isotype control, Immunoglobulin control, Control
Catalog # Size Price (AUD) Your Price (AUD) Qty
31933 126.00
Check your price
  • Rabbit anti-Rat IgG Secondary Antibody, FITC conjugate for ICC, Flow and ELISA
  • Host Rabbit
  • Target Species Rat
  • Conjugate FITC
  • Catalog # PA128569
  • Size 1mg
Applications
Flow Cytometry (Assay-dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay-dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128569 199.00
Check your price
  • Sheep anti-Rat IgG Secondary Antibody, FITC conjugate for IHC (F), IHC (P), Flow and IM
  • Host Sheep
  • Target Species Rat
  • Conjugate FITC
  • Catalog # PA128638
  • Size 1mg
Applications
Flow Cytometry (Assay-dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay-dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128638 197.00
Check your price
  • F(ab')2-Rabbit anti-Rat IgG Secondary Antibody, HRP conjugate for IHC, IHC (F), IHC (P) and ELISA
  • Host Rabbit
  • Target Species Rat
  • Conjugate HRP
  • Catalog # PA533281
  • Size 1mg
Applications
ELISA (1:500-1:1000), Immunohistochemistry (1:50), Immunohistochemistry (Frozen) (1:50), Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533281 375.00
Check your price
  • Goat anti-Rat IgG Secondary Antibody, WesternDot® 585 conjugate for Western Blot
  • Host Goat
  • Target Species Rat
  • Conjugate WesternDot® 585
  • Catalog # W10817
  • Size 25blots
Applications
Western Blot (Assay-Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10817 183.00
Check your price
  • Goat anti-Rat IgG Secondary Antibody, WesternDot® 800 conjugate for Western Blot
  • Host Goat
  • Target Species Rat
  • Conjugate WesternDot® 800
  • Catalog # W10821
  • Size 25blots
Applications
Western Blot (Assay-Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10821 180.00
Check your price
  • Goat anti-Rat IgG Secondary Antibody, WesternDot® 655 conjugate for Western Blot
  • Host Goat
  • Target Species Rat
  • Conjugate WesternDot® 655
  • Catalog # W10819
  • Size 25blots
Applications
Western Blot (Assay-Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10819 183.00
Check your price
  • Goat anti-Rat IgG Secondary Antibody, WesternDot® 625 conjugate for Western Blot
  • Host Goat
  • Target Species Rat
  • Conjugate WesternDot® 625
  • Catalog # W10818
  • Size 25blots
Applications
Western Blot (Assay-Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10818 180.00
Check your price
  • Mouse anti-Rat IgG (Kappa light chain) Secondary Antibody for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # SA170012
  • Size 250ug
Applications
Flow Cytometry (1 µg/1 million cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA170012 231.00
Check your price
  • Chicken anti-Rat IgG Secondary Antibody, FITC conjugate for IHC (F), IHC (P), Flow and IM
  • Host Chicken
  • Target Species Rat
  • Conjugate FITC
  • Catalog # PA129948
  • Size 1mg
Applications
Flow Cytometry (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129948 321.00
Check your price
  • Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 647 conjugate for Western Blot, IF, ICC, IHC and IP
  • Host Goat
  • Target Species Rat
  • Conjugate Alexa Fluor® 647
  • Catalog # A21247
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21247 169.00
Check your price
  • Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 488 conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Rat
  • Conjugate Alexa Fluor® 488
  • Catalog # A11006
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11006 169.00
Check your price
  • Goat anti-Rat IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Goat
  • Target Species Rat
  • Conjugate HRP
  • Catalog # 31470
  • Size 2mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31470 149.00
Check your price
  • Goat anti-Rat IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Rat
  • Conjugate HRP
  • Catalog # 31471
  • Size 0.2mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31471 149.00
Check your price
  • Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, Texas Red®-X conjugate for IF
  • Host Goat
  • Target Species Rat
  • Conjugate Texas Red®-X
  • Catalog # T6392
  • Size 500µL
Applications
Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
T6392 138.00
Check your price
  • Rabbit anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # 31219
  • Size 500ug
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31219 197.00
Check your price
  • Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 594 conjugate for IF and Flow
  • Host Goat
  • Target Species Rat
  • Conjugate Alexa Fluor® 594
  • Catalog # A11007
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11007 169.00
Check your price
  • Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 568 conjugate for IF, ICC and IHC
  • Host Goat
  • Target Species Rat
  • Conjugate Alexa Fluor® 568
  • Catalog # A11077
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11077 169.00
Check your price