1 - 30 of 30 products displayed
  • Calcium Sensing Receptor Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA137213
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Parathyroid cell calcium-sensing receptor 1; Parathyroid cell calcium-sensing receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137213 475.00
Check your price
  • Calcium Sensing Receptor Antibody (Monoclonal, HL 1499)
  • Host Mouse
  • Target Species Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA116940
  • Size 100 µL
Applications
ELISA (1:100-1:2000), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Frozen) (1:50-1:200), Immunoprecipitation (5-10 µg), Radioimmune Assays (Assay-Dependent), Western Blot (1:50-1:200)
Alternate names:
Parathyroid cell calcium-sensing receptor; Parathyroid cell calcium-sensing receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116940 375.00
Check your price
  • Calcium Sensing Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132832
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Parathyroid cell calcium-sensing receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132832 425.00
Check your price
  • Progesterone Receptor Antibody (Monoclonal, Alpha PR6)
  • Host Mouse
  • Target Species Bovine, Chicken, Guinea pig, Human, Mouse, Sheep, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA1411
  • Size 100 µg
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (10 µg/ml), Immunohistochemistry (Frozen) (20 µg/ml), Immunohistochemistry (Paraffin) (5 µg/ml), Immunoprecipitation (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1411 375.00
Check your price
  • Calcium Sensing Receptor Antibody (Monoclonal, 5C10, ADD)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1934
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (2 ug/ml), Western Blot (2 ug/ml)
Alternate names:
Parathyroid cell calcium-sensing receptor; Parathyroid cell calcium-sensing receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1934 375.00
Check your price
  • Androgen Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517771
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Nuclear receptor subfamily 3 group C member 4; Dihydrotestosterone receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517771 275.00
Check your price
  • Calcitonin Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533381
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (6.8 µg/ml)
Alternate names:
CALCR, Calcitonin receptor, CRT, CT-R, CTR, CT receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533381 349.00
Check your price
  • Calcitonin Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533379
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (9.2 µg/ml)
Alternate names:
CALCR, Calcitonin receptor, CRT, CT-R, CTR, CT receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533379 349.00
Check your price
  • Calcitonin Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533380
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (9 µg/ml)
Alternate names:
CALCR, Calcitonin receptor, CRT, CT-R, CTR, CT receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533380 349.00
Check your price
  • Progesterone Receptor Antibody (Monoclonal, alpha PR-22)
  • Host Mouse
  • Target Species Chicken, Human, Reptile
  • Conjugate Unconjugated
  • Catalog # MA1412
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Gel Shift (Assay Dependent), Immunofluorescence (10 µg/ml), Immunoprecipitation (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1412 349.00
Check your price
  • Endothelin B Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120534
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Endothelin receptor non-selective type
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120534 518.00
Check your price
  • Oxytocin Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534065
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (9 µg/ml)
Alternate names:
OXTR, Oxytocin receptor, Oxt-r, OT-R
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534065 349.00
Check your price
  • Oxytocin Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534066
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml)
Alternate names:
OXTR, Oxytocin receptor, Oxt-r, OT-R
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534066 349.00
Check your price
  • Glucagon Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533595
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (11 µg/ml)
Alternate names:
GCGR, GL-R, Glucagon receptor, GGR
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533595 349.00
Check your price
  • Glucagon Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533594
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (13 µg/ml)
Alternate names:
GCGR, GL-R, Glucagon receptor, GGR
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533594 349.00
Check your price
  • Glucagon Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533596
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (15 µg/ml)
Alternate names:
GCGR, GL-R, Glucagon receptor, GGR
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533596 349.00
Check your price
  • beta-2 Adrenergic Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OPA115269
  • Size 25ug
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Beta-2 adrenoreceptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA115269 399.00
Check your price
  • Prolactin Receptor Antibody (Monoclonal, B6.2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511955
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml)
Alternate names:
PRL-R, delta 4-SF1b truncated prolactin receptor, delta 4-delta 7/11 truncated prolactin receptor, hPRL receptor, prolactin receptor delta 7/11, secreted prolactin binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511955 349.00
Check your price
  • alpha-1b Adrenergic Receptor Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA527120
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
ADRA1, ALPHA1BAR, alpha-1B adrenergic receptor, alpha-1B adrenoceptor, alpha-1B adrenoreceptor, alpha-1B-adrenergic receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527120 349.00
Check your price
  • Apelin Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533343
  • Size 50 ug
Applications
Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (8 µg/ml)
Alternate names:
G-protein coupled receptor HG11; G-protein coupled receptor APJ; Angiotensin receptor-like 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533343 349.00
Check your price
  • Apelin Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531978
  • Size 100 µL
Applications
Western Blot (1:5000-1:20,000)
Alternate names:
G-protein coupled receptor HG11; G-protein coupled receptor APJ; Angiotensin receptor-like 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531978 349.00
Check your price
  • Androgen Receptor Antibody, Biotin (Monoclonal, AR 441)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA513423
  • Size 500 µL
Applications
Immunocytochemistry (1:25-1:50), Immunofluorescence (1:25-1:50), Immunohistochemistry (Paraffin) (1:25), Immunoprecipitation (2 ug), Western Blot (1:100-1:200)
Alternate names:
Nuclear receptor subfamily 3 group C member 4; Dihydrotestosterone receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513423 420.00
Check your price
  • Androgen Receptor Antibody (Monoclonal, AR 441)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513426
  • Size 500 µL
Applications
Immunocytochemistry (1:25-1:50), Immunofluorescence (1:25-1:50), Immunohistochemistry (Paraffin) (1:25), Immunoprecipitation (2 ug), Western Blot (1:100-1:200)
Alternate names:
Nuclear receptor subfamily 3 group C member 4; Dihydrotestosterone receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513426 349.00
Check your price
  • Androgen Receptor Antibody (Monoclonal, AN1-15)
  • Host Rat
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA1150
  • Size 50 µg
Applications
Gel Shift (Assay dependent), Immunocytochemistry (5 µg/ml), Immunohistochemistry (Paraffin, Frozen) (2-4 ug/ml), Immunoprecipitation (Assay dependent), Western Blot (1:50-1:500)
Alternate names:
Nuclear receptor subfamily 3 group C member 4; Dihydrotestosterone receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1150 375.00
Check your price
  • Androgen Receptor Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA1110
  • Size 200 µL
Applications
Immunocytochemistry (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (1:10-1:100), Immunoprecipitation (Assay dependent), Western Blot (1:500)
Alternate names:
Nuclear receptor subfamily 3 group C member 4; Dihydrotestosterone receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1110 349.00
Check your price
  • alpha-2c Adrenergic Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527166
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Alpha-2C adrenoreceptor; Alpha-2 adrenergic receptor subtype C4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527166 349.00
Check your price
  • alpha-2c Adrenergic Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA123562
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Alpha-2C adrenoreceptor; Alpha-2 adrenergic receptor subtype C4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123562 425.00
Check your price
  • alpha-2c Adrenergic Receptor Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA14518
  • Size 100 µg
Applications
Immunohistochemistry (1:100-1:300), Western Blot (1:500-1:1000)
Alternate names:
Alpha-2C adrenoreceptor; Alpha-2 adrenergic receptor subtype C4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14518 375.00
Check your price
  • Progesterone Receptor Antibody (Monoclonal, PgR636)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512581
  • Size 500 µL
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:10-1:20), Western Blot (1:500-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512581 850.00
Check your price
  • Progesterone Receptor Antibody (Monoclonal, SP42)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516394
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:400)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516394 700.00
Check your price