1 - 30 of 30 products displayed
  • DYKDDDDK Tag Monoclonal Antibody for Western Blot and ELISA
  • Host Rat
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA11421MG
  • Size 1mg
Applications
ELISA (100ng/ml), Western Blot (1ug/ml)
Alternate names:
FLAG Tag, DYKDDDDK tag, FLAG antibody, ECS epitope tag, DDDDK epitope tag, DDDDK, DYKDDDDK epitope tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, ECS tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA11421MG 900.00
Check your price
  • DYKDDDDK Tag Monoclonal Antibody for Western Blot and ELISA
  • Host Rat
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA1142
  • Size 100ug
Applications
ELISA (100ng/ml), Western Blot (1ug/ml)
Alternate names:
FLAG Tag, DYKDDDDK tag, FLAG antibody, ECS epitope tag, DDDDK epitope tag, DDDDK, DYKDDDDK epitope tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, ECS tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1142 159.00
Check your price
  • RAD51 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # PA534905
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
BRCA1/BRCA2-containing complex, subunit 5, DNA repair protein RAD51 homolog 1, RAD51 homolog A, RecA, E. coli, homolog of, RecA-like protein, recombination protein A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534905 349.00
Check your price
  • MRE11 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517261
  • Size 100µL
Applications
Immunoprecipitation (2 ug), Western Blot (0.2 µg/ml)
Alternate names:
Meiotic recombination 11 homolog A; Meiotic recombination 11 homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517261 349.00
Check your price
  • MRE11 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA531262
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Meiotic recombination 11 homolog A; Meiotic recombination 11 homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531262 349.00
Check your price
  • RAG1 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517163
  • Size 100ug
Applications
Western Blot (1:500-1:2000)
Alternate names:
RING finger protein 74
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517163 309.00
Check your price
  • RAG2 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515622
  • Size 100µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
RAG-2; RAG2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515622 349.00
Check your price
  • DMC1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521472
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
RP1-199H16.4, DMC1H, HsLim15, LIM15, dJ199H16.1, DMC1 homologue, disrupted meiotic cDNA1, yeast, homolog of, meiotic recombination protein DMC1/LIM15 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521472 349.00
Check your price
  • DMC1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521283
  • Size 100ug
Applications
Western Blot (1:500-1:3000)
Alternate names:
RP1-199H16.4, DMC1H, HsLim15, LIM15, dJ199H16.1, DMC1 homologue, disrupted meiotic cDNA1, yeast, homolog of, meiotic recombination protein DMC1/LIM15 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521283 349.00
Check your price
  • KPNA1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529474
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
SRP1-beta; RAG cohort protein 2; Nucleoprotein interactor 1; Karyopherin subunit alpha-1; Importin alpha-S1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529474 349.00
Check your price
  • RBPJ Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA522226
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
RBP-J kappa; J kappa-recombination signal-binding protein; Renal carcinoma antigen NY-REN-30; CBF-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522226 349.00
Check your price
  • CD99 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137370
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
T-cell surface glycoprotein E2; Protein MIC2; E2 antigen; 120000000
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137370 349.00
Check your price
  • RAG2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA522048
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
RAG-2, V(D)J recombination-activating protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522048 349.00
Check your price
  • RAD51B Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528351
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
RAD51-like protein 1; RAD51 homolog B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528351 349.00
Check your price
  • SPO11 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530549
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Cancer/testis antigen 35
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530549 349.00
Check your price
  • RAD54B Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527877
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
RAD54 homolog B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527877 349.00
Check your price
  • RAD54B Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529881
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
RAD54 homolog B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529881 349.00
Check your price
  • RAD51 Monoclonal Antibody, Biotin conjugate for Western Blot, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA514416
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
RAD51 homolog A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514416 399.00
Check your price
  • RAD51 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA514419
  • Size 500µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1:50-1:500)
Alternate names:
RAD51 homolog A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514419 349.00
Check your price
  • RAD51 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527195
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
RAD51 homolog A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527195 Not Orderable Online
Check your price
  • RAD51 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531144
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
RAD51 homolog A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531144 349.00
Check your price
  • RAD54L Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527232
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
RAD54 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527232 349.00
Check your price
  • RAD52 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA123334
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123334 309.00
Check your price
  • SSRP1 Polyclonal Antibody for Western Blot, IHC (P) and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522186
  • Size 100µL
Applications
ChIP assay (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
T160; Structure-specific recognition protein 1; Recombination signal sequence recognition protein 1; Facilitates chromatin transcription complex subunit SSRP1; Facilitates chromatin transcription complex 80 kDa subunit; Chromatin-specific transcription elongation factor 80 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522186 349.00
Check your price
  • CD59 Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517046
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:50 -1:200), Immunofluorescence (1:50-200), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Protectin; Membrane inhibitor of reactive lysis; Membrane attack complex inhibition factor; MEM43 antigen; MAC-inhibitory protein; 20 kDa homologous restriction factor; 1F5 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517046 309.00
Check your price
  • CD200 Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MA523575
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
My033, MOX1, MOX2, MRC, OX-2, CD200 antigen, MRC OX-2 antigen, OX-2 membrane glycoprotein, antigen identified by monoclonal antibody MRC OX-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523575 249.00
Check your price
  • CD200 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA523576
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
My033, MOX1, MOX2, MRC, OX-2, CD200 antigen, MRC OX-2 antigen, OX-2 membrane glycoprotein, antigen identified by monoclonal antibody MRC OX-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523576 249.00
Check your price
  • CD200 Polyclonal Antibody, PE conjugate for Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # PA546880
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
My033, MOX1, MOX2, MRC, OX-2, CD200 antigen, MRC OX-2 antigen, OX-2 membrane glycoprotein, antigen identified by monoclonal antibody MRC OX-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546880 209.00
Check your price
  • CD200 Polyclonal Antibody, APC conjugate for Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate APC
  • Catalog # PA546878
  • Size 100Tests
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
My033, MOX1, MOX2, MRC, OX-2, CD200 antigen, MRC OX-2 antigen, OX-2 membrane glycoprotein, antigen identified by monoclonal antibody MRC OX-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546878 199.00
Check your price
  • CD200 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524048
  • Size 100µg
Applications
Flow Cytometry (0.25 µg/10^6 cells)
Alternate names:
My033, MOX1, MOX2, MRC, OX-2, CD200 antigen, MRC OX-2 antigen, OX-2 membrane glycoprotein, antigen identified by monoclonal antibody MRC OX-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524048 275.00
Check your price