1 - 30 of 30 products displayed
  • RFP Tag Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # PA1986
  • Size 100µg
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (1:1000)
Alternate names:
Red Fluorescent Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1986 249.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Rhodamine Red-X conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Rhodamine Red-X
  • Catalog # R6393
  • Size 500µL
Applications
Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
R6393 137.00
Check your price
  • eGFP Tag Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA1952
  • Size 100µg
Applications
Immunocytochemistry (1:20-1:100), Immunofluorescence (1:20-1:100), Western Blot (1:1,000)
Alternate names:
Enhanced Green Fluorescence Protein, Venus, enhanced GFP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1952 159.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 594 conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 594
  • Catalog # A11012
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11012 169.00
Check your price
  • F(ab')2-Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 594 conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 594
  • Catalog # A11020
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11020 149.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 594 conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 594
  • Catalog # A11005
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11005 169.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 594 conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 594
  • Catalog # A11072
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11072 149.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor® 594 conjugate for IF, ICC, IHC and Flow
  • Host Donkey
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 594
  • Catalog # A21207
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21207 209.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor® 555 conjugate for IF, ICC, IHC and Flow
  • Host Donkey
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 555
  • Catalog # A31572
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A31572 209.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 633 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 633
  • Catalog # A21070
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21070 169.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor® 633 conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 633
  • Catalog # A21071
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21071 209.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor® 594 conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 594
  • Catalog # A11037
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11037 209.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 568 conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 568
  • Catalog # A11011
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11011 169.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor® 568 conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 568
  • Catalog # A11036
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11036 209.00
Check your price
  • Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor® 594 conjugate for IF, ICC, IHC and Flow
  • Host Donkey
  • Target Species Mouse
  • Conjugate Alexa Fluor® 594
  • Catalog # A21203
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21203 209.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 647 conjugate for IF and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 647
  • Catalog # A21235
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21235 169.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 633 conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 633
  • Catalog # A21050
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21050 169.00
Check your price
  • Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor® 633 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 633
  • Catalog # A21052
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21052 209.00
Check your price
  • Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor® 594 conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 594
  • Catalog # A11032
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11032 209.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 568 conjugate for IF, ICC, IHC (F) and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 568
  • Catalog # A11004
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11004 169.00
Check your price
  • Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor® 568 conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 568
  • Catalog # A11031
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11031 209.00
Check your price
  • Goat anti-Mouse IgG1 Cross-Adsorbed Secondary Antibody, Alexa Fluor® 568 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 568
  • Catalog # A21124
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21124 149.00
Check your price
  • Goat anti-Mouse IgG1 Cross-Adsorbed Secondary Antibody, Alexa Fluor® 488 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 488
  • Catalog # A21121
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21121 149.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor® 568 conjugate for IF, ICC, IHC and Flow
  • Host Donkey
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 568
  • Catalog # A10042
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A10042 209.00
Check your price
  • Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor® 568 conjugate for IF, ICC, IHC and Flow
  • Host Donkey
  • Target Species Mouse
  • Conjugate Alexa Fluor® 568
  • Catalog # A10037
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A10037 209.00
Check your price
  • Goat anti-Mouse IgG / IgM (H+L) Secondary Antibody, Alexa Fluor® 488 conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 488
  • Catalog # A10680
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A10680 149.00
Check your price
  • Goat anti-Mouse IgM Heavy Chain Cross-Adsorbed Secondary Antibody, Alexa Fluor® 488 conjugate for IF, ICC and IHC
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 488
  • Catalog # A21042
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21042 149.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, Alexa Fluor® 594 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Chicken
  • Conjugate Alexa Fluor® 594
  • Catalog # A11042
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11042 169.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, Alexa Fluor® 568 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Chicken
  • Conjugate Alexa Fluor® 568
  • Catalog # A11041
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11041 169.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, Alexa Fluor® 488 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Chicken
  • Conjugate Alexa Fluor® 488
  • Catalog # A11039
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11039 169.00
Check your price