1 - 3 of 3 products displayed
  • RSV M2 Antibody
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA123415
  • Size 100 µg
Applications
ELISA (Assay dependent), Western Blot (1:500-1:1000)
Alternate names:
human respiratory syncytial virus, RSV, RSV M2, RSV Matrix protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123415 375.00
Check your price
  • RSV M2 Antibody
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA522828
  • Size 100 ug
Applications
Western Blot (1:500-1:1000)
Alternate names:
human respiratory syncytial virus, RSV, RSV M2, RSV Matrix protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522828 349.00
Check your price
  • RSV M2 Antibody
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA14183
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
human respiratory syncytial virus, RSV, RSV M2, RSV Matrix protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14183 349.00
Check your price