1 - 30 of 30 products displayed
  • PPAR alpha Polyclonal Antibody for Western Blot, IF, Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1822A
  • Size 100µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunofluorescence (1:100), Immunoprecipitation (Assay dependent), Western Blot (1.0 ug/ml)
Alternate names:
Nuclear receptor subfamily 1 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1822A 375.00
Check your price
  • Phospho-PPAR alpha (Ser12) Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1820
  • Size 100µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:100-1:1000)
Alternate names:
Nuclear receptor subfamily 1 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1820 375.00
Check your price
  • PPAR alpha Monoclonal Antibody for Western Blot, IF, ICC, Flow, IP, ChIP and GS
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1822
  • Size 100µg
Applications
ChIP assay (Assay Dependent), Flow Cytometry (1µg per 10^6 cells), Gel Shift (Assay dependent), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunoprecipitation (Assay dependent), Western Blot (2-5 µg/ml)
Alternate names:
Nuclear receptor subfamily 1 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1822 375.00
Check your price
  • PPAR alpha Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 424600
  • Size 100µg
Applications
Flow Cytometry (1:100-1:500), Western Blot (1-3 µg/ml)
Alternate names:
Nuclear receptor subfamily 1 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
424600 309.00
Check your price
  • LRP1 Monoclonal Antibody for Western Blot, IF, IHC, IHC (F) and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 373800
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (1:100), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Alpha-2-macroglobulin receptor; Apolipoprotein E receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
373800 305.00
Check your price
  • PPAR alpha/gamma Polyclonal Antibody for Western Blot, IP and GS
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA3820
  • Size 100µL
Applications
Gel Shift (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:2,000)
Alternate names:
Nuclear receptor subfamily 1 group C member 1; Nuclear receptor subfamily 1 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3820 375.00
Check your price
  • LXR alpha/beta Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1331B
  • Size 200µg
Applications
Western Blot (1:500)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha; Ubiquitously-expressed nuclear receptor; Nuclear receptor subfamily 1 group H member 2; Nuclear receptor NER; Liver X receptor beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1331B 349.00
Check your price
  • LXR alpha Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529868
  • Size 100µL
Applications
Western Blot (1:1000-1:10,000)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529868 349.00
Check your price
  • LXR alpha Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528439
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528439 349.00
Check your price
  • LXR alpha Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA1332
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1332 375.00
Check your price
  • RORA Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534158
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (11-17 µg/ml)
Alternate names:
Retinoid-related orphan receptor-alpha; RAR-related orphan receptor A; Nuclear receptor subfamily 1 group F member 1; Nuclear receptor RZR-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534158 349.00
Check your price
  • RORA Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534159
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (3 µg/ml)
Alternate names:
Retinoid-related orphan receptor-alpha; RAR-related orphan receptor A; Nuclear receptor subfamily 1 group F member 1; Nuclear receptor RZR-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534159 349.00
Check your price
  • RORA Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1812
  • Size 100µg
Applications
Western Blot (1 ug/ml)
Alternate names:
Retinoid-related orphan receptor-alpha; RAR-related orphan receptor A; Nuclear receptor subfamily 1 group F member 1; Nuclear receptor RZR-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1812 349.00
Check your price
  • RXRA Monoclonal Antibody for Western Blot, IHC, IP, ChIP, ELISA and GS
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 433900
  • Size 100µg
Applications
ChIP assay (Assay Dependent), ELISA (Assay Dependent), Gel Shift (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Retinoid X receptor alpha; Nuclear receptor subfamily 2 group B member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
433900 426.00
Check your price
  • LXR alpha Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 418300
  • Size 100µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
418300 426.00
Check your price
  • RORA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Chicken, Horse, Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523268
  • Size 100ug
Applications
Western Blot (0.01-0.25 µg/ml)
Alternate names:
Retinoid-related orphan receptor-alpha; RAR-related orphan receptor A; Nuclear receptor subfamily 1 group F member 1; Nuclear receptor RZR-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523268 349.00
Check your price
  • RORA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Chicken, Horse, Human, Marsupial, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523267
  • Size 100ug
Applications
Western Blot (0.025-0.05 µg/ml)
Alternate names:
Retinoid-related orphan receptor-alpha; RAR-related orphan receptor A; Nuclear receptor subfamily 1 group F member 1; Nuclear receptor RZR-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523267 349.00
Check your price
  • LRP1 Monoclonal Antibody for Western Blot, IF, IHC (F), Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA180192
  • Size 200µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1/50-1/100), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay dependent (Non-redu)
Alternate names:
Apolipoprotein E receptor; Alpha-2-macroglobulin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180192 499.00
Check your price
  • PPAR alpha Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA186844
  • Size 50µg
Applications
ELISA (1:8000-1:32,000), Western Blot (1:500-1:1000)
Alternate names:
Nuclear receptor subfamily 1 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186844 375.00
Check your price
  • LRP1 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA135979
  • Size 100tests
Applications
Flow Cytometry (Neat)
Alternate names:
Apolipoprotein E receptor; Alpha-2-macroglobulin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135979 800.00
Check your price
  • RXRG Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # P21939
  • Size 100µL
Applications
Western Blot (Assay Dependent)
Alternate names:
Retinoid X receptor gamma; Nuclear receptor subfamily 2 group B member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
P21939 309.00
Check your price
  • LXR alpha/beta Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA19066
  • Size 100ug
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha; Ubiquitously-expressed nuclear receptor; Retinoid X receptor-interacting protein No.15; Nuclear receptor subfamily 1 group H member 2; Liver X receptor beta; Nuclear receptor NER; Orphan nuclear receptor OR-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19066 349.00
Check your price
  • RXRA Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA19096
  • Size 100ug
Applications
Western Blot (0.5-3 µg/ml)
Alternate names:
Retinoid X receptor alpha; Nuclear receptor subfamily 2 group B member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19096 309.00
Check your price
  • LXR alpha Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520880
  • Size 100µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520880 349.00
Check your price
  • LXR alpha Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526951
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526951 349.00
Check your price
  • LXR alpha Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Pig
  • Conjugate Unconjugated
  • Catalog # PA116824
  • Size 100µL
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116824 349.00
Check your price
  • RXRA Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524845
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Retinoid X receptor alpha; Nuclear receptor subfamily 2 group B member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524845 349.00
Check your price
  • RORA Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511224
  • Size 400µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
Retinoid-related orphan receptor-alpha; RAR-related orphan receptor A; Nuclear receptor subfamily 1 group F member 1; Nuclear receptor RZR-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511224 349.00
Check your price
  • LRP1 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA135977
  • Size 100µg
Applications
Flow Cytometry (1:50)
Alternate names:
Apolipoprotein E receptor; Alpha-2-macroglobulin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135977 750.00
Check your price
  • RXRB Monoclonal Antibody for Western Blot, IF, IP and GS
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA3812
  • Size 100µg
Applications
Gel Shift (Assay dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (5 µg/ml)
Alternate names:
Retinoid X receptor beta; Nuclear receptor subfamily 2 group B member 2; MHC class I regulatory element-binding protein H-2RIIBP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3812 375.00
Check your price