1 - 30 of 30 products displayed
  • Fibrillarin Antibody (Monoclonal, 38F3)
  • Host Mouse
  • Target Species C. elegans, Fruit fly, Human, Mouse, Plant, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # MA122000
  • Size 250 µL
Applications
Immunocytochemistry (1:500-1:5000), Immunofluorescence (1:500-1:5000), Immunohistochemistry (Paraffin) (Assay dependent), Western Blot (1:1000-1:2000)
Alternate names:
Nucleolar protein 1; Histone-glutamine methyltransferase; 34 kDa nucleolar scleroderma antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122000 349.00
Check your price
  • NOP1 Antibody (Monoclonal, 28F2)
  • Host Mouse
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # MA110025
  • Size 500 µL
Applications
Immunocytochemistry (1:2000-1:20,000), Immunofluorescence (1:2000-1:20,000), Immunoprecipitation (2-4 µl/lysate), Western Blot (1:10,000-1:25,000)
Alternate names:
U3 small nucleolar RNA-associated protein NOP1; Histone-glutamine methyltransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110025 375.00
Check your price
  • Fibrillarin Antibody (Monoclonal, 38F3)
  • Host Mouse
  • Target Species Fruit fly, Human, Rat
  • Conjugate Unconjugated
  • Catalog # 480009
  • Size 100 µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Western Blot (1:1,000)
Alternate names:
Nucleolar protein 1; Histone-glutamine methyltransferase; 34 kDa nucleolar scleroderma antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
480009 1.98
Check your price
  • Fibrillarin Antibody (Monoclonal, 38F3)
  • Host Mouse
  • Target Species C. elegans, Chicken, Fruit fly, Human, Mouse, Plant, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # MA316771
  • Size 250 µL
Applications
Flow Cytometry (1:20-1:400), Immunocytochemistry (1:500-1:1000), Immunofluorescence (1:500-1:1000), Immunohistochemistry (Frozen) (1:500-1:1000), Immunohistochemistry (Paraffin) (1:500-1:1000), Western Blot (1:1000-1:5000)
Alternate names:
Nucleolar protein 1; Histone-glutamine methyltransferase; 34 kDa nucleolar scleroderma antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA316771 375.00
Check your price
  • Calprotectin Antibody (Monoclonal, MAC 387)
  • Host Mouse
  • Target Species Dog, Cat, Guinea pig, Human, Mouse, Non-human primate, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA512213
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:1000)
Alternate names:
S100 calcium-binding protein A9; Migration inhibitory factor-related protein 14; Leukocyte L1 complex heavy chain; Calgranulin-B; S100 calcium-binding protein A8; Migration inhibitory factor-related protein 8; Calgranulin-A; Urinary stone protein band A; Leukocyte L1 complex light chain; Cystic fibrosis antigen; Calprotectin L1L subunit; Myeloid-related protein 14; Calprotectin L1H subunit; Pro-inflammatory S100 cytokine; Chemotactic cytokine CP-10; JNK-46
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512213 269.00
Check your price
  • MED4 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA543593
  • Size 100 µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (0.5 µg/ml)
Alternate names:
Mediator complex subunit 4; Vitamin D3 receptor-interacting protein complex 36 kDa component; TRAP/SMCC/PC2 subunit p36 subunit; Activator-recruited cofactor 36 kDa component
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543593 309.00
Check your price
  • MED4 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA543592
  • Size 100 µl
Applications
Immunohistochemistry (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Mediator complex subunit 4; Vitamin D3 receptor-interacting protein complex 36 kDa component; TRAP/SMCC/PC2 subunit p36 subunit; Activator-recruited cofactor 36 kDa component
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543592 309.00
Check your price
  • MED4 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528847
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Vitamin D3 receptor-interacting protein complex 36 kDa component; TRAP/SMCC/PC2 subunit p36 subunit; Mediator complex subunit 4; Activator-recruited cofactor 36 kDa component
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528847 349.00
Check your price
  • MUC1 Antibody (Monoclonal, LH39)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA190931
  • Size 100 µg
Applications
Immunohistochemistry (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Tumor-associated mucin; Tumor-associated epithelial membrane antigen; Polymorphic epithelial mucin; Peanut-reactive urinary mucin; PEMT; Krebs von den Lungen-6; H23AG; Episialin; Carcinoma-associated mucin; Cancer antigen 15-3; Breast carcinoma-associated antigen DF3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190931 375.00
Check your price
  • MPM2 Antibody Monoclonal, FITC conjugate (0.T.166) (MA1-91857)
  • Host Mouse
  • Target Species Human
  • Conjugate
  • Catalog # MA191857
  • Size 50µg
Applications
Immunocytochemistry (2-10 µg/ml), Immunofluorescence (2-10 ug/ml)
Alternate names:
Winged-helix factor from INS-1 cells; Transcription factor Trident; MPM-2 reactive phosphoprotein 2; M-phase phosphoprotein 2; Hepatocyte nuclear factor 3 forkhead homolog 11; Forkhead-related protein FKHL16
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191857 349.00
Check your price
  • MPM2 Antibody Monoclonal (0.T.181) (MA1-91855)
  • Host Mouse
  • Target Species Human, Yeast
  • Conjugate
  • Catalog # MA191855
  • Size 50µg
Applications
ELISA (Assay dependent), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 ug/ml), Western Blot (Assay Dependent)
Alternate names:
Winged-helix factor from INS-1 cells; Transcription factor Trident; MPM-2 reactive phosphoprotein 2; M-phase phosphoprotein 2; Hepatocyte nuclear factor 3 forkhead homolog 11; Forkhead-related protein FKHL16
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191855 359.00
Check your price
  • MUC1 Antibody (Monoclonal, 126E7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135688
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (Assay dependent.)
Alternate names:
Tumor-associated mucin; Tumor-associated epithelial membrane antigen; Polymorphic epithelial mucin; Peanut-reactive urinary mucin; PEMT; Krebs von den Lungen-6; H23AG; Episialin; Carcinoma-associated mucin; Cancer antigen 15-3; Breast carcinoma-associated antigen DF3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135688 349.00
Check your price
  • Cytokeratin 18 Antibody (Monoclonal, RGE53)
  • Host Mouse
  • Target Species Dog, Chicken, Hamster, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA106326
  • Size 100 µL
Applications
Flow Cytometry (Assay dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Western Blot (1:100-1:1000)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; DreK18; Dorsal aorta proneprin kinesin-1; Cytokeratin endo B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106326 375.00
Check your price
  • Cytokeratin 8/18 Antibody (Monoclonal, K8.8 + DC10)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512281
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512281 309.00
Check your price
  • CD56 Antibody (Monoclonal, 56C04)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA511563
  • Size 500 µL
Applications
Flow Cytometry (0.5 ug/test), Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1-2 µg/ml), Western Blot (1:100-1:500)
Alternate names:
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511563 309.00
Check your price
  • Nop5/Nop6 Antibody (Monoclonal, 37C12)
  • Host Mouse
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # MA110028
  • Size 500 µL
Applications
Immunocytochemistry (1:1000-1:10,000), Immunofluorescence (1:1000-1:10,000), Immunohistochemistry (1:1000-1:10,000), Immunoprecipitation (2-4 ul), Western Blot (1:10,000)
Alternate names:
Nucleolar protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110028 309.00
Check your price
  • H-2Dd Antibody (Monoclonal, 34-5-8S)
  • Host Mouse
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA170108
  • Size 250 ug
Applications
Flow Cytometry (1 ug/1 million cells), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
H-2D, H2-D
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA170108 309.00
Check your price
  • Transferrin Receptor Antibody (Monoclonal, MEM-189)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121562
  • Size 100 µg
Applications
Flow Cytometry (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
p90; T9
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121562 349.00
Check your price
  • NSR1 Antibody (Monoclonal, 31C4)
  • Host Mouse
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # MA110030
  • Size 500 µL
Applications
Immunofluorescence (1:500-1:5000), Western Blot (1:1000-1:10,000)
Alternate names:
p67
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110030 309.00
Check your price
  • Cytokeratin Pan Antibody (Monoclonal, C-11)
  • Host Mouse
  • Target Species Amphibian, Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA512231
  • Size 500 µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (1-2 µg/ml), Western Blot (1:10-1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512231 225.00
Check your price
  • Cytokeratin Pan Antibody, Alexa Fluor® 488 (Monoclonal, C-11)
  • Host Mouse
  • Target Species Mammal
  • Conjugate Alexa Fluor® 488
  • Catalog # MA518156
  • Size 100 µg
Applications
Flow Cytometry (10ug/ml)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Keratin-10; Cytokeratin-10; Type-II keratin Kb8; Keratin-8; Cytokeratin-8; Type-II keratin Kb5; Keratin-5; Cytokeratin-5; 58 kDa cytokeratin; Type-II keratin Kb6; Keratin-6A; Cytokeratin-6D; Cytokeratin-6A; Keratin-13; Cytokeratin-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518156 309.00
Check your price
  • Cytokeratin Pan Antibody, APC (Monoclonal, C-11)
  • Host Mouse
  • Target Species Mammal
  • Conjugate APC
  • Catalog # MA110325
  • Size 100 ug
Applications
Flow Cytometry (5 ug/ml), Immunocytochemistry (1:100-1:400), Immunofluorescence (1:100-1:400)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Keratin-10; Cytokeratin-10; Type-II keratin Kb8; Keratin-8; Cytokeratin-8; Type-II keratin Kb5; Keratin-5; Cytokeratin-5; 58 kDa cytokeratin; Type-II keratin Kb6; Keratin-6A; Cytokeratin-6D; Cytokeratin-6A; Keratin-13; Cytokeratin-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110325 349.00
Check your price
  • EpCAM Antibody (Monoclonal, AUA1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA146104
  • Size 100 µl
Applications
ELISA (1:100-1:2000), Flow Cytometry (1 µg/10^6 cells), Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Paraffin) (1:10-1:500), Western Blot (2 µg/ml)
Alternate names:
Tumor-associated calcium signal transducer 1; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein 314; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146104 309.00
Check your price
  • Cytokeratin 5/18 Antibody (Monoclonal, C-50)
  • Host Mouse
  • Target Species Bovine, Dog, Hamster, Human, Mouse, Sheep, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA119031
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Type-II keratin Kb5; Keratin-5; Cytokeratin-5; 58 kDa cytokeratin; Cytokeratin endo B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119031 199.00
Check your price
  • Cytokeratin 18 Antibody, Alexa Fluor® 488 (Monoclonal, C-04)
  • Host Mouse
  • Target Species Mammal
  • Conjugate Alexa Fluor® 488
  • Catalog # MA518157
  • Size 100 µg
Applications
Flow Cytometry (7ulx10^6 cells)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; DreK18; Dorsal aorta proneprin kinesin-1; Cytokeratin endo B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518157 309.00
Check your price
  • Cytokeratin 18 Antibody, Biotin (Monoclonal, C-04)
  • Host Mouse
  • Target Species Mammal
  • Conjugate Biotin
  • Catalog # MA119475
  • Size 100 µg
Applications
Flow Cytometry (1:500)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Cytokeratin endo B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119475 225.00
Check your price
  • Cytokeratin 18 Antibody, FITC (Monoclonal, C-04)
  • Host Mouse
  • Target Species Mammal
  • Conjugate FITC
  • Catalog # MA110326
  • Size 100 ug
Applications
Flow Cytometry (10 ul/10^6 cells)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Cytokeratin endo B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110326 275.00
Check your price
  • Cytokeratin 18 Antibody (Monoclonal, C-04)
  • Host Mouse
  • Target Species Mammal
  • Conjugate Unconjugated
  • Catalog # MA119039
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (10 ug/ml), Immunoprecipitation (Assay Dependent), Western Blot (1-2 ug/ml)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Cytokeratin endo B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119039 375.00
Check your price
  • HSP70 Antibody (Monoclonal, MB-H1)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 333800
  • Size 200 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (Assay Dependent)
Alternate names:
Heat shock 70 kDa protein 2; Heat shock 70 kDa protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
333800 304.00
Check your price
  • Cytokeratin Pan Antibody (Monoclonal, C11)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA112594
  • Size 100 µg
Applications
Immunohistochemistry (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p53-related protein; p53-like transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112594 375.00
Check your price