1 - 5 of 5 products displayed
  • PKA gamma Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528148
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
KAPG, PKACg, PKA C-gamma, cAMP-dependent protein kinase catalytic subunit gamma, serine(threonine) protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528148 349.00
Check your price
  • CDw293 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511861
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Serine(threonine) protein kinase, activin receptor-like kinase 6, ALK6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511861 349.00
Check your price
  • CDw293 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511862
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Serine(threonine) protein kinase, activin receptor-like kinase 6, ALK6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511862 349.00
Check your price
  • CDw293 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA511860
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Serine/threonine-protein kinase receptor R6; Activin receptor-like kinase 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511860 349.00
Check your price
  • PKA gamma Antibody (Monoclonal, 2E4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517158
  • Size 100 µL
Applications
Flow Cytometry (1:200-1:400), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517158 309.00
Check your price