1 - 8 of 8 products displayed
  • PIM2 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA522315
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-dependent), Western Blot (1:500-1:3000)
Alternate names:
Pim-2h
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522315 349.00
Check your price
  • PIM1 Oligoclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 710504
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (1 ug/ml)
Alternate names:
Oncogene PIM1, pim-1 kinase 44 kDa isoform, pim-1 oncogene (proviral integration site 1), proto-oncogene serine/threonine-protein kinase pim-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710504 303.00
Check your price
  • PIM1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536392
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Serine/threonine-protein kinase pim-1, Name: Pim1, Pim-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536392 309.00
Check your price
  • PIM1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515124
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Proto-oncogene serine/threonine-protein kinase pim-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515124 349.00
Check your price
  • PIM2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532552
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Pim-2h
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532552 349.00
Check your price
  • PIM1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515123
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Proto-oncogene serine/threonine-protein kinase pim-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515123 349.00
Check your price
  • PIM3 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513976
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Serine/threonine-protein kinase Pim-3; PIM3;
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513976 349.00
Check your price
  • PIM2 Polyclonal Antibody for Western Blot, IHC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515125
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Immunoprecipitation (1:50-1:100), Western Blot (1:1000)
Alternate names:
Pim-2h
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515125 349.00
Check your price