1 - 10 of 10 products displayed
  • HTR2B Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120706
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Serotonin receptor 2B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120706 425.00
Check your price
  • HTR2B Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533303
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (4-6 µg/ml)
Alternate names:
Serotonin receptor 2B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533303 349.00
Check your price
  • HTR2B Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533304
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (2 µg/ml)
Alternate names:
Serotonin receptor 2B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533304 349.00
Check your price
  • HTR2B Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533305
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (3 µg/ml)
Alternate names:
Serotonin receptor 2B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533305 349.00
Check your price
  • HTR2B Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132494
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200), Western Blot (Assay-dependent)
Alternate names:
Serotonin receptor 2B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132494 399.00
Check your price
  • NMDAR2A/NMDAR2B Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538506
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
N-methyl-D-aspartate receptor subunit 3; N-methyl D-aspartate receptor subtype 2B; Glutamate [NMDA] receptor subunit epsilon-2; N-methyl D-aspartate receptor subtype 2A; Glutamate [NMDA] receptor subunit epsilon-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538506 309.00
Check your price
  • NMDAR2A Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # A6473
  • Size 10 µg
Applications
Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
N-methyl D-aspartate receptor subtype 2A; Glutamate [NMDA] receptor subunit epsilon-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
A6473 392.00
Check your price
  • PSD95 Antibody (Monoclonal, 7E3-1B8)
  • Host Mouse
  • Target Species Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA1046
  • Size 100 µL
Applications
Flow Cytometry (2 ug/test), Immunocytochemistry (1:500), Immunofluorescence (1:200), Immunohistochemistry (Frozen) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:2,000)
Alternate names:
Synapse-associated protein 90; Postsynaptic density protein 95
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1046 375.00
Check your price
  • PSD95 Antibody (Monoclonal, 6G6-1C9)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1045
  • Size 100 µL
Applications
Flow Cytometry (2 ug/test), Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunoprecipitation (Assay dependent), Western Blot (1:2,000)
Alternate names:
Synapse-associated protein 90; Postsynaptic density protein 95
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1045 375.00
Check your price
  • PMCA ATPase Antibody (Monoclonal, 5F10)
  • Host Mouse
  • Target Species Amphibian, Dog, Chicken, Cat, Human, Mouse, Non-human primate, Sheep, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3914
  • Size 100 µL
Applications
Flow Cytometry (1ug/test), Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin, Frozen) (1:500), Immunoprecipitation (Assay dependent), Western Blot (1:1000-1:5000)
Alternate names:
Plasma membrane calcium pump isoform 1; Plasma membrane calcium ATPase isoform 1; Plasma membrane calcium pump isoform 2; Plasma membrane calcium ATPase isoform 2; Plasma membrane calcium pump isoform 3; Plasma membrane calcium ATPase isoform 3; Plasma membrane calcium pump isoform 4; Plasma membrane calcium ATPase isoform 4; Matrix-remodeling-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3914 375.00
Check your price