1 - 30 of 30 products displayed
  • C1 inhibitor Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFMA0136
  • Size 100µL
Applications
Immunoprecipitation (2 µg), Western Blot (0.5 ug/ml)
Alternate names:
Serpin G1; C1-inhibiting factor; C1 esterase inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0136 309.00
Check your price
  • C1 inhibitor Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531541
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Serpin G1; C1-inhibiting factor; C1 esterase inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531541 349.00
Check your price
  • C1 inhibitor Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB2880502
  • Size 200 µl
Applications
ELISA (Assay Dependent)
Alternate names:
Serpin G1; C1-inhibiting factor; C1 esterase inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB2880502 399.00
Check your price
  • C1 inhibitor Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # PA521652
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Serpin G1; C1-inhibiting factor; C1 esterase inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521652 349.00
Check your price
  • C1 inhibitor Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA172519
  • Size 100 µg
Applications
ELISA (1 µg/ml)
Alternate names:
Serpin G1; C1-inhibiting factor; C1 esterase inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA172519 199.00
Check your price
  • C1 inhibitor Polyclonal Antibody, HRP conjugate for ELISA and IE
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # PA174017
  • Size 200ug
Applications
ELISA (Assay Dependent), Immunoelectrophoresis (Assay Dependent)
Alternate names:
Serpin G1; C1-inhibiting factor; C1 esterase inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA174017 309.00
Check your price
  • C1 inhibitor Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513627
  • Size 400 µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Serpin G1; C1-inhibiting factor; C1 esterase inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513627 349.00
Check your price
  • SERPINA4 Polyclonal Antibody for Western Blot, IHC, IP and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547180
  • Size 100 µg
Applications
Immunohistochemistry (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Neutralization (10.8 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Serpin A4; Peptidase inhibitor 4; Kallikrein inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547180 349.00
Check your price
  • SERPINB8 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517647
  • Size 200 ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Peptidase inhibitor 8; Cytoplasmic antiproteinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517647 309.00
Check your price
  • SERPINB8 Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547589
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Peptidase inhibitor 8; Cytoplasmic antiproteinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547589 349.00
Check your price
  • PAI1 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523790
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.3 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Serpin E1; Endothelial plasminogen activator inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523790 349.00
Check your price
  • SERPINB9 Monoclonal Antibody for Western Blot, ICC and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135771
  • Size 1 mL
Applications
Immunocytochemistry (Assay dependent.), Immunohistochemistry (Paraffin) (Assay dependent.), Western Blot (Assay dependent.)
Alternate names:
Peptidase inhibitor 9; Cytoplasmic antiproteinase 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135771 550.00
Check your price
  • SERPINA6 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA548038
  • Size 100 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Transcortin; Serpin A6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA548038 309.00
Check your price
  • SERPINA5 Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547109
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Serpin A5; Protein C inhibitor; Plasminogen activator inhibitor 3; Acrosomal serine protease inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547109 349.00
Check your price
  • SERPIND1 Polyclonal Antibody for Western Blot, IP and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547466
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Neutralization (6.6 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Serpin D1; Protease inhibitor leuserpin-2; Heparin cofactor II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547466 349.00
Check your price
  • SERPINA3 Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546907
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Serpin A3; Cell growth-inhibiting gene 24/25 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546907 375.00
Check your price
  • PAI1 Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701096
  • Size 100 µg
Applications
Western Blot (0.1-2 ug/ml)
Alternate names:
Serpin E1; Endothelial plasminogen activator inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
701096 309.00
Check your price
  • SERPINF2 Polyclonal Antibody for Western Blot, IP and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547142
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Neutralization (4.2 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Serpin F2; Alpha-2-plasmin inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547142 349.00
Check your price
  • Serpina12 Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA547725
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
OL-64, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 12, serpin A12, vaspin, visceral adipose tissue-derived serine protease inhibitor, visceral adipose-specific SERPIN
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547725 349.00
Check your price
  • SERPINA12 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524197
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Visceral adipose-specific serpin; Visceral adipose tissue-derived serine protease inhibitor; OL-64
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524197 275.00
Check your price
  • SERPINA12 Polyclonal Antibody for Western Blot and IP
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546948
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Visceral adipose-specific serpin; Visceral adipose tissue-derived serine protease inhibitor; OL-64
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546948 375.00
Check your price
  • PAI1 Oligoclonal Antibody for ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710121
  • Size 100 µg
Applications
ELISA (Assay-Dependent)
Alternate names:
Serpin E1; Endothelial plasminogen activator inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
710121 309.00
Check your price
  • SERPINA9 Polyclonal Antibody for IHC (P) and IP
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547025
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Immunoprecipitation (25 µg/ml)
Alternate names:
Germinal center B-cell-expressed transcript 1 protein; Centerin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547025 375.00
Check your price
  • Neuroserpin Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529751
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (Assay-dependent), Western Blot (1:500-1:3000)
Alternate names:
Serpin I1; Peptidase inhibitor 12; Serine protease inhibitor 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529751 349.00
Check your price
  • Neuroserpin Polyclonal Antibody for Western Blot, IF and IHC (F)
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519610
  • Size 200 µL
Applications
Immunofluorescence (1 µg/ml), Immunohistochemistry (Frozen) (1 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Serpin I1; Serine protease inhibitor 17; Peptidase inhibitor 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519610 349.00
Check your price
  • SERPINB9 Monoclonal Antibody for Western Blot, Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517648
  • Size 100 ug
Applications
ELISA (Assay Dependent), Flow Cytometry (0.5 ug/10^6 cells), Western Blot (Assay Dependent)
Alternate names:
Peptidase inhibitor 9; Cytoplasmic antiproteinase 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517648 249.00
Check your price
  • SERPINE2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA532163
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Serpin E2; Protease nexin I; Protease nexin 1; Peptidase inhibitor 7; Serine protease-inhibitor 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532163 349.00
Check your price
  • alpha-1 Antitrypsin Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515521
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Serpin A1; Alpha-1-antiproteinase; Alpha-1 protease inhibitor; Serine protease inhibitor A1a; Serine protease inhibitor 1-1; Alpha-1 protease inhibitor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515521 349.00
Check your price
  • SERPINI2 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518360
  • Size 100 ug
Applications
Immunohistochemistry (3µg/ml), Western Blot (0.3-1 ug/ml)
Alternate names:
Peptidase inhibitor 14; Pancreas-specific protein TSA2004; Pancpin; Myoepithelium-derived serine protease inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518360 349.00
Check your price
  • SERPINB13 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14286
  • Size 100 µg
Applications
Western Blot (1:500)
Alternate names:
Proteinase inhibitor 13; Peptidase inhibitor 13; Headpin; HaCaT UV-repressible serpin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14286 309.00
Check your price