1 - 10 of 10 products displayed
  • SHIP1 Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110449
  • Size 100 ug
Applications
Flow Cytometry (2.5 ug/ml), Western Blot (Assay Dependent)
Alternate names:
p150Ship; SH2 domain-containing inositol 5'-phosphatase 1; Inositol polyphosphate-5-phosphatase of 145 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110449 225.00
Check your price
  • SHIP1 Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110450
  • Size 100 ug
Applications
Flow Cytometry (2.5 ug/ml), Western Blot (Assay Dependent)
Alternate names:
p150Ship; SH2 domain-containing inositol 5'-phosphatase 1; Inositol polyphosphate-5-phosphatase of 145 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110450 225.00
Check your price
  • SHIP1 Monoclonal Antibody for Western Blot, Flow and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514896
  • Size 100 µL
Applications
Flow Cytometry (1:200), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
p150Ship; SH2 domain-containing inositol 5'-phosphatase 1; Inositol polyphosphate-5-phosphatase of 145 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514896 275.00
Check your price
  • SHIP1 Monoclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514893
  • Size 100 µL
Applications
Flow Cytometry (1:200), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
p150Ship; SH2 domain-containing inositol 5'-phosphatase 1; Inositol polyphosphate-5-phosphatase of 145 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514893 275.00
Check your price
  • SHIP1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512463
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
p150Ship; SH2 domain-containing inositol 5'-phosphatase 1; Inositol polyphosphate-5-phosphatase of 145 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512463 349.00
Check your price
  • SHIP1 Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517390
  • Size 100 µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:25), Immunofluorescence (1:25), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
p150Ship; SH2 domain-containing inositol 5'-phosphatase 1; Inositol polyphosphate-5-phosphatase of 145 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517390 275.00
Check your price
  • SHIP1 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517392
  • Size 100 µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
SH2 domain-containing inositol 5'-phosphatase 1; p150Ship; Inositol polyphosphate-5-phosphatase of 145 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517392 275.00
Check your price
  • Phospho-SHIP1 (Tyr1021) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538393
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
p150Ship; SH2 domain-containing inositol 5'-phosphatase 1; Inositol polyphosphate-5-phosphatase of 145 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538393 309.00
Check your price
  • SHIP1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532582
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
p150Ship; SH2 domain-containing inositol 5'-phosphatase 1; Inositol polyphosphate-5-phosphatase of 145 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532582 349.00
Check your price
  • Phospho-SHIP1 (Tyr1021) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA540220
  • Size 100 µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
p150Ship; SH2 domain-containing inositol 5'-phosphatase 1; Inositol polyphosphate-5-phosphatase of 145 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540220 309.00
Check your price