1 - 3 of 3 products displayed
  • MCP-5 Antibody (Monoclonal, 63205)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523841
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.5-2.5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
MCP-5, Scya12
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523841 299.00
Check your price
  • MCP-5 Antibody, Biotin
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # PA132934
  • Size 50 µg
Applications
ELISA (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132934 600.00
Check your price
  • MCP-5 Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA132933
  • Size 100 µg
Applications
ELISA (Assay-dependent), Functional Assay (Assay-dependent), Neutralization (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132933 600.00
Check your price