1 - 30 of 30 products displayed
  • CD3z Antibody (Monoclonal, 6B10)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 137700
  • Size 100 µg
Applications
ELISA (0.1-1.0 ug/ml), Immunoprecipitation (5 ug), Western Blot (1-3 ug/ml)
Alternate names:
T-cell receptor T3 zeta chain; T-cell receptor T3 eta chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
137700 309.00
Check your price
  • TCR alpha/beta Antibody, FITC (Monoclonal, R73)
  • Host Mouse
  • Target Species Non-human primate, Rat
  • Conjugate FITC
  • Catalog # A18461
  • Size 50 µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Tcra, Tcrb, FLJ22602, MGC117436, MGC22624, MGC23964, MGC71411, TCRA, TCRD, TRA, T-cell antigen receptor, alpha polypeptide, T-cell receptor, TCR beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18461 106.00
Check your price
  • TCR gamma/delta Antibody, Alexa Fluor® 488 (Monoclonal, UC7-13D5)
  • Host Hamster
  • Target Species Mouse
  • Conjugate Alexa Fluor® 488
  • Catalog # A15407
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
T-cell receptor gamma, t-cell receptor delta, TCRd, TCRg, TCR delta gamma, TCR-CD3 complex
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15407 211.00
Check your price
  • TCR gamma/delta Antibody, FITC (Monoclonal, UC7-13D5)
  • Host Hamster
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # A15406
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
T-cell receptor gamma, t-cell receptor delta, TCRd, TCRg, TCR delta gamma, TCR-CD3 complex
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15406 259.00
Check your price
  • TCR gamma/delta Antibody, FITC (Monoclonal, 5A6.E9)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC See Additional Formats PE TRI-COLOR
  • Catalog # MHGD014
  • Size 2 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
T-cell receptor gamma, t-cell receptor delta, TCRd, TCRg, TCR delta gamma, TCR-CD3 complex
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHGD014 876.00
Check your price
  • TCR gamma/delta Antibody, TRI-COLOR® (Monoclonal, 5A6.E9)
  • Host Mouse
  • Target Species Human
  • Conjugate TRI-COLOR® See Additional Formats FITC PE
  • Catalog # MHGD06
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
T-cell receptor gamma, t-cell receptor delta, TCRd, TCRg, TCR delta gamma, TCR-CD3 complex
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHGD06 394.00
Check your price
  • TCR gamma/delta Antibody, FITC (Monoclonal, 5A6.E9)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC See Additional Formats PE TRI-COLOR
  • Catalog # MHGD01
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
T-cell receptor gamma, t-cell receptor delta, TCRd, TCRg, TCR delta gamma, TCR-CD3 complex
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHGD01 257.00
Check your price
  • TCR gamma/delta Antibody, PE (Monoclonal, 5A6.E9)
  • Host Mouse
  • Target Species Human
  • Conjugate PE See Additional Formats FITC TRI-COLOR
  • Catalog # MHGD04
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
T-cell receptor gamma, t-cell receptor delta, TCRd, TCRg, TCR delta gamma, TCR-CD3 complex
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHGD04 428.00
Check your price
  • Progesterone Receptor Antibody (Monoclonal, alpha PR-22)
  • Host Mouse
  • Target Species Chicken, Human, Reptile
  • Conjugate Unconjugated
  • Catalog # MA1412
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Gel Shift (Assay Dependent), Immunofluorescence (10 µg/ml), Immunoprecipitation (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1412 349.00
Check your price
  • Transferrin Receptor Antibody (Monoclonal, OX-26)
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA170033
  • Size 250 ug
Applications
Flow Cytometry (1 ug/1 million cells), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
CD71, T9, TFR, TFR1, TR, TRFR, p90, transferrin receptor protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA170033 199.00
Check your price
  • Phospho-Progesterone Receptor (Ser294) Antibody (Monoclonal, 608)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA1414
  • Size 200 µg
Applications
ELISA (Assay dependent), Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1:20), Immunofluorescence (1:20), Immunohistochemistry (Paraffin) (1:500), Immunoprecipitation (Assay dependent), Western Blot (10 ug/ml)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1414 275.00
Check your price
  • CD3/HLA Class 2 Antibody, FITC, PE (Cocktail, UCHT1/WR18)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC , PE
  • Catalog # MA516958
  • Size 50 TESTS
Applications
Flow Cytometry (Neat)
Alternate names:
MHC class II antigen DPB1; HLA class II histocompatibility antigen, DP(W4) beta chain; MHC class II antigen DRB1*15; DW2.2/DR2.2; T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain; MHC class II antigen DRB1*16; T-cell surface antigen T3/Leu-4 epsilon chain; MHC class II antigen DQB1; MHC class II antigen DRB1*3; Clone P2-beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516958 399.00
Check your price
  • TGF beta Receptor I Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate
  • Catalog # PA527785
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Transforming growth factor-beta receptor type I; TGF-beta type I receptor; Serine/threonine-protein kinase receptor R4; Activin receptor-like kinase 5; Activin A receptor type II-like protein kinase of 53kD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527785 349.00
Check your price
  • Adiponectin Receptor 2 Antibody
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA11071
  • Size 100 µg
Applications
Western Blot (1:50-1:500)
Alternate names:
ADIPOR2, ACDCR2, Adiponectin receptor protein 2, FLJ21432, MGC4640, PAQR2, Progestin and adipoQ receptor family member II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11071 349.00
Check your price
  • CD3/CD28 Antibody, FITC, PE (Cocktail, M2AB, B-23)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC , PE
  • Catalog # MA112098
  • Size 50 Tests
Applications
Flow Cytometry (10 ul/10^6 cells)
Alternate names:
T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain; TP44; T-cell surface antigen T3/Leu-4 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112098 309.00
Check your price
  • Estrogen Receptor alpha Antibody, Biotin (Monoclonal, AER314)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA512325
  • Size 500 µL
Applications
Affinity Purification (Assay Dependent), Gel Shift (1mg/mL), Immunocytochemistry (1:500-1:2000), Immunofluorescence (1:500-1:2000), Immunoprecipitation (2 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512325 387.00
Check your price
  • Estrogen Receptor alpha Antibody (Cocktail, 1D5 + 6F11)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA514104
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:100-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514104 550.00
Check your price
  • Estrogen Receptor alpha Antibody (Monoclonal, AER314)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA512328
  • Size 500 µL
Applications
Gel Shift (1mg/mL), Immunoprecipitation (2 µg/ml), Western Blot (1:100-1:400)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512328 309.00
Check your price
  • Estrogen Receptor alpha Antibody (Monoclonal, SP1)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514501
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:100-1:400), Western Blot (1:200-1:500)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514501 650.00
Check your price
  • Estrogen Receptor alpha Antibody (Monoclonal, 1D5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513191
  • Size 500 µL
Applications
Immunocytochemistry (1:500-1:1000), Immunofluorescence (1:500-1:1000), Immunohistochemistry (Paraffin) (0.4-0.8 µg/mL), Western Blot (0.5-1.0 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513191 550.00
Check your price
  • Estrogen Receptor alpha Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Sheep, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA516476
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (10-20 µg/mL), Immunoprecipitation (10 µg/ml), Western Blot (2.5-5.0 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516476 349.00
Check your price
  • Estrogen Receptor alpha Antibody
  • Host Rabbit
  • Target Species Hamster, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA516440
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:250-500), Immunoprecipitation (10 µg/ml), Western Blot (2-5µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516440 349.00
Check your price
  • Transferrin Receptor Antibody, Biotin (Monoclonal, OX-26)
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # MA517556
  • Size 100 ug
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
CD71, T9, TFR, TFR1, TR, TRFR, p90, transferrin receptor protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517556 159.00
Check your price
  • Transferrin Receptor Antibody, Biotin (Monoclonal, OX-26)
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # MA517557
  • Size 500 ug
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
CD71, T9, TFR, TFR1, TR, TRFR, p90, transferrin receptor protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517557 309.00
Check your price
  • Transferrin Receptor Antibody, FITC (Monoclonal, OX-26)
  • Host Mouse
  • Target Species Rat
  • Conjugate FITC
  • Catalog # MA517558
  • Size 100 ug
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
CD71, T9, TFR, TFR1, TR, TRFR, p90, transferrin receptor protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517558 159.00
Check your price
  • Transferrin Receptor Antibody, FITC (Monoclonal, OX-26)
  • Host Mouse
  • Target Species Rat
  • Conjugate FITC
  • Catalog # MA517559
  • Size 500 ug
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
CD71, T9, TFR, TFR1, TR, TRFR, p90, transferrin receptor protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517559 475.00
Check your price
  • Transferrin Receptor Antibody (Monoclonal, OX-26)
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA517555
  • Size 500 ug
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
CD71, T9, TFR, TFR1, TR, TRFR, p90, transferrin receptor protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517555 309.00
Check your price
  • Progesterone Receptor Antibody (Monoclonal, SP2)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514505
  • Size 500 µL
Applications
Immunofluorescence (1-2 µg/ml), Immunohistochemistry (Paraffin) (1:100-1:400), Western Blot (1:1000-1:2000)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514505 700.00
Check your price
  • Progesterone Receptor Antibody Polyclonal (PA5-16709)
  • Host Rabbit
  • Target Species Human
  • Conjugate
  • Catalog # PA516709
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516709 339.00
Check your price
  • Estrogen Receptor beta Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 517700
  • Size 100 µg
Applications
ELISA (1.0 ug/ml), Flow Cytometry (3-5 µg/million cells), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (1-2 ug/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
517700 305.00
Check your price