1 - 12 of 12 products displayed
  • Phospho-TERT (Ser227) Polyclonal Antibody for IF, ICC and pep-ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538817
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), peptide-ELISA (1:1000)
Alternate names:
Telomerase-associated protein 2; Telomerase catalytic subunit; HEST2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538817 309.00
Check your price
  • TERT Polyclonal Antibody for Western Blot, IF, ICC, IHC, Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate
  • Catalog # PA511447
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Telomerase-associated protein 2; Telomerase catalytic subunit; HEST2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511447 339.00
Check your price
  • Phospho-TERT (Ser1125) Polyclonal Antibody for DB
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512973
  • Size 400 µL
Applications
Dot blot (1:500)
Alternate names:
Telomerase-associated protein 2; Telomerase catalytic subunit; HEST2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512973 349.00
Check your price
  • Phospho-TERT (Ser824) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538820
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Telomerase-associated protein 2; Telomerase catalytic subunit; HEST2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538820 309.00
Check your price
  • Phospho-TERT (Tyr707) Polyclonal Antibody for IF, ICC and DB
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513048
  • Size 400 µL
Applications
Dot blot (1:500), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50)
Alternate names:
Telomerase-associated protein 2; Telomerase catalytic subunit; HEST2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513048 349.00
Check your price
  • TERT Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532195
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Telomerase-associated protein 2; Telomerase catalytic subunit; HEST2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532195 349.00
Check your price
  • TERT Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511446
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
Telomerase-associated protein 2; Telomerase catalytic subunit; HEST2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511446 349.00
Check your price
  • TERT Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516033
  • Size 100 µL
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Assay-Dependent), Western Blot (1:500-1:1000)
Alternate names:
Telomerase-associated protein 2; Telomerase catalytic subunit; HEST2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516033 349.00
Check your price
  • TERT Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA516034
  • Size 100 µL
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50), Immunoprecipitation (1:10-1:500), Western Blot (1:500)
Alternate names:
Telomerase catalytic subunit; Telomerase-associated protein 2; HEST2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516034 349.00
Check your price
  • TEP1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520354
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
p80 telomerase homolog; p240; Telomerase-associated protein 1; p230
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520354 349.00
Check your price
  • SCYL1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529295
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Teratoma-associated tyrosine kinase; Telomerase transcriptional element-interacting factor; Telomerase regulation-associated protein; SCY1-like protein 1; Coated vesicle-associated kinase of 90 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529295 349.00
Check your price
  • TAK1 Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 700113
  • Size 100 µg
Applications
Flow Cytometry (3-5 ug x 10^6 cells), Immunocytochemistry (2-3 ug/ml), Immunofluorescence (2-3 ug/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
Transforming growth factor-beta-activated kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
700113 303.00
Check your price