1 - 3 of 3 products displayed
  • BERP Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA540957
  • Size 100 µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (1 µg/ml)
Alternate names:
RING finger protein 22; Brain-expressed RING finger protein; RING finger protein HAC1; RING finger protein 97
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540957 309.00
Check your price
  • BERP Antibody
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518136
  • Size 100 ug
Applications
Western Blot (0.1-1 ug/ml)
Alternate names:
RING finger protein HAC1; RING finger protein 22; RING finger protein 97; Brain-expressed RING finger protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518136 349.00
Check your price
  • BERP Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538542
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
RING finger protein 22; Brain-expressed RING finger protein; RING finger protein HAC1; RING finger protein 97
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538542 309.00
Check your price