1 - 30 of 30 products displayed
  • Collagen IV Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Mammal
  • Conjugate Unconjugated
  • Catalog # PA128534
  • Size 100 µg
Applications
ELISA (1:4000-1:8000), Immunohistochemistry (Frozen) (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (1:5000-1:10,000), Western Blot (1:5000-1:10,000)
Alternate names:
arresten
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128534 375.00
Check your price
  • Collagen III (soluble) Antibody (Monoclonal, 1E7-D7/Col3)
  • Host Mouse
  • Target Species Dog, Human, Marsupial, Rat
  • Conjugate Unconjugated
  • Catalog # CSI0070102
  • Size 200 µl
Applications
ELISA (1:800), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay dependent)
Alternate names:
COL3, Collagen type III alpha 1, EDS4A
Catalog # Size Price (AUD) Your Price (AUD) Qty
CSI0070102 425.00
Check your price
  • Collagen II Antibody (Monoclonal, 5B2.5)
  • Host Mouse
  • Target Species Chicken, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA121807
  • Size 500µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Western Blot (1-2 µg/ml)
Alternate names:
Alpha-1 type II collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121807 375.00
Check your price
  • Collagen I Antibody, Biotin
  • Host Goat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # PA186054
  • Size 200 µg
Applications
ELISA (1:200-1:2000)
Alternate names:
Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186054 800.00
Check your price
  • Collagen IV Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA136063
  • Size 1 mL
Applications
ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
arresten
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136063 309.00
Check your price
  • Collagen IV Antibody (Monoclonal, MC4-HA)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135333
  • Size 200 µg
Applications
ELISA (Assay dependent.), Immunohistochemistry (Paraffin, Frozen) (1:20)
Alternate names:
arresten
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135333 425.00
Check your price
  • Collagen IV Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA136065
  • Size 1 mL
Applications
ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
arresten
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136065 349.00
Check your price
  • Collagen IV Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA136001
  • Size 100 µg
Applications
ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
arresten
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136001 349.00
Check your price
  • Collagen IV Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA126148
  • Size 100 µL
Applications
ELISA (1:200), Immunocytochemistry (1:80), Immunofluorescence (1:80), Immunohistochemistry (Paraffin) (1:500), Radioimmune Assays (Assay Dependent)
Alternate names:
Type IV Collagen, Collagen 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126148 399.00
Check your price
  • Collagen I Antibody (Monoclonal, 5D8-G9)
  • Host Mouse
  • Target Species Bovine, Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA1141
  • Size 100 µg
Applications
ELISA (0.4-50 µg/ml), Immunohistochemistry (Frozen) (1:200-1:1000), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (1:500-1:2000)
Alternate names:
Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1141 349.00
Check your price
  • Collagen I Antibody, Biotin
  • Host Rabbit
  • Target Species Bovine, Human
  • Conjugate Biotin
  • Catalog # PA128530
  • Size 50 µg
Applications
ELISA (1:4000-1:8000), Immunohistochemistry (Frozen) (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Immunomicroscopy (Assay Dependent), Immunoprecipitation (1:5000-1:10,000), Western Blot (1:5000-1:10,000)
Alternate names:
Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128530 349.00
Check your price
  • Collagen III Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534787
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
EDS4A, Ehlers-Danlos syndrome type IV, autosomal dominant, alpha1 (III) collagen, collagen alpha-1(III) chain, COL3 alpha 1, Collagen 3, Collagen3, COL 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534787 349.00
Check your price
  • Collagen Pan Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA185324
  • Size 50 µg
Applications
ELISA (1:2000-1:8000), Immunofluorescence (1:10-1:40), Immunohistochemistry (Frozen) (1:10-1:40)
Alternate names:
Alpha-1 type II collagen; Alpha-2 type I collagen; Goodpasture antigen; Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185324 449.00
Check your price
  • Collagen IV Antibody (Monoclonal, COL-94)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA122148
  • Size 100 µL
Applications
Dot blot (Assay dependent), ELISA (Assay dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (1:500), Immunohistochemistry (Paraffin) (1:500), Western Blot (Assay dependent)
Alternate names:
arresten
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122148 375.00
Check your price
  • Collagen Pan Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA136058
  • Size 1 mL
Applications
ELISA (Assay dependent), Immunohistochemistry (Frozen) (1:200), Western Blot (Assay dependent)
Alternate names:
Alpha-1 type II collagen; Alpha-2 type I collagen; Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136058 375.00
Check your price
  • Collagen I Antibody (Monoclonal, NFI/20)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA183073
  • Size 100 µg
Applications
ELISA (1:1000), Immunohistochemistry (Frozen) (1:20)
Alternate names:
Alpha-2 type I collagen; Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183073 349.00
Check your price
  • Collagen III Antibody
  • Host Rabbit
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # PA128870
  • Size 100 µg
Applications
ELISA (1:5000-1:10,000), Immunohistochemistry (Frozen) (1:50-1:200), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunoprecipitation (Assay dependent), Western Blot (1:5000-1:10,000)
Alternate names:
COL3, Collagen type III alpha 1, EDS4A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128870 309.00
Check your price
  • Collagen III Antibody (Monoclonal, NLI/42)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516891
  • Size 1 mL
Applications
ELISA (1:500-1:2000), Immunohistochemistry (Frozen) (1:20-1:80)
Alternate names:
EDS4A,Ehlers-Danlos syndrome type IV, autosomal dominant; alpha1 (III) collagen; collagen alpha-1(III) chain; collagen, fetal
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516891 309.00
Check your price
  • Collagen I/III Antibody
  • Host Rabbit
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # PA135035
  • Size 100 µg
Applications
ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
Alpha-1 type I collagen; Alpha-2 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA135035 399.00
Check your price
  • Collagen I/III Antibody
  • Host Rabbit
  • Target Species Dog
  • Conjugate Unconjugated
  • Catalog # PA136069
  • Size 100 µg
Applications
ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
Alpha-1 type I collagen; Alpha-2 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136069 325.00
Check your price
  • Collagen III Antibody, Biotin
  • Host Rabbit
  • Target Species Bovine, Human, Mammal
  • Conjugate Biotin
  • Catalog # PA128532
  • Size 50 µg
Applications
ELISA (1:10,000-1:50,000), Immunohistochemistry (Frozen) (1:200-1:1000), Immunohistochemistry (Paraffin) (1:200-1:1000), Immunoprecipitation (Assay dependent), Western Blot (1:5000-1:10,000)
Alternate names:
EDS4A, FLJ34534
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128532 349.00
Check your price
  • Collagen III Antibody (Monoclonal, FH-7A)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA122147
  • Size 100 µL
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (1:4000), Immunohistochemistry (Paraffin) (Assay-dependent), Western Blot (1:100-1:1000)
Alternate names:
EDS4A, FLJ34534
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122147 425.00
Check your price
  • Collagen I/III Antibody
  • Host Rabbit
  • Target Species Pig
  • Conjugate Unconjugated
  • Catalog # PA185328
  • Size 500 µL
Applications
ELISA (1:500-1:2000), Immunofluorescence (1:10-1:40), Immunohistochemistry (Frozen) (1:10-1:40)
Alternate names:
COL1A1, COL1A2, COL3A1, Collagen 1, Collagen 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185328 425.00
Check your price
  • Collagen I/III Antibody
  • Host Rabbit
  • Target Species Pig
  • Conjugate Unconjugated
  • Catalog # PA136140
  • Size 100 µg
Applications
Dot blot (Assay dependent), ELISA (Assay dependent.), Immunohistochemistry (Frozen) (1:20)
Alternate names:
COL1A1, COL1A2, COL3A1, Collagen 1, Collagen 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136140 349.00
Check your price
  • Collagen III Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA185314
  • Size 500 µL
Applications
ELISA (1/2000-1/8000), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1/10-1/40)
Alternate names:
EDS4A, FLJ34534
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185314 550.00
Check your price
  • Collagen III Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA136062
  • Size 1 mL
Applications
ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
EDS4A, FLJ34534
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136062 375.00
Check your price
  • Collagen III Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA136000
  • Size 100 µg
Applications
ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
EDS4A, FLJ34534
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136000 349.00
Check your price
  • Collagen III Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA136061
  • Size 1 mL
Applications
ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
EDS4A, FLJ34534
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136061 309.00
Check your price
  • Collagen IV Antibody
  • Host Rabbit
  • Target Species Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA185320
  • Size 100 µL
Applications
ELISA (1:100-1:400), Immunofluorescence (1:40-1:160), Immunohistochemistry (Paraffin) (1:250-1:1000)
Alternate names:
arresten
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185320 425.00
Check your price
  • MMP9 Antibody
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA122528
  • Size 50 µg
Applications
Immunoprecipitation (1:500), Western Blot (1:2000)
Alternate names:
Gelatinase B; 92 kDa type IV collagenase; 92 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA122528 349.00
Check your price