1 - 24 of 24 products displayed
  • Phospho-TEK (Tyr1113) Polyclonal Antibody for DB
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512992
  • Size 400µL
Applications
Dot blot (1:500)
Alternate names:
p140 TEK; Tyrosine-protein kinase receptor TIE-2; Tyrosine-protein kinase receptor TEK; Tyrosine kinase with Ig and EGF homology domains-2; Tunica interna endothelial cell kinase; Endothelial tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512992 349.00
Check your price
  • TEK Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528582
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
p140 TEK; Tyrosine-protein kinase receptor TIE-2; Tyrosine-protein kinase receptor TEK; Tyrosine kinase with Ig and EGF homology domains-2; Tunica interna endothelial cell kinase; Endothelial tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528582 349.00
Check your price
  • TEK Polyclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523082
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:200), Western Blot (1:2500)
Alternate names:
p140 TEK; Tyrosine-protein kinase receptor TIE-2; Tyrosine-protein kinase receptor TEK; Tyrosine kinase with Ig and EGF homology domains-2; Tunica interna endothelial cell kinase; Endothelial tyrosine kinase; STK1; HYK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523082 349.00
Check your price
  • TEK Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514742
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
p140 TEK; Tyrosine-protein kinase receptor TIE-2; Tyrosine-protein kinase receptor TEK; Tyrosine kinase with Ig and EGF homology domains-2; Tunica interna endothelial cell kinase; Endothelial tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514742 349.00
Check your price
  • TEK Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 425100
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
p140 TEK; Tyrosine-protein kinase receptor TIE-2; Tyrosine-protein kinase receptor TEK; Tyrosine kinase with Ig and EGF homology domains-2; Tunica interna endothelial cell kinase; STK1; HYK; Endothelial tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
425100 309.00
Check your price
  • Phospho-TEK (Tyr992) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537754
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
p140 TEK; Tyrosine-protein kinase receptor TIE-2; Tyrosine-protein kinase receptor TEK; Tyrosine kinase with Ig and EGF homology domains-2; Tunica interna endothelial cell kinase; Endothelial tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537754 309.00
Check your price
  • Phospho-TEK (Tyr1108) Polyclonal Antibody for Western Blot and IF
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538339
  • Size 100µg
Applications
Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
p140 TEK; Tyrosine-protein kinase receptor TIE-2; Tyrosine-protein kinase receptor TEK; Tyrosine kinase with Ig and EGF homology domains-2; Tunica interna endothelial cell kinase; Endothelial tyrosine kinase; STK1; HYK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538339 309.00
Check your price
  • Phospho-TEK (Tyr1102) Polyclonal Antibody for IHC (P) and pep-ELISA
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538394
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), peptide-ELISA (1:40,000)
Alternate names:
p140 TEK; Tyrosine-protein kinase receptor TIE-2; Tyrosine-protein kinase receptor TEK; Tyrosine kinase with Ig and EGF homology domains-2; Tunica interna endothelial cell kinase; Endothelial tyrosine kinase; STK1; HYK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538394 309.00
Check your price
  • TEK Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # A18380
  • Size 50µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
p140 TEK; Tyrosine-protein kinase receptor TIE-2; Tyrosine-protein kinase receptor TEK; Tyrosine kinase with Ig and EGF homology domains-2; Tunica interna endothelial cell kinase; STK1; HYK; Endothelial tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18380 168.00
Check your price
  • TIE1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532618
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
JTK14, TIE, tyrosine kinase with immunoglobulin and epidermal growth factor homology domains 1, tyrosine-protein kinase receptor Tie-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532618 349.00
Check your price
  • TIE2 Polyclonal Antibody for Western Blot, IHC (F) and Neu
  • Host Goat
  • Target Species Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA547963
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Tyrosine kinase with Ig and EGF homology domains-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547963 349.00
Check your price
  • SHC Monoclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFMA0201
  • Size 100µL
Applications
ELISA (Assay Dependent), Immunoprecipitation (2 ug), Western Blot (0.1 ug/ml)
Alternate names:
Src homology 2 domain-containing-transforming protein C1; SHC-transforming protein A; SHC-transforming protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0201 309.00
Check your price
  • Phospho-SHC (Tyr239, Tyr240) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44830
  • Size 10blots
Applications
Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (0.5-2 µg/ml)
Alternate names:
Src homology 2 domain-containing-transforming protein C1; SHC-transforming protein A; SHC-transforming protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
44830 309.00
Check your price
  • Phospho-SHC (Tyr427) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537738
  • Size 100µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Src homology 2 domain-containing-transforming protein C1; SHC-transforming protein A; SHC-transforming protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537738 309.00
Check your price
  • Phospho-SHC (Tyr349) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA537737
  • Size 100µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Western Blot (1:500-1:1000)
Alternate names:
Src homology 2 domain-containing-transforming protein C1; SHC-transforming protein A; SHC-transforming protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537737 309.00
Check your price
  • Phospho-SHC (Ser36) Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Dog, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44828M
  • Size 50µL
Applications
Western Blot (1µg/ml)
Alternate names:
Src homology 2 domain-containing-transforming protein C1; SHC-transforming protein A; SHC-transforming protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
44828M 304.00
Check your price
  • SHC Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44829M
  • Size 50µL
Applications
Western Blot (0.5µg/ml)
Alternate names:
Src homology 2 domain-containing-transforming protein C1; SHC-transforming protein A; SHC-transforming protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
44829M 309.00
Check your price
  • SHC Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534647
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Src homology 2 domain-containing-transforming protein C1; SHC-transforming protein A; SHC-transforming protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534647 349.00
Check your price
  • Angiopoietin 2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527297
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
AGPT2, ANG2, ANG-2, Tie2-ligand, angiopoietin-2, angiopoietin-2B, angiopoietin-2a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527297 349.00
Check your price
  • Angiopoietin 4 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534646
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Angiopoietin-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534646 349.00
Check your price
  • Angiopoietin 2 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523612
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
AGPT2, ANG2, ANG-2, Tie2-ligand, angiopoietin-2, angiopoietin-2B, angiopoietin-2a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523612 349.00
Check your price
  • Angiopoietin 2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA129640
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (Assay-dependent), Western Blot (1:500)
Alternate names:
AGPT2, ANG2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129640 375.00
Check your price
  • Angiopoietin 2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527299
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
AGPT2, ANG2, ANG-2, Tie2-ligand, angiopoietin-2, angiopoietin-2B, angiopoietin-2a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527299 349.00
Check your price
  • Angiopoietin 4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534780
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Angiopoietin-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534780 349.00
Check your price