1 - 17 of 17 products displayed
  • TYRO3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA514737
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Tyrosine-protein kinase TIF; Tyrosine-protein kinase SKY; Tyrosine-protein kinase RSE; Tyrosine-protein kinase DTK; Tyrosine-protein kinase BYK; TK19-2; Etk2/tyro3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514737 349.00
Check your price
  • TYRO3 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515340
  • Size 100µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Western Blot (1/500-1/2000)
Alternate names:
Tyrosine-protein kinase TIF; Tyrosine-protein kinase SKY; Tyrosine-protein kinase RSE; Tyrosine-protein kinase DTK; Tyrosine-protein kinase BYK
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515340 349.00
Check your price
  • TYRO3 Monoclonal Antibody for IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515404
  • Size 100µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000)
Alternate names:
Tyrosine-protein kinase TIF; Tyrosine-protein kinase SKY; Tyrosine-protein kinase RSE; Tyrosine-protein kinase DTK; Tyrosine-protein kinase BYK
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515404 349.00
Check your price
  • TYRO3 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515505
  • Size 100µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Tyrosine-protein kinase TIF; Tyrosine-protein kinase SKY; Tyrosine-protein kinase RSE; Tyrosine-protein kinase DTK; Tyrosine-protein kinase BYK
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515505 349.00
Check your price
  • TYRO3 Monoclonal Antibody for IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511171
  • Size 400µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100)
Alternate names:
Tyrosine-protein kinase TIF; Tyrosine-protein kinase SKY; Tyrosine-protein kinase RSE; Tyrosine-protein kinase DTK; Tyrosine-protein kinase BYK
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511171 349.00
Check your price
  • TYRO3 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514738
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Tyrosine-protein kinase TIF; Tyrosine-protein kinase SKY; Tyrosine-protein kinase RSE; Tyrosine-protein kinase DTK; Tyrosine-protein kinase BYK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514738 349.00
Check your price
  • TYRO3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528682
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Tyrosine-protein kinase TIF; Tyrosine-protein kinase SKY; Tyrosine-protein kinase RSE; Tyrosine-protein kinase DTK; Tyrosine-protein kinase BYK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528682 349.00
Check your price
  • TYRO3 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA522999
  • Size 100µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Western Blot (2 µg/ml)
Alternate names:
Tyrosine-protein kinase TIF; Tyrosine-protein kinase SKY; Tyrosine-protein kinase RSE; Tyrosine-protein kinase DTK; Tyrosine-protein kinase BYK; TK19-2; Etk2/tyro3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522999 349.00
Check your price
  • TYRO3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA523244
  • Size 100ug
Applications
Western Blot (0.25-1 µg/ml)
Alternate names:
Tyrosine-protein kinase SKY; Tyrosine-protein kinase TIF; Tyrosine-protein kinase RSE; Tyrosine-protein kinase DTK; Tyrosine-protein kinase BYK; TK19-2; Etk2/tyro3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523244 349.00
Check your price
  • Phospho-MER/SKY (Tyr749, Tyr681) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA540270
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Receptor tyrosine kinase MerTK; Proto-oncogene c-Mer; Tyrosine-protein kinase TIF; Tyrosine-protein kinase SKY; Tyrosine-protein kinase RSE; Tyrosine-protein kinase DTK; Tyrosine-protein kinase BYK; TK19-2; Etk2/tyro3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540270 309.00
Check your price
  • Phospho-MER/SKY (Tyr749, Tyr681) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537808
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Tyrosine-protein kinase SKY; Receptor tyrosine kinase MerTK; Proto-oncogene c-Mer; Tyrosine-protein kinase TIF; Tyrosine-protein kinase RSE; Tyrosine-protein kinase DTK; Tyrosine-protein kinase BYK; TK19-2; Etk2/tyro3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537808 309.00
Check your price
  • MERTK Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Human, Non-human primate
  • Conjugate
  • Catalog # PA523280
  • Size 100ug
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Receptor tyrosine kinase MerTK; Proto-oncogene c-Mer
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523280 Not Orderable Online
Check your price
  • MERTK Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515028
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:10), Western Blot (1:1000)
Alternate names:
Receptor tyrosine kinase MerTK; Proto-oncogene c-Mer
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515028 349.00
Check your price
  • MERTK Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518028
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Receptor tyrosine kinase MerTK; Proto-oncogene c-Mer
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518028 425.00
Check your price
  • MERTK Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515358
  • Size 100µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Receptor tyrosine kinase MerTK; Proto-oncogene c-Mer
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515358 349.00
Check your price
  • MERTK Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131203
  • Size 100µg
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (1:200), Western Blot (1:500)
Alternate names:
Receptor tyrosine kinase MerTK; Proto-oncogene c-Mer
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131203 499.00
Check your price
  • MERTK Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131204
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
Receptor tyrosine kinase MerTK; Proto-oncogene c-Mer
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131204 449.00
Check your price