1 - 30 of 30 products displayed
  • VEGF Receptor 2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 441053G
  • Size 10 blots
Applications
Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Protein-tyrosine kinase receptor flk-1; Kinase insert domain receptor; Fetal liver kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
441053G 268.00
Check your price
  • Phospho-VEGF Receptor 2 (Tyr1175) Antibody (Monoclonal, S.657.3)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515170
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:300), Western Blot (1:1000)
Alternate names:
Protein-tyrosine kinase receptor flk-1; Kinase insert domain receptor; Fetal liver kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515170 309.00
Check your price
  • Phospho-VEGF Receptor 2 (Tyr951) Antibody (Monoclonal, H.266.3)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515149
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (1:1000)
Alternate names:
Protein-tyrosine kinase receptor flk-1; Kinase insert domain receptor; Fetal liver kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515149 309.00
Check your price
  • VEGF Receptor 2 Antibody (Monoclonal, B.309.4)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515157
  • Size 100 µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (Frozen) (1:200), Immunohistochemistry (Paraffin) (1:600), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Protein-tyrosine kinase receptor flk-1; Kinase insert domain receptor; Fetal liver kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515157 275.00
Check your price
  • VEGF Receptor 2 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA120207
  • Size 25 µg
Applications
ELISA (0.5-1 µg/ml), Immunohistochemistry (5-15 µg/ml), Neutralization (5-50 ng/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
Protein-tyrosine kinase receptor flk-1; Kinase insert domain receptor; Fetal liver kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120207 349.00
Check your price
  • VEGF Receptor 2 Antibody (Monoclonal, ZV001)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 392400
  • Size 100 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Protein-tyrosine kinase receptor flk-1; Kinase insert domain receptor; Fetal liver kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
392400 304.00
Check your price
  • VEGF Receptor 2 Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 360900
  • Size 100 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
6130401C07, Flk-1, Flk1, Krd-1, Ly73, VEGFR-2, VEGFR2, sVEGFR-2, CD309, KDR
Catalog # Size Price (AUD) Your Price (AUD) Qty
360900 304.00
Check your price
  • VEGF Receptor 1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121731
  • Size 500 µL
Applications
Immunohistochemistry (Frozen) (1:50-1:100), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Vascular permeability factor receptor; Tyrosine-protein kinase receptor FLT; Tyrosine-protein kinase FRT; Fms-like tyrosine kinase 1; Embryonic receptor kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121731 499.00
Check your price
  • VEGF Receptor 3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527277
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Tyrosine-protein kinase receptor FLT4; Fms-like tyrosine kinase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527277 349.00
Check your price
  • VEGF Receptor 1 (soluble) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 361100
  • Size 100 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Vascular permeability factor receptor; Tyrosine-protein kinase receptor FLT; Tyrosine-protein kinase FRT; Fms-like tyrosine kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
361100 304.00
Check your price
  • VEGF Receptor 2 Antibody, Biotin (Monoclonal, KDR-2 or 260.4)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA120208
  • Size 50 µL
Applications
ELISA (Assay dependent), Flow Cytometry (Assay dependent), Immunohistochemistry (Assay dependent), Western Blot (1:1000)
Alternate names:
Protein-tyrosine kinase receptor flk-1; Kinase insert domain receptor; Fetal liver kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120208 375.00
Check your price
  • VEGF Receptor 3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 360200
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (1:20), Western Blot (Assay Dependent)
Alternate names:
Tyrosine-protein kinase receptor FLT4; Fms-like tyrosine kinase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
360200 309.00
Check your price
  • VEGF Receptor 2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121025
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (Assay dependent)
Alternate names:
Protein-tyrosine kinase receptor flk-1; Kinase insert domain receptor; Fetal liver kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121025 349.00
Check your price
  • Phospho-VEGF Receptor 2 (Tyr1054) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538540
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Protein-tyrosine kinase receptor flk-1; Kinase insert domain receptor; Fetal liver kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538540 309.00
Check your price
  • Phospho-VEGF Receptor 2 (Tyr1214) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539704
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Protein-tyrosine kinase receptor flk-1; Kinase insert domain receptor; Fetal liver kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539704 309.00
Check your price
  • Phospho-VEGF Receptor 2 (Tyr951) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA536700
  • Size 100 µl
Applications
Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Protein-tyrosine kinase receptor flk-1; Kinase insert domain receptor; Fetal liver kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536700 349.00
Check your price
  • VEGF Receptor 2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116613
  • Size 100 µL
Applications
Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Western Blot (1:250-1:1000)
Alternate names:
Protein-tyrosine kinase receptor flk-1; Kinase insert domain receptor; Fetal liver kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116613 349.00
Check your price
  • VEGF Receptor 2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA526064
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Protein-tyrosine kinase receptor flk-1; Kinase insert domain receptor; Fetal liver kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526064 349.00
Check your price
  • Phospho-VEGF Receptor 2 (Tyr1054, Tyr1059) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 441047G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
Protein-tyrosine kinase receptor flk-1; Kinase insert domain receptor; Fetal liver kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
441047G 304.00
Check your price
  • Phospho-VEGF Receptor 2 (Tyr1214) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 441052
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Western Blot (0.35-1.0 ug/ml)
Alternate names:
Protein-tyrosine kinase receptor flk-1; Kinase insert domain receptor; Fetal liver kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
441052 309.00
Check your price
  • VEGF Receptor 2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA316624
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:10-1:500), Immunoprecipitation (2-5 µl), Western Blot (1:2000-1:10,000)
Alternate names:
Protein-tyrosine kinase receptor flk-1; Kinase insert domain receptor; Fetal liver kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316624 375.00
Check your price
  • VEGF Receptor 2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA516487
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (5-10 µg/mL)
Alternate names:
Protein-tyrosine kinase receptor flk-1; Kinase insert domain receptor; Fetal liver kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516487 349.00
Check your price
  • VEGF Receptor 2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA524710
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Protein-tyrosine kinase receptor flk-1; Kinase insert domain receptor; Fetal liver kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524710 349.00
Check your price
  • Phospho-VEGF Receptor 2 (Tyr1054) Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 441046
  • Size 10 blots
Applications
Western Blot (0.35-1.0 ug/ml)
Alternate names:
6130401C07, Flk-1, Flk1, Krd-1, Ly73, VEGFR-2, VEGFR2, sVEGFR-2, CD309, KDR
Catalog # Size Price (AUD) Your Price (AUD) Qty
441046 309.00
Check your price
  • VEGF Receptor 2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA132431
  • Size 100 µL
Applications
Immunoprecipitation (2-5 µl), Western Blot (1:2000-1:10,000)
Alternate names:
Protein-tyrosine kinase receptor flk-1; Kinase insert domain receptor; Fetal liver kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132431 550.00
Check your price
  • VEGF Antibody (Monoclonal, SP28)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514573
  • Size 7 mL
Applications
Immunohistochemistry (Paraffin) (Ready-to-use)
Alternate names:
Vascular permeability factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514573 309.00
Check your price
  • VEGF Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA516754
  • Size 500 µL
Applications
Immunocytochemistry (1:40-1:200), Immunofluorescence (1:40-1:200), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (3 µg/ml), Western Blot (1 µg/mL)
Alternate names:
Vascular permeability factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516754 309.00
Check your price
  • VEGF Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # P931
  • Size 500 ug
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:20-1:100), Immunofluorescence (1:20-1:100), Western Blot (1:500-1:1000)
Alternate names:
Vascular permeability factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
P931 375.00
Check your price
  • VEGF Antibody (Monoclonal, 5C3.F8)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512949
  • Size 500 µL
Applications
Immunocytochemistry (1:20-1:100), Immunofluorescence (1:20-1:100), Western Blot (1-2 µg/ml)
Alternate names:
Vascular permeability factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512949 309.00
Check your price
  • VEGF Antibody, Biotin
  • Host Rabbit
  • Target Species Human
  • Conjugate Biotin
  • Catalog # P802B
  • Size 100 µg
Applications
ELISA (0.125-0.5µg/ml), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (1 ug/ml)
Alternate names:
Vascular permeability factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
P802B 349.00
Check your price