1 - 2 of 2 products displayed
  • VIP Antibody
  • Host Guinea Pig
  • Target Species Guinea pig, Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA136030
  • Size 50 µL
Applications
Immunofluorescence (1:1000), Immunohistochemistry (Paraffin, Frozen) (1:1000-1:2000)
Alternate names:
MGC13587, PHM27
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136030 950.00
Check your price
  • VIP Antibody (Monoclonal, 2)
  • Host Mouse
  • Target Species Bovine, Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # ABS0230202
  • Size 200 µL
Applications
ELISA (1:1000), Western Blot (Assay dependent)
Alternate names:
VIP-R-1, Pituitary adenylate cyclase-activating polypeptide type II receptor_x000D_ PACAP type II receptor, PACAP-R-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0230202 550.00
Check your price