1 - 3 of 3 products displayed
  • SCN2A Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536545
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Voltage-gated sodium channel subunit alpha Nav1.2; Sodium channel protein type II subunit alpha; Sodium channel protein brain II subunit alpha; HBSC II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536545 309.00
Check your price
  • Na+ CP- Pan Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536074
  • Size 100 µl
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
SCN2A, NAC2, SCN2A1, SCN2A2, Nav1.1, NAC1, SCN1, SCN1A, Nav1.2, SCN3A, KIAA1356, NAC3, Nav1.3, Nav1.4, SCN4A, Nav1.5, SCN5A, Nav1.6, SCN8A, MED, Nav1.7, SCN9A, NENA, Nav1.8, SCN10A, Nav1.9, hNaN, SCN11A, SCN12A, SNS2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536074 309.00
Check your price
  • SCN4A Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539462
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Voltage-gated sodium channel subunit alpha Nav1.4; Sodium channel protein type IV subunit alpha; Sodium channel protein skeletal muscle subunit alpha; SkM1; Mu-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539462 309.00
Check your price