1 - 4 of 4 products displayed
  • SCN5A Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534190
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (4 µg/ml)
Alternate names:
Voltage-gated sodium channel subunit alpha Nav1.5; Sodium channel protein type V subunit alpha; Sodium channel protein cardiac muscle subunit alpha; HH1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534190 349.00
Check your price
  • SCN5A Antibody (Monoclonal, 4G8)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA127429
  • Size 100 µg
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (2 µg/ml)
Alternate names:
Voltage-gated sodium channel subunit alpha Nav1.5; Sodium channel protein type V subunit alpha; Sodium channel protein cardiac muscle subunit alpha; HH1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA127429 375.00
Check your price
  • Na+ CP- Pan Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536074
  • Size 100 µl
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
SCN2A, NAC2, SCN2A1, SCN2A2, Nav1.1, NAC1, SCN1, SCN1A, Nav1.2, SCN3A, KIAA1356, NAC3, Nav1.3, Nav1.4, SCN4A, Nav1.5, SCN5A, Nav1.6, SCN8A, MED, Nav1.7, SCN9A, NENA, Nav1.8, SCN10A, Nav1.9, hNaN, SCN11A, SCN12A, SNS2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536074 309.00
Check your price
  • SCN4A Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539462
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Voltage-gated sodium channel subunit alpha Nav1.4; Sodium channel protein type IV subunit alpha; Sodium channel protein skeletal muscle subunit alpha; SkM1; Mu-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539462 309.00
Check your price