1 - 2 of 2 products displayed
  • Vomeronasal 1 Receptor B2/B4 Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # OSR00235W
  • Size 100 µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Vomeronasal type-1 receptor B2; Vomeronasal receptor VRi2; Vomeronasal type-1 receptor B4; Vomeronasal type-1 receptor B11; Vomeronasal type-1 receptor A10
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00235W 309.00
Check your price
  • Vomeronasal 1 Receptor B2/B4 Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # OSR00236W
  • Size 100 µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Vomeronasal type-1 receptor B2; Vomeronasal receptor VRi2; Vomeronasal type-1 receptor B4; Vomeronasal type-1 receptor B11; Vomeronasal type-1 receptor A10
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00236W 309.00
Check your price